Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159587734-159588235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCS
Entrez Gene ID
325
Gene Name
amyloid P component, serum
Gene Aliases
HEL-S-92n, PTX2, SAP
UniGene
Hs.507080
Gene Chromosome Location
Chr.1: 159587826-159588871 on Build GRCh38
Transcripts
NM_001639.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602117 c.44T>A p.L15Q Chr.1: 159587965-159587965 on Build GRCh38
6602116 c.11C>T p.P4L Chr.1: 159587932-159587932 on Build GRCh38
1601087 c.35C>A p.T12N Chr.1: 159587956-159587956 on Build GRCh38
5438586 c.131C>T p.P44L Chr.1: 159588167-159588167 on Build GRCh38
675954 c.169C>A p.R57R Chr.1: 159588205-159588205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781175160 Chr.1: 159588059-159588059 on Build GRCh38
rs200052053 Chr.1: 159587779-159587779 on Build GRCh38
rs994697399 Chr.1: 159587966-159587966 on Build GRCh38
rs574058350 Chr.1: 159587770-159587770 on Build GRCh38
rs899066950 Chr.1: 159587965-159587965 on Build GRCh38
rs368966907 Chr.1: 159587874-159587874 on Build GRCh38
rs200588785 Chr.1: 159588034-159588034 on Build GRCh38
rs371650621 Chr.1: 159588168-159588168 on Build GRCh38
rs746825256 Chr.1: 159587960-159587960 on Build GRCh38
rs200185691 Chr.1: 159587824-159587824 on Build GRCh38
rs778808269 Chr.1: 159588151-159588152 on Build GRCh38
rs765738916 Chr.1: 159588107-159588107 on Build GRCh38
rs199652311 Chr.1: 159587929-159587929 on Build GRCh38
rs368672294 Chr.1: 159587852-159587853 on Build GRCh38
rs145110984 Chr.1: 159588169-159588169 on Build GRCh38
rs775940749 Chr.1: 159587897-159587897 on Build GRCh38
rs748544651 Chr.1: 159588200-159588200 on Build GRCh38
rs11545902 Chr.1: 159588130-159588130 on Build GRCh38
rs777880716 Chr.1: 159588163-159588163 on Build GRCh38
rs764444754 Chr.1: 159587944-159587949 on Build GRCh38
rs11545901 Chr.1: 159587918-159587918 on Build GRCh38
rs201292757 Chr.1: 159587828-159587828 on Build GRCh38
rs745529059 Chr.1: 159588177-159588177 on Build GRCh38
rs200993550 Chr.1: 159588083-159588083 on Build GRCh38
rs777039478 Chr.1: 159588097-159588097 on Build GRCh38
rs949575893 Chr.1: 159588213-159588213 on Build GRCh38
rs771860703 Chr.1: 159587853-159587853 on Build GRCh38
rs890282790 Chr.1: 159588009-159588009 on Build GRCh38
rs202054475 Chr.1: 159588055-159588055 on Build GRCh38
rs148924046 Chr.1: 159588158-159588158 on Build GRCh38
rs377010258 Chr.1: 159588178-159588178 on Build GRCh38
rs200362255 Chr.1: 159587842-159587842 on Build GRCh38
rs772980505 Chr.1: 159587914-159587914 on Build GRCh38
rs947549677 Chr.1: 159587876-159587876 on Build GRCh38
rs201124053 Chr.1: 159588139-159588139 on Build GRCh38
rs200849361 Chr.1: 159587797-159587797 on Build GRCh38
rs1010186302 Chr.1: 159588091-159588091 on Build GRCh38
rs755226192 Chr.1: 159588152-159588152 on Build GRCh38
rs182065199 Chr.1: 159587933-159587933 on Build GRCh38
rs746907835 Chr.1: 159587891-159587891 on Build GRCh38
rs776111690 Chr.1: 159588183-159588183 on Build GRCh38
rs767765934 Chr.1: 159587923-159587923 on Build GRCh38
rs938055440 Chr.1: 159587848-159587848 on Build GRCh38
rs201554469 Chr.1: 159587887-159587887 on Build GRCh38
rs201879111 Chr.1: 159587814-159587814 on Build GRCh38
rs916131600 Chr.1: 159587867-159587867 on Build GRCh38
rs983599155 Chr.1: 159588215-159588215 on Build GRCh38
rs968982265 Chr.1: 159588064-159588064 on Build GRCh38
rs202064188 Chr.1: 159587837-159587837 on Build GRCh38
rs757409232 Chr.1: 159588136-159588137 on Build GRCh38
rs201726959 Chr.1: 159587927-159587927 on Build GRCh38
rs200190182 Chr.1: 159587948-159587948 on Build GRCh38
rs758706848 Chr.1: 159587944-159587944 on Build GRCh38
rs141583738 Chr.1: 159588172-159588172 on Build GRCh38
rs762252459 Chr.1: 159588100-159588100 on Build GRCh38
rs943375054 Chr.1: 159588124-159588124 on Build GRCh38
rs200668921 Chr.1: 159588096-159588096 on Build GRCh38
rs200874405 Chr.1: 159587843-159587843 on Build GRCh38
rs202233773 Chr.1: 159588212-159588212 on Build GRCh38
rs748148834 Chr.1: 159587977-159587977 on Build GRCh38
rs756046861 Chr.1: 159588161-159588161 on Build GRCh38
rs138308518 Chr.1: 159588167-159588167 on Build GRCh38
rs959473527 Chr.1: 159588166-159588166 on Build GRCh38
rs750515137 Chr.1: 159588032-159588032 on Build GRCh38
rs763118993 Chr.1: 159588144-159588144 on Build GRCh38
rs759742496 Chr.1: 159588208-159588208 on Build GRCh38
rs11545899 Chr.1: 159587946-159587946 on Build GRCh38
rs544861970 Chr.1: 159587907-159587907 on Build GRCh38
rs201717830 Chr.1: 159587981-159587981 on Build GRCh38
rs995868029 Chr.1: 159587961-159587961 on Build GRCh38
rs768652881 Chr.1: 159587892-159587892 on Build GRCh38
rs199983893 Chr.1: 159588060-159588060 on Build GRCh38
rs200567682 Chr.1: 159587904-159587904 on Build GRCh38
rs1032316858 Chr.1: 159588128-159588128 on Build GRCh38
rs766659239 Chr.1: 159588052-159588052 on Build GRCh38
rs770626377 Chr.1: 159588004-159588004 on Build GRCh38
rs777098679 Chr.1: 159588016-159588016 on Build GRCh38
rs775646603 Chr.1: 159588010-159588010 on Build GRCh38
rs777355943 Chr.1: 159588075-159588075 on Build GRCh38
rs756176594 Chr.1: 159588073-159588073 on Build GRCh38
rs866268741 Chr.1: 159587990-159587990 on Build GRCh38
rs534841533 Chr.1: 159587819-159587819 on Build GRCh38
rs375578988 Chr.1: 159587996-159587996 on Build GRCh38
rs886274993 Chr.1: 159588199-159588199 on Build GRCh38
rs200374022 Chr.1: 159587866-159587866 on Build GRCh38
rs773689788 Chr.1: 159588205-159588205 on Build GRCh38
rs753977844 Chr.1: 159587956-159587956 on Build GRCh38
rs200015430 Chr.1: 159588206-159588206 on Build GRCh38
rs767855061 Chr.1: 159588211-159588211 on Build GRCh38
rs751779055 Chr.1: 159588151-159588151 on Build GRCh38
rs11545897 Chr.1: 159587931-159587931 on Build GRCh38
rs200137779 Chr.1: 159588126-159588126 on Build GRCh38
rs200643478 Chr.1: 159587964-159587964 on Build GRCh38
rs376772214 Chr.1: 159587963-159587963 on Build GRCh38
rs149736856 Chr.1: 159587932-159587932 on Build GRCh38
rs753158535 Chr.1: 159587924-159587924 on Build GRCh38
rs199985915 Chr.1: 159588013-159588013 on Build GRCh38
rs761444612 Chr.1: 159587899-159587899 on Build GRCh38
rs992155380 Chr.1: 159587851-159587851 on Build GRCh38
rs767403078 Chr.1: 159588153-159588153 on Build GRCh38
rs200370168 Chr.1: 159588189-159588189 on Build GRCh38
rs377631985 Chr.1: 159588149-159588149 on Build GRCh38
rs1043223982 Chr.1: 159587928-159587928 on Build GRCh38
rs981872636 Chr.1: 159587830-159587830 on Build GRCh38
rs771831189 Chr.1: 159587885-159587885 on Build GRCh38
rs779656352 Chr.1: 159587953-159587953 on Build GRCh38
rs979861757 Chr.1: 159588111-159588111 on Build GRCh38
rs560956584 Chr.1: 159588031-159588031 on Build GRCh38
rs746407359 Chr.1: 159588185-159588185 on Build GRCh38
rs924754759 Chr.1: 159587930-159587930 on Build GRCh38
rs751267985 Chr.1: 159588057-159588057 on Build GRCh38
rs201356482 Chr.1: 159587880-159587880 on Build GRCh38
rs925363766 Chr.1: 159587804-159587804 on Build GRCh38
rs546428006 Chr.1: 159588056-159588056 on Build GRCh38
rs745554963 Chr.1: 159588092-159588092 on Build GRCh38
rs770449950 Chr.1: 159588079-159588079 on Build GRCh38
rs528177690 Chr.1: 159588035-159588035 on Build GRCh38
rs530720208 Chr.1: 159588074-159588074 on Build GRCh38
rs748832145 Chr.1: 159588076-159588076 on Build GRCh38
rs369633878 Chr.1: 159588022-159588022 on Build GRCh38
rs769400183 Chr.1: 159587988-159587988 on Build GRCh38
rs202143231 Chr.1: 159587852-159587852 on Build GRCh38
rs773237391 Chr.1: 159588121-159588121 on Build GRCh38

View Full Product Details