Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159588323-159588835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCS
Entrez Gene ID
325
Gene Name
amyloid P component, serum
Gene Aliases
HEL-S-92n, PTX2, SAP
UniGene
Hs.507080
Gene Chromosome Location
Chr.1: 159587826-159588871 on Build GRCh38
Transcripts
NM_001639.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668126 c.412C>A p.L138M Chr.1: 159588448-159588448 on Build GRCh38
5548283 c.671G>C p.*224S Chr.1: 159588707-159588707 on Build GRCh38
3863222 c.473C>T p.S158F Chr.1: 159588509-159588509 on Build GRCh38
5580238 c.420G>A p.Q140Q Chr.1: 159588456-159588456 on Build GRCh38
675953 c.362C>T p.S121L Chr.1: 159588398-159588398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201186513 Chr.1: 159588722-159588722 on Build GRCh38
rs377148645 Chr.1: 159588746-159588746 on Build GRCh38
rs746424116 Chr.1: 159588397-159588397 on Build GRCh38
rs200793718 Chr.1: 159588693-159588693 on Build GRCh38
rs866542574 Chr.1: 159588509-159588509 on Build GRCh38
rs1012271392 Chr.1: 159588747-159588747 on Build GRCh38
rs748443924 Chr.1: 159588528-159588528 on Build GRCh38
rs202236596 Chr.1: 159588458-159588458 on Build GRCh38
rs749615402 Chr.1: 159588375-159588375 on Build GRCh38
rs572958646 Chr.1: 159588784-159588784 on Build GRCh38
rs774786617 Chr.1: 159588362-159588362 on Build GRCh38
rs754687176 Chr.1: 159588373-159588373 on Build GRCh38
rs943497399 Chr.1: 159588361-159588361 on Build GRCh38
rs759032262 Chr.1: 159588500-159588500 on Build GRCh38
rs781402077 Chr.1: 159588638-159588638 on Build GRCh38
rs757884673 Chr.1: 159588359-159588359 on Build GRCh38
rs144053685 Chr.1: 159588660-159588660 on Build GRCh38
rs780225259 Chr.1: 159588502-159588502 on Build GRCh38
rs201642560 Chr.1: 159588703-159588703 on Build GRCh38
rs201564694 Chr.1: 159588626-159588626 on Build GRCh38
rs767103503 Chr.1: 159588476-159588476 on Build GRCh38
rs200258131 Chr.1: 159588609-159588609 on Build GRCh38
rs772059311 Chr.1: 159588760-159588760 on Build GRCh38
rs771476250 Chr.1: 159588674-159588674 on Build GRCh38
rs772956949 Chr.1: 159588748-159588748 on Build GRCh38
rs760187041 Chr.1: 159588697-159588697 on Build GRCh38
rs540285209 Chr.1: 159588803-159588803 on Build GRCh38
rs201797159 Chr.1: 159588546-159588546 on Build GRCh38
rs374061734 Chr.1: 159588579-159588579 on Build GRCh38
rs754554557 Chr.1: 159588473-159588473 on Build GRCh38
rs754180281 Chr.1: 159588487-159588487 on Build GRCh38
rs145831547 Chr.1: 159588603-159588603 on Build GRCh38
rs577581014 Chr.1: 159588568-159588568 on Build GRCh38
rs894362593 Chr.1: 159588730-159588730 on Build GRCh38
rs761743822 Chr.1: 159588349-159588349 on Build GRCh38
rs868528129 Chr.1: 159588531-159588531 on Build GRCh38
rs765118018 Chr.1: 159588353-159588353 on Build GRCh38
rs773806334 Chr.1: 159588574-159588574 on Build GRCh38
rs201740730 Chr.1: 159588430-159588430 on Build GRCh38
rs752007161 Chr.1: 159588710-159588710 on Build GRCh38
rs199571947 Chr.1: 159588639-159588639 on Build GRCh38
rs1041530915 Chr.1: 159588384-159588384 on Build GRCh38
rs28383572 Chr.1: 159588360-159588360 on Build GRCh38
rs190916394 Chr.1: 159588765-159588765 on Build GRCh38
rs768896856 Chr.1: 159588624-159588624 on Build GRCh38
rs1031882412 Chr.1: 159588351-159588351 on Build GRCh38
rs201875721 Chr.1: 159588768-159588768 on Build GRCh38
rs987625787 Chr.1: 159588357-159588357 on Build GRCh38
rs200503726 Chr.1: 159588643-159588643 on Build GRCh38
rs908021545 Chr.1: 159588545-159588545 on Build GRCh38
rs747570470 Chr.1: 159588624-159588624 on Build GRCh38
rs749558737 Chr.1: 159588742-159588742 on Build GRCh38
rs746370899 Chr.1: 159588499-159588499 on Build GRCh38
rs151221908 Chr.1: 159588435-159588435 on Build GRCh38
rs200387564 Chr.1: 159588518-159588518 on Build GRCh38
rs201199341 Chr.1: 159588475-159588475 on Build GRCh38
rs201306055 Chr.1: 159588653-159588653 on Build GRCh38
rs1025071956 Chr.1: 159588472-159588472 on Build GRCh38
rs749107842 Chr.1: 159588652-159588652 on Build GRCh38
rs920646149 Chr.1: 159588782-159588782 on Build GRCh38
rs200923462 Chr.1: 159588443-159588443 on Build GRCh38
rs755860498 Chr.1: 159588606-159588606 on Build GRCh38
rs200473312 Chr.1: 159588799-159588799 on Build GRCh38
rs778744768 Chr.1: 159588750-159588750 on Build GRCh38
rs553201156 Chr.1: 159588493-159588493 on Build GRCh38
rs767100828 Chr.1: 159588585-159588585 on Build GRCh38
rs199816861 Chr.1: 159588620-159588620 on Build GRCh38
rs201686668 Chr.1: 159588505-159588505 on Build GRCh38
rs866060863 Chr.1: 159588420-159588420 on Build GRCh38
rs200112420 Chr.1: 159588402-159588402 on Build GRCh38
rs768765833 Chr.1: 159588424-159588424 on Build GRCh38
rs763843176 Chr.1: 159588702-159588702 on Build GRCh38
rs201641715 Chr.1: 159588392-159588392 on Build GRCh38
rs971247491 Chr.1: 159588759-159588759 on Build GRCh38
rs200193910 Chr.1: 159588712-159588712 on Build GRCh38
rs199954466 Chr.1: 159588757-159588757 on Build GRCh38
rs148975228 Chr.1: 159588615-159588615 on Build GRCh38
rs201860037 Chr.1: 159588648-159588648 on Build GRCh38
rs747220810 Chr.1: 159588423-159588423 on Build GRCh38
rs775122176 Chr.1: 159588682-159588682 on Build GRCh38
rs777989899 Chr.1: 159588737-159588737 on Build GRCh38
rs879661012 Chr.1: 159588451-159588451 on Build GRCh38
rs778663293 Chr.1: 159588513-159588514 on Build GRCh38
rs1004949853 Chr.1: 159588762-159588762 on Build GRCh38
rs866421868 Chr.1: 159588810-159588810 on Build GRCh38
rs771425771 Chr.1: 159588379-159588379 on Build GRCh38
rs756247140 Chr.1: 159588724-159588724 on Build GRCh38
rs1000102078 Chr.1: 159588417-159588417 on Build GRCh38
rs200753101 Chr.1: 159588426-159588426 on Build GRCh38
rs1055711865 Chr.1: 159588725-159588725 on Build GRCh38
rs200465094 Chr.1: 159588622-159588622 on Build GRCh38
rs773053482 Chr.1: 159588556-159588556 on Build GRCh38
rs758934326 Chr.1: 159588619-159588619 on Build GRCh38
rs41317511 Chr.1: 159588504-159588504 on Build GRCh38
rs567804198 Chr.1: 159588452-159588452 on Build GRCh38
rs766891924 Chr.1: 159588709-159588709 on Build GRCh38
rs984038523 Chr.1: 159588495-159588495 on Build GRCh38
rs267598114 Chr.1: 159588673-159588673 on Build GRCh38
rs200788109 Chr.1: 159588374-159588374 on Build GRCh38
rs763388925 Chr.1: 159588704-159588704 on Build GRCh38
rs1012036141 Chr.1: 159588578-159588578 on Build GRCh38
rs367910989 Chr.1: 159588580-159588580 on Build GRCh38
rs765505336 Chr.1: 159588607-159588607 on Build GRCh38
rs199934882 Chr.1: 159588437-159588437 on Build GRCh38
rs759555245 Chr.1: 159588667-159588667 on Build GRCh38
rs199669232 Chr.1: 159588371-159588371 on Build GRCh38
rs775779605 Chr.1: 159588416-159588416 on Build GRCh38
rs2808661 Chr.1: 159588468-159588468 on Build GRCh38
rs1056323078 Chr.1: 159588480-159588480 on Build GRCh38
rs765864471 Chr.1: 159588465-159588465 on Build GRCh38
rs201507609 Chr.1: 159588723-159588723 on Build GRCh38
rs749901363 Chr.1: 159588354-159588354 on Build GRCh38
rs200973914 Chr.1: 159588534-159588534 on Build GRCh38
rs144056771 Chr.1: 159588404-159588404 on Build GRCh38

View Full Product Details