Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160040796-160041293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ10
Entrez Gene ID
3766
Gene Name
potassium voltage-gated channel subfamily J member 10
Gene Aliases
BIRK-10, KCNJ13-PEN, KIR1.2, KIR4.1, SESAME
UniGene
Hs.408960
Gene Chromosome Location
Chr.1: 160037467-160070261 on Build GRCh38
Transcripts
NM_002241.4

SNP Detail

SNP ID Chromosome Location
rs986130357 Chr.1: 160040843-160040843 on Build GRCh38
rs116016979 Chr.1: 160040876-160040876 on Build GRCh38
rs930429118 Chr.1: 160041181-160041181 on Build GRCh38
rs953182006 Chr.1: 160041167-160041167 on Build GRCh38
rs1049083650 Chr.1: 160040951-160040951 on Build GRCh38
rs961700784 Chr.1: 160041147-160041147 on Build GRCh38
rs942950199 Chr.1: 160041240-160041240 on Build GRCh38
rs927662373 Chr.1: 160040836-160040836 on Build GRCh38
rs370064318 Chr.1: 160041151-160041151 on Build GRCh38
rs768946463 Chr.1: 160041251-160041251 on Build GRCh38
rs909225416 Chr.1: 160040846-160040846 on Build GRCh38
rs144487288 Chr.1: 160041127-160041127 on Build GRCh38
rs775931517 Chr.1: 160041126-160041126 on Build GRCh38
rs551940976 Chr.1: 160040954-160040954 on Build GRCh38
rs986014209 Chr.1: 160041224-160041224 on Build GRCh38
rs901146903 Chr.1: 160041242-160041242 on Build GRCh38
rs571511216 Chr.1: 160041217-160041217 on Build GRCh38
rs528816212 Chr.1: 160041031-160041031 on Build GRCh38
rs770312767 Chr.1: 160041106-160041106 on Build GRCh38
rs1010735588 Chr.1: 160041040-160041040 on Build GRCh38
rs1031107325 Chr.1: 160041034-160041034 on Build GRCh38
rs546202393 Chr.1: 160040952-160040952 on Build GRCh38
rs1041714461 Chr.1: 160041241-160041241 on Build GRCh38
rs886045412 Chr.1: 160041156-160041156 on Build GRCh38
rs184816558 Chr.1: 160040962-160040962 on Build GRCh38
rs927609860 Chr.1: 160041134-160041134 on Build GRCh38
rs920348321 Chr.1: 160041165-160041165 on Build GRCh38
rs918725037 Chr.1: 160040899-160040899 on Build GRCh38
rs777312868 Chr.1: 160041046-160041046 on Build GRCh38
rs546624560 Chr.1: 160041143-160041143 on Build GRCh38
rs971753248 Chr.1: 160041154-160041154 on Build GRCh38
rs967137131 Chr.1: 160041086-160041086 on Build GRCh38
rs567333107 Chr.1: 160041070-160041070 on Build GRCh38
rs957817123 Chr.1: 160041037-160041037 on Build GRCh38
rs1020483757 Chr.1: 160041074-160041074 on Build GRCh38
rs115725914 Chr.1: 160041256-160041256 on Build GRCh38
rs192502647 Chr.1: 160041097-160041097 on Build GRCh38
rs1016110817 Chr.1: 160041229-160041229 on Build GRCh38
rs931489971 Chr.1: 160040911-160040911 on Build GRCh38
rs1030158376 Chr.1: 160041160-160041160 on Build GRCh38
rs539110996 Chr.1: 160041222-160041222 on Build GRCh38
rs982461639 Chr.1: 160041133-160041133 on Build GRCh38
rs190544797 Chr.1: 160040975-160040975 on Build GRCh38
rs79238495 Chr.1: 160040980-160040980 on Build GRCh38
rs748563412 Chr.1: 160040946-160040946 on Build GRCh38
rs747830339 Chr.1: 160040839-160040839 on Build GRCh38
rs911410860 Chr.1: 160041235-160041235 on Build GRCh38

View Full Product Details