Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103321893-103322407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC2
Entrez Gene ID
27000
Gene Name
DnaJ heat shock protein family (Hsp40) member C2
Gene Aliases
MPHOSPH11, MPP11, ZRF1, ZUO1
UniGene
Hs.558476
Gene Chromosome Location
Chr.7: 103312474-103345362 on Build GRCh38
Transcripts
NM_001129887.1 NM_014377.1 XM_005250269.2 XM_011516028.2 XM_011516029.2 XM_011516030.2 XM_017011968.1 XM_005250717.3 XM_006716181.3

Gene Symbol
PMPCB
Entrez Gene ID
9512
Gene Name
peptidase, mitochondrial processing beta subunit
Gene Aliases
Beta-MPP, MPP11, MPPB, MPPP52, P-52
UniGene
Hs.184211
Gene Chromosome Location
Chr.7: 103297412-103329902 on Build GRCh38
Transcripts
NM_001129887.1 NM_014377.1 XM_005250269.2 XM_011516028.2 XM_011516029.2 XM_011516030.2 XM_017011968.1 XM_005250717.3 XM_006716181.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817399 c.1060C>G p.Q354E Chr.7: 103321955-103321955 on Build GRCh38
6564326 c.1035G>C p.Q345H Chr.7: 103321980-103321980 on Build GRCh38
4678731 c.1041_1042insA p.A348fs*3 Chr.7: 103321973-103321974 on Build GRCh38
4678733 c.959C>T p.A320V Chr.7: 103322056-103322056 on Build GRCh38
1204202 c.1042G>A p.A348T Chr.7: 103321973-103321973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905722768 Chr.7: 103322177-103322177 on Build GRCh38
rs749082545 Chr.7: 103322086-103322086 on Build GRCh38
rs929980120 Chr.7: 103322071-103322071 on Build GRCh38
rs202134632 Chr.7: 103322064-103322064 on Build GRCh38
rs970442830 Chr.7: 103322146-103322146 on Build GRCh38
rs555904836 Chr.7: 103322037-103322037 on Build GRCh38
rs117776402 Chr.7: 103322357-103322357 on Build GRCh38
rs1020456758 Chr.7: 103322222-103322222 on Build GRCh38
rs770209528 Chr.7: 103322092-103322093 on Build GRCh38
rs142265339 Chr.7: 103322317-103322317 on Build GRCh38
rs922668058 Chr.7: 103322325-103322325 on Build GRCh38
rs200342518 Chr.7: 103322041-103322041 on Build GRCh38
rs767069455 Chr.7: 103321986-103321986 on Build GRCh38
rs1033569865 Chr.7: 103322160-103322160 on Build GRCh38
rs754335124 Chr.7: 103321998-103321998 on Build GRCh38
rs527541360 Chr.7: 103322283-103322283 on Build GRCh38
rs545198035 Chr.7: 103322237-103322237 on Build GRCh38
rs959257494 Chr.7: 103322193-103322193 on Build GRCh38
rs531378430 Chr.7: 103322352-103322352 on Build GRCh38
rs773105471 Chr.7: 103322077-103322077 on Build GRCh38
rs993295720 Chr.7: 103322087-103322087 on Build GRCh38
rs142695020 Chr.7: 103322031-103322031 on Build GRCh38
rs987104945 Chr.7: 103322247-103322247 on Build GRCh38
rs147653871 Chr.7: 103322311-103322311 on Build GRCh38
rs372358510 Chr.7: 103321970-103321970 on Build GRCh38
rs781715794 Chr.7: 103322085-103322086 on Build GRCh38
rs772351667 Chr.7: 103321937-103321937 on Build GRCh38
rs148392195 Chr.7: 103321921-103321921 on Build GRCh38
rs200390973 Chr.7: 103322160-103322160 on Build GRCh38
rs758507524 Chr.7: 103322016-103322016 on Build GRCh38
rs994725732 Chr.7: 103322374-103322374 on Build GRCh38
rs779946577 Chr.7: 103321938-103321938 on Build GRCh38
rs1026480114 Chr.7: 103322119-103322119 on Build GRCh38
rs920015659 Chr.7: 103322298-103322298 on Build GRCh38
rs774036735 Chr.7: 103322104-103322104 on Build GRCh38
rs934021264 Chr.7: 103322328-103322328 on Build GRCh38
rs766944568 Chr.7: 103322115-103322115 on Build GRCh38
rs911435717 Chr.7: 103321963-103321963 on Build GRCh38
rs768667928 Chr.7: 103321946-103321946 on Build GRCh38
rs759061813 Chr.7: 103321985-103321985 on Build GRCh38
rs746220591 Chr.7: 103322091-103322092 on Build GRCh38
rs761908434 Chr.7: 103321953-103321953 on Build GRCh38
rs955621216 Chr.7: 103322223-103322223 on Build GRCh38
rs746021935 Chr.7: 103321925-103321925 on Build GRCh38
rs1047501505 Chr.7: 103322377-103322377 on Build GRCh38
rs747255126 Chr.7: 103322034-103322034 on Build GRCh38
rs555429309 Chr.7: 103321968-103321968 on Build GRCh38
rs988445598 Chr.7: 103322206-103322206 on Build GRCh38
rs781426067 Chr.7: 103322053-103322053 on Build GRCh38
rs1024550450 Chr.7: 103322135-103322135 on Build GRCh38
rs896522612 Chr.7: 103322353-103322353 on Build GRCh38
rs747142313 Chr.7: 103321944-103321944 on Build GRCh38
rs560402985 Chr.7: 103322254-103322254 on Build GRCh38
rs201242580 Chr.7: 103322017-103322017 on Build GRCh38
rs759175105 Chr.7: 103322106-103322106 on Build GRCh38
rs776698613 Chr.7: 103321947-103321947 on Build GRCh38
rs775509565 Chr.7: 103322114-103322114 on Build GRCh38
rs774992993 Chr.7: 103322123-103322123 on Build GRCh38
rs1001671003 Chr.7: 103322179-103322180 on Build GRCh38
rs1003149273 Chr.7: 103322147-103322147 on Build GRCh38
rs942908046 Chr.7: 103322279-103322279 on Build GRCh38
rs750821694 Chr.7: 103322009-103322009 on Build GRCh38
rs574399299 Chr.7: 103322233-103322233 on Build GRCh38
rs755104279 Chr.7: 103322042-103322042 on Build GRCh38
rs544373831 Chr.7: 103322003-103322003 on Build GRCh38
rs769647623 Chr.7: 103321962-103321962 on Build GRCh38
rs765526786 Chr.7: 103322004-103322004 on Build GRCh38
rs763268401 Chr.7: 103322116-103322117 on Build GRCh38
rs897360669 Chr.7: 103322370-103322370 on Build GRCh38
rs769877689 Chr.7: 103322059-103322059 on Build GRCh38
rs529603989 Chr.7: 103322198-103322199 on Build GRCh38
rs748044603 Chr.7: 103322054-103322054 on Build GRCh38
rs989274793 Chr.7: 103322219-103322219 on Build GRCh38
rs770677068 Chr.7: 103322102-103322102 on Build GRCh38
rs1038625938 Chr.7: 103322362-103322362 on Build GRCh38
rs180788647 Chr.7: 103322361-103322361 on Build GRCh38
rs1014219713 Chr.7: 103322134-103322134 on Build GRCh38
rs1049735761 Chr.7: 103322338-103322338 on Build GRCh38
rs751214764 Chr.7: 103321984-103321984 on Build GRCh38

View Full Product Details