Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 105605092-105605602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L1
Entrez Gene ID
222255
Gene Name
ataxin 7 like 1
Gene Aliases
ATXN7L4
UniGene
Hs.745056
Gene Chromosome Location
Chr.7: 105604774-105877232 on Build GRCh38
Transcripts
NM_020725.1 NM_138495.1 XM_005250220.3 XM_006715901.2 XM_011515935.1 XM_011515939.1 XM_011515940.1

SNP Detail

SNP ID Chromosome Location
rs576263636 Chr.7: 105605475-105605476 on Build GRCh38
rs911191057 Chr.7: 105605137-105605137 on Build GRCh38
rs894090725 Chr.7: 105605499-105605499 on Build GRCh38
rs918855944 Chr.7: 105605222-105605222 on Build GRCh38
rs1001877582 Chr.7: 105605468-105605468 on Build GRCh38
rs1046062812 Chr.7: 105605466-105605466 on Build GRCh38
rs74744964 Chr.7: 105605532-105605532 on Build GRCh38
rs929375726 Chr.7: 105605115-105605115 on Build GRCh38
rs200622868 Chr.7: 105605476-105605476 on Build GRCh38
rs941344214 Chr.7: 105605155-105605155 on Build GRCh38
rs370383193 Chr.7: 105605477-105605477 on Build GRCh38
rs777207566 Chr.7: 105605477-105605487 on Build GRCh38
rs569046128 Chr.7: 105605362-105605362 on Build GRCh38
rs186750876 Chr.7: 105605345-105605345 on Build GRCh38
rs12535264 Chr.7: 105605307-105605307 on Build GRCh38
rs182802460 Chr.7: 105605130-105605130 on Build GRCh38
rs755822501 Chr.7: 105605476-105605477 on Build GRCh38
rs909148731 Chr.7: 105605382-105605382 on Build GRCh38
rs200955030 Chr.7: 105605480-105605480 on Build GRCh38
rs143738311 Chr.7: 105605340-105605340 on Build GRCh38
rs974087881 Chr.7: 105605516-105605516 on Build GRCh38
rs1001826709 Chr.7: 105605203-105605203 on Build GRCh38
rs543116849 Chr.7: 105605487-105605488 on Build GRCh38
rs897561474 Chr.7: 105605459-105605459 on Build GRCh38
rs1034570697 Chr.7: 105605495-105605495 on Build GRCh38
rs1034602888 Chr.7: 105605225-105605225 on Build GRCh38
rs957491682 Chr.7: 105605305-105605305 on Build GRCh38
rs957626866 Chr.7: 105605515-105605515 on Build GRCh38
rs536522646 Chr.7: 105605474-105605475 on Build GRCh38
rs904820986 Chr.7: 105605467-105605467 on Build GRCh38
rs538202920 Chr.7: 105605487-105605487 on Build GRCh38
rs951658997 Chr.7: 105605543-105605543 on Build GRCh38
rs529304714 Chr.7: 105605400-105605400 on Build GRCh38
rs777124679 Chr.7: 105605477-105605477 on Build GRCh38
rs745382605 Chr.7: 105605475-105605475 on Build GRCh38
rs1013234458 Chr.7: 105605126-105605126 on Build GRCh38
rs202117184 Chr.7: 105605486-105605487 on Build GRCh38
rs913292017 Chr.7: 105605337-105605337 on Build GRCh38
rs941846082 Chr.7: 105605395-105605395 on Build GRCh38
rs576844333 Chr.7: 105605484-105605487 on Build GRCh38
rs569559260 Chr.7: 105605481-105605481 on Build GRCh38
rs930247685 Chr.7: 105605237-105605237 on Build GRCh38
rs1020899338 Chr.7: 105605544-105605544 on Build GRCh38
rs1054561088 Chr.7: 105605381-105605381 on Build GRCh38
rs918121083 Chr.7: 105605329-105605329 on Build GRCh38
rs200779433 Chr.7: 105605485-105605487 on Build GRCh38
rs1018387270 Chr.7: 105605482-105605482 on Build GRCh38
rs17150293 Chr.7: 105605430-105605430 on Build GRCh38
rs535179062 Chr.7: 105605463-105605463 on Build GRCh38
rs746724851 Chr.7: 105605477-105605478 on Build GRCh38
rs199658092 Chr.7: 105605479-105605479 on Build GRCh38
rs117515381 Chr.7: 105605124-105605124 on Build GRCh38
rs1025091833 Chr.7: 105605127-105605127 on Build GRCh38
rs370062610 Chr.7: 105605521-105605521 on Build GRCh38
rs111736885 Chr.7: 105605205-105605205 on Build GRCh38
rs77521628 Chr.7: 105605343-105605343 on Build GRCh38
rs79987178 Chr.7: 105605361-105605361 on Build GRCh38
rs67686948 Chr.7: 105605487-105605487 on Build GRCh38

View Full Product Details