Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 105605499-105606007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L1
Entrez Gene ID
222255
Gene Name
ataxin 7 like 1
Gene Aliases
ATXN7L4
UniGene
Hs.745056
Gene Chromosome Location
Chr.7: 105604774-105877232 on Build GRCh38
Transcripts
NM_020725.1 NM_138495.1 XM_005250220.3 XM_006715901.2 XM_011515935.1 XM_011515939.1 XM_011515940.1

SNP Detail

SNP ID Chromosome Location
rs367941083 Chr.7: 105605669-105605669 on Build GRCh38
rs571699895 Chr.7: 105605798-105605798 on Build GRCh38
rs951658997 Chr.7: 105605543-105605543 on Build GRCh38
rs962808817 Chr.7: 105605897-105605897 on Build GRCh38
rs867381541 Chr.7: 105605630-105605630 on Build GRCh38
rs796537985 Chr.7: 105605636-105605636 on Build GRCh38
rs867168492 Chr.7: 105605745-105605745 on Build GRCh38
rs996033843 Chr.7: 105605917-105605917 on Build GRCh38
rs74744964 Chr.7: 105605532-105605532 on Build GRCh38
rs949250739 Chr.7: 105605780-105605780 on Build GRCh38
rs918181904 Chr.7: 105605976-105605976 on Build GRCh38
rs373501617 Chr.7: 105605945-105605945 on Build GRCh38
rs893377993 Chr.7: 105605921-105605921 on Build GRCh38
rs571781606 Chr.7: 105605621-105605621 on Build GRCh38
rs937636471 Chr.7: 105605881-105605881 on Build GRCh38
rs1020899338 Chr.7: 105605544-105605544 on Build GRCh38
rs190961073 Chr.7: 105605660-105605660 on Build GRCh38
rs1053471698 Chr.7: 105605892-105605892 on Build GRCh38
rs935774945 Chr.7: 105605675-105605677 on Build GRCh38
rs890937822 Chr.7: 105605833-105605833 on Build GRCh38
rs1012274962 Chr.7: 105605931-105605931 on Build GRCh38
rs1006738552 Chr.7: 105605876-105605876 on Build GRCh38
rs1051010089 Chr.7: 105605793-105605793 on Build GRCh38
rs951463025 Chr.7: 105605959-105605959 on Build GRCh38
rs992829532 Chr.7: 105605595-105605595 on Build GRCh38
rs986114330 Chr.7: 105605644-105605644 on Build GRCh38
rs904956340 Chr.7: 105605878-105605878 on Build GRCh38
rs1046536542 Chr.7: 105605830-105605830 on Build GRCh38
rs941970995 Chr.7: 105605720-105605720 on Build GRCh38
rs992399820 Chr.7: 105605972-105605972 on Build GRCh38
rs976610938 Chr.7: 105605622-105605622 on Build GRCh38
rs971915875 Chr.7: 105605744-105605744 on Build GRCh38

View Full Product Details