Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 105606329-105606838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L1
Entrez Gene ID
222255
Gene Name
ataxin 7 like 1
Gene Aliases
ATXN7L4
UniGene
Hs.745056
Gene Chromosome Location
Chr.7: 105604774-105877232 on Build GRCh38
Transcripts
NM_020725.1 NM_138495.1 XM_005250220.3 XM_006715901.2 XM_011515935.1 XM_011515939.1 XM_011515940.1

SNP Detail

SNP ID Chromosome Location
rs933609699 Chr.7: 105606785-105606785 on Build GRCh38
rs113369791 Chr.7: 105606638-105606638 on Build GRCh38
rs747242010 Chr.7: 105606569-105606569 on Build GRCh38
rs912196514 Chr.7: 105606779-105606779 on Build GRCh38
rs1026625731 Chr.7: 105606493-105606493 on Build GRCh38
rs959107329 Chr.7: 105606597-105606597 on Build GRCh38
rs1021146635 Chr.7: 105606696-105606696 on Build GRCh38
rs898306117 Chr.7: 105606411-105606411 on Build GRCh38
rs945016112 Chr.7: 105606653-105606653 on Build GRCh38
rs146898491 Chr.7: 105606649-105606649 on Build GRCh38
rs141967130 Chr.7: 105606637-105606637 on Build GRCh38
rs963726390 Chr.7: 105606784-105606784 on Build GRCh38
rs1024925558 Chr.7: 105606573-105606573 on Build GRCh38
rs560573340 Chr.7: 105606490-105606490 on Build GRCh38
rs571784941 Chr.7: 105606778-105606778 on Build GRCh38
rs986581281 Chr.7: 105606760-105606760 on Build GRCh38
rs771197449 Chr.7: 105606605-105606605 on Build GRCh38
rs1039650046 Chr.7: 105606375-105606375 on Build GRCh38
rs532015633 Chr.7: 105606702-105606702 on Build GRCh38
rs529446678 Chr.7: 105606544-105606544 on Build GRCh38
rs988700108 Chr.7: 105606669-105606669 on Build GRCh38
rs540672820 Chr.7: 105606403-105606403 on Build GRCh38
rs887175055 Chr.7: 105606532-105606532 on Build GRCh38
rs532070058 Chr.7: 105606534-105606534 on Build GRCh38
rs1016305287 Chr.7: 105606377-105606377 on Build GRCh38
rs759700392 Chr.7: 105606743-105606743 on Build GRCh38
rs963675454 Chr.7: 105606466-105606466 on Build GRCh38
rs551803572 Chr.7: 105606734-105606734 on Build GRCh38
rs12154239 Chr.7: 105606374-105606374 on Build GRCh38
rs1046920806 Chr.7: 105606487-105606487 on Build GRCh38
rs926474783 Chr.7: 105606566-105606566 on Build GRCh38
rs375021267 Chr.7: 105606756-105606756 on Build GRCh38
rs1002370593 Chr.7: 105606625-105606625 on Build GRCh38
rs958201834 Chr.7: 105606666-105606666 on Build GRCh38
rs931153250 Chr.7: 105606453-105606453 on Build GRCh38
rs557634908 Chr.7: 105606697-105606697 on Build GRCh38

View Full Product Details