Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 105638074-105638578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L1
Entrez Gene ID
222255
Gene Name
ataxin 7 like 1
Gene Aliases
ATXN7L4
UniGene
Hs.745056
Gene Chromosome Location
Chr.7: 105604774-105877232 on Build GRCh38
Transcripts
NM_001318229.1 NM_020725.1 NM_138495.1 XM_005250220.3 XM_006715900.2 XM_006715901.2 XM_006715902.3 XM_006715903.3 XM_006715904.3 XM_011515935.1 XM_011515936.2 XM_011515937.2 XM_011515938.2 XM_011515939.1 XM_011515940.1 XM_011515941.2 XM_011515942.2 XM_011515943.2 XM_011515944.1 XM_011515945.1 XM_011515946.1 XM_011515947.1 XM_011515950.1 XM_017011858.1 XM_017011859.1 XM_017011860.1 XM_017011861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357125 c.1198C>T p.P400S Chr.7: 105638357-105638357 on Build GRCh38
3254263 c.1110G>T p.Q370H Chr.7: 105638445-105638445 on Build GRCh38
4595797 c.1151C>T p.P384L Chr.7: 105638404-105638404 on Build GRCh38
5170077 c.1085T>C p.L362P Chr.7: 105638470-105638470 on Build GRCh38
4384193 c.1115C>G p.S372C Chr.7: 105638440-105638440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370326878 Chr.7: 105638506-105638506 on Build GRCh38
rs528334599 Chr.7: 105638312-105638312 on Build GRCh38
rs1008588700 Chr.7: 105638103-105638103 on Build GRCh38
rs769933011 Chr.7: 105638504-105638504 on Build GRCh38
rs569896299 Chr.7: 105638200-105638200 on Build GRCh38
rs948646886 Chr.7: 105638330-105638330 on Build GRCh38
rs752247008 Chr.7: 105638356-105638356 on Build GRCh38
rs924631591 Chr.7: 105638318-105638318 on Build GRCh38
rs982970743 Chr.7: 105638512-105638512 on Build GRCh38
rs938744402 Chr.7: 105638309-105638309 on Build GRCh38
rs751942706 Chr.7: 105638479-105638479 on Build GRCh38
rs952925035 Chr.7: 105638501-105638501 on Build GRCh38
rs200723756 Chr.7: 105638451-105638451 on Build GRCh38
rs150248213 Chr.7: 105638336-105638336 on Build GRCh38
rs376104641 Chr.7: 105638525-105638525 on Build GRCh38
rs952894398 Chr.7: 105638207-105638207 on Build GRCh38
rs904441453 Chr.7: 105638341-105638341 on Build GRCh38
rs778355356 Chr.7: 105638231-105638231 on Build GRCh38
rs1050105183 Chr.7: 105638430-105638430 on Build GRCh38
rs961967645 Chr.7: 105638102-105638102 on Build GRCh38
rs751432843 Chr.7: 105638373-105638373 on Build GRCh38
rs867234926 Chr.7: 105638422-105638422 on Build GRCh38
rs765755744 Chr.7: 105638435-105638435 on Build GRCh38
rs759536093 Chr.7: 105638313-105638314 on Build GRCh38
rs1017280061 Chr.7: 105638418-105638418 on Build GRCh38
rs932829023 Chr.7: 105638402-105638402 on Build GRCh38
rs914670411 Chr.7: 105638253-105638253 on Build GRCh38
rs138918501 Chr.7: 105638475-105638475 on Build GRCh38
rs796892847 Chr.7: 105638486-105638486 on Build GRCh38
rs982937984 Chr.7: 105638214-105638214 on Build GRCh38
rs977469520 Chr.7: 105638314-105638314 on Build GRCh38
rs915867749 Chr.7: 105638322-105638322 on Build GRCh38
rs530507362 Chr.7: 105638248-105638248 on Build GRCh38
rs749761476 Chr.7: 105638456-105638456 on Build GRCh38
rs1009036288 Chr.7: 105638412-105638412 on Build GRCh38
rs1017665612 Chr.7: 105638140-105638140 on Build GRCh38
rs958565098 Chr.7: 105638195-105638195 on Build GRCh38
rs759168490 Chr.7: 105638335-105638335 on Build GRCh38
rs1035517827 Chr.7: 105638186-105638186 on Build GRCh38
rs745640458 Chr.7: 105638524-105638524 on Build GRCh38
rs975630706 Chr.7: 105638181-105638181 on Build GRCh38
rs758798783 Chr.7: 105638467-105638467 on Build GRCh38
rs770886022 Chr.7: 105638550-105638550 on Build GRCh38
rs556888064 Chr.7: 105638500-105638500 on Build GRCh38
rs769920897 Chr.7: 105638318-105638326 on Build GRCh38
rs142922391 Chr.7: 105638172-105638172 on Build GRCh38
rs1053345851 Chr.7: 105638371-105638371 on Build GRCh38
rs781521927 Chr.7: 105638481-105638481 on Build GRCh38
rs780914400 Chr.7: 105638457-105638457 on Build GRCh38
rs755288185 Chr.7: 105638480-105638480 on Build GRCh38

View Full Product Details