Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107201251-107201757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG5
Entrez Gene ID
10466
Gene Name
component of oligomeric golgi complex 5
Gene Aliases
CDG2I, GOLTC1, GTC90
UniGene
Hs.239631
Gene Chromosome Location
Chr.7: 107201743-107564514 on Build GRCh38
Transcripts
NM_006348.3 NM_181733.2 NM_001244262.1 NM_012257.3 XM_005250266.2 XM_005250267.2 XM_017011967.1

Gene Symbol
HBP1
Entrez Gene ID
26959
Gene Name
HMG-box transcription factor 1
Gene Aliases
-
UniGene
Hs.162032
Gene Chromosome Location
Chr.7: 107168961-107202529 on Build GRCh38
Transcripts
NM_006348.3 NM_181733.2 NM_001244262.1 NM_012257.3 XM_005250266.2 XM_005250267.2 XM_017011967.1

SNP Detail

SNP ID Chromosome Location
rs987092102 Chr.7: 107201484-107201484 on Build GRCh38
rs748994897 Chr.7: 107201380-107201381 on Build GRCh38
rs1001374263 Chr.7: 107201467-107201467 on Build GRCh38
rs1044367246 Chr.7: 107201546-107201546 on Build GRCh38
rs1009791925 Chr.7: 107201584-107201584 on Build GRCh38
rs529549920 Chr.7: 107201679-107201679 on Build GRCh38
rs570071183 Chr.7: 107201549-107201549 on Build GRCh38
rs140869666 Chr.7: 107201625-107201625 on Build GRCh38
rs996635341 Chr.7: 107201638-107201638 on Build GRCh38
rs1055630710 Chr.7: 107201723-107201723 on Build GRCh38
rs544694864 Chr.7: 107201331-107201331 on Build GRCh38
rs778763756 Chr.7: 107201365-107201365 on Build GRCh38
rs1041263171 Chr.7: 107201627-107201638 on Build GRCh38
rs975003649 Chr.7: 107201347-107201347 on Build GRCh38
rs1056277387 Chr.7: 107201578-107201578 on Build GRCh38
rs765212427 Chr.7: 107201731-107201734 on Build GRCh38
rs760187814 Chr.7: 107201452-107201453 on Build GRCh38
rs907251834 Chr.7: 107201563-107201563 on Build GRCh38
rs770526035 Chr.7: 107201529-107201529 on Build GRCh38
rs771774154 Chr.7: 107201401-107201402 on Build GRCh38
rs769329259 Chr.7: 107201458-107201458 on Build GRCh38
rs561139290 Chr.7: 107201499-107201499 on Build GRCh38
rs760018179 Chr.7: 107201464-107201464 on Build GRCh38
rs571520604 Chr.7: 107201713-107201713 on Build GRCh38
rs776238759 Chr.7: 107201426-107201426 on Build GRCh38
rs371194533 Chr.7: 107201375-107201375 on Build GRCh38
rs200187637 Chr.7: 107201394-107201394 on Build GRCh38
rs767877405 Chr.7: 107201466-107201466 on Build GRCh38
rs550225636 Chr.7: 107201507-107201507 on Build GRCh38
rs746675115 Chr.7: 107201386-107201386 on Build GRCh38
rs747785547 Chr.7: 107201400-107201401 on Build GRCh38
rs781036742 Chr.7: 107201678-107201679 on Build GRCh38
rs191952814 Chr.7: 107201643-107201643 on Build GRCh38
rs866959074 Chr.7: 107201306-107201306 on Build GRCh38
rs1011398658 Chr.7: 107201492-107201493 on Build GRCh38
rs745768937 Chr.7: 107201367-107201367 on Build GRCh38
rs575270872 Chr.7: 107201440-107201440 on Build GRCh38
rs547734050 Chr.7: 107201462-107201462 on Build GRCh38
rs774715082 Chr.7: 107201703-107201703 on Build GRCh38
rs908800575 Chr.7: 107201490-107201490 on Build GRCh38
rs1045903251 Chr.7: 107201684-107201684 on Build GRCh38
rs865956584 Chr.7: 107201287-107201287 on Build GRCh38
rs774062919 Chr.7: 107201395-107201400 on Build GRCh38
rs745972145 Chr.7: 107201337-107201338 on Build GRCh38
rs34979723 Chr.7: 107201412-107201413 on Build GRCh38
rs954989032 Chr.7: 107201671-107201671 on Build GRCh38
rs144592852 Chr.7: 107201362-107201362 on Build GRCh38
rs768464934 Chr.7: 107201391-107201392 on Build GRCh38
rs1002830899 Chr.7: 107201575-107201575 on Build GRCh38
rs746984593 Chr.7: 107201581-107201584 on Build GRCh38
rs752989307 Chr.7: 107201479-107201479 on Build GRCh38
rs761182181 Chr.7: 107201437-107201437 on Build GRCh38
rs772059782 Chr.7: 107201368-107201368 on Build GRCh38
rs565198969 Chr.7: 107201352-107201352 on Build GRCh38
rs922059701 Chr.7: 107201354-107201354 on Build GRCh38
rs967842590 Chr.7: 107201399-107201399 on Build GRCh38
rs966846011 Chr.7: 107201510-107201510 on Build GRCh38
rs774665382 Chr.7: 107201419-107201421 on Build GRCh38
rs562248212 Chr.7: 107201629-107201629 on Build GRCh38
rs908767852 Chr.7: 107201685-107201685 on Build GRCh38
rs530266824 Chr.7: 107201503-107201503 on Build GRCh38
rs768129883 Chr.7: 107201425-107201425 on Build GRCh38
rs886223426 Chr.7: 107201409-107201409 on Build GRCh38
rs1022152894 Chr.7: 107201596-107201596 on Build GRCh38
rs906880981 Chr.7: 107201465-107201465 on Build GRCh38
rs1000038820 Chr.7: 107201574-107201575 on Build GRCh38
rs974634233 Chr.7: 107201705-107201705 on Build GRCh38
rs948859334 Chr.7: 107201538-107201538 on Build GRCh38
rs775413724 Chr.7: 107201369-107201369 on Build GRCh38
rs529514798 Chr.7: 107201344-107201345 on Build GRCh38
rs907598098 Chr.7: 107201652-107201652 on Build GRCh38
rs940154744 Chr.7: 107201498-107201498 on Build GRCh38
rs892397863 Chr.7: 107201581-107201581 on Build GRCh38

View Full Product Details