Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107202694-107203239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG5
Entrez Gene ID
10466
Gene Name
component of oligomeric golgi complex 5
Gene Aliases
CDG2I, GOLTC1, GTC90
UniGene
Hs.239631
Gene Chromosome Location
Chr.7: 107201743-107564514 on Build GRCh38
Transcripts
NM_006348.3 NM_181733.2

SNP Detail

SNP ID Chromosome Location
rs1020899034 Chr.7: 107203046-107203046 on Build GRCh38
rs6958137 Chr.7: 107202904-107202904 on Build GRCh38
rs15002 Chr.7: 107202852-107202852 on Build GRCh38
rs987482953 Chr.7: 107203084-107203084 on Build GRCh38
rs904650312 Chr.7: 107203144-107203144 on Build GRCh38
rs938737403 Chr.7: 107202855-107202859 on Build GRCh38
rs912491229 Chr.7: 107202811-107202811 on Build GRCh38
rs532794584 Chr.7: 107202799-107202799 on Build GRCh38
rs534463628 Chr.7: 107202806-107202806 on Build GRCh38
rs998632979 Chr.7: 107202722-107202722 on Build GRCh38
rs193127052 Chr.7: 107203079-107203079 on Build GRCh38
rs965080444 Chr.7: 107202751-107202751 on Build GRCh38
rs984051958 Chr.7: 107202972-107202974 on Build GRCh38
rs942082962 Chr.7: 107202813-107202813 on Build GRCh38
rs1031724777 Chr.7: 107202738-107202738 on Build GRCh38
rs771283784 Chr.7: 107203132-107203132 on Build GRCh38
rs373785950 Chr.7: 107202824-107202824 on Build GRCh38
rs563727099 Chr.7: 107202761-107202761 on Build GRCh38
rs972558203 Chr.7: 107202980-107202980 on Build GRCh38
rs976625406 Chr.7: 107202781-107202781 on Build GRCh38
rs527645957 Chr.7: 107202882-107202882 on Build GRCh38
rs934884770 Chr.7: 107203150-107203150 on Build GRCh38
rs1017147054 Chr.7: 107202973-107202973 on Build GRCh38
rs61088959 Chr.7: 107202972-107202972 on Build GRCh38
rs569737967 Chr.7: 107202918-107202918 on Build GRCh38
rs919381974 Chr.7: 107202987-107202987 on Build GRCh38
rs1031422675 Chr.7: 107202913-107202913 on Build GRCh38
rs894757078 Chr.7: 107202899-107202899 on Build GRCh38
rs968822130 Chr.7: 107203054-107203054 on Build GRCh38
rs1057305902 Chr.7: 107202861-107202861 on Build GRCh38
rs1057284923 Chr.7: 107202969-107202969 on Build GRCh38
rs547054779 Chr.7: 107202730-107202730 on Build GRCh38
rs550092854 Chr.7: 107202914-107202914 on Build GRCh38
rs932552080 Chr.7: 107202782-107202782 on Build GRCh38
rs185768753 Chr.7: 107203165-107203165 on Build GRCh38
rs547910604 Chr.7: 107202847-107202847 on Build GRCh38
rs978770946 Chr.7: 107202732-107202732 on Build GRCh38
rs767126074 Chr.7: 107202958-107202958 on Build GRCh38
rs772464238 Chr.7: 107202910-107202910 on Build GRCh38
rs896087446 Chr.7: 107202970-107202970 on Build GRCh38
rs931075544 Chr.7: 107203203-107203203 on Build GRCh38
rs1036459118 Chr.7: 107202817-107202817 on Build GRCh38
rs918324534 Chr.7: 107202879-107202879 on Build GRCh38
rs1010383711 Chr.7: 107203023-107203023 on Build GRCh38
rs114915566 Chr.7: 107202992-107202992 on Build GRCh38
rs761301900 Chr.7: 107203082-107203082 on Build GRCh38
rs34913407 Chr.7: 107203137-107203137 on Build GRCh38
rs546302907 Chr.7: 107202812-107202812 on Build GRCh38
rs992810506 Chr.7: 107203013-107203013 on Build GRCh38
rs558203688 Chr.7: 107202983-107202983 on Build GRCh38
rs904702211 Chr.7: 107202907-107202907 on Build GRCh38
rs538476088 Chr.7: 107202966-107202966 on Build GRCh38
rs921960082 Chr.7: 107202774-107202774 on Build GRCh38
rs548613467 Chr.7: 107202894-107202894 on Build GRCh38
rs554269208 Chr.7: 107203060-107203060 on Build GRCh38
rs1037291827 Chr.7: 107202878-107202878 on Build GRCh38
rs886061871 Chr.7: 107203048-107203048 on Build GRCh38
rs912460078 Chr.7: 107203107-107203107 on Build GRCh38
rs905577615 Chr.7: 107202922-107202922 on Build GRCh38
rs149142658 Chr.7: 107202838-107202838 on Build GRCh38
rs1000725722 Chr.7: 107203055-107203055 on Build GRCh38
rs918451375 Chr.7: 107203171-107203171 on Build GRCh38
rs534277881 Chr.7: 107202993-107202993 on Build GRCh38
rs961241552 Chr.7: 107202978-107202978 on Build GRCh38

View Full Product Details