Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107203126-107203629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG5
Entrez Gene ID
10466
Gene Name
component of oligomeric golgi complex 5
Gene Aliases
CDG2I, GOLTC1, GTC90
UniGene
Hs.239631
Gene Chromosome Location
Chr.7: 107201743-107564514 on Build GRCh38
Transcripts
NM_006348.3 NM_181733.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4155364 c.2470G>T p.G824* Chr.7: 107203629-107203629 on Build GRCh38
5434193 c.2513G>A p.G838D Chr.7: 107203586-107203586 on Build GRCh38
5979418 c.2578C>T p.Q860* Chr.7: 107203521-107203521 on Build GRCh38
1568708 c.2489T>G p.V830G Chr.7: 107203610-107203610 on Build GRCh38
1083840 c.2518G>T p.E840* Chr.7: 107203581-107203581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965317962 Chr.7: 107203388-107203388 on Build GRCh38
rs746763077 Chr.7: 107203524-107203524 on Build GRCh38
rs193161476 Chr.7: 107203392-107203392 on Build GRCh38
rs900944884 Chr.7: 107203220-107203220 on Build GRCh38
rs775853693 Chr.7: 107203573-107203573 on Build GRCh38
rs952620946 Chr.7: 107203424-107203424 on Build GRCh38
rs1802326 Chr.7: 107203361-107203361 on Build GRCh38
rs937117794 Chr.7: 107203464-107203464 on Build GRCh38
rs1016893906 Chr.7: 107203295-107203295 on Build GRCh38
rs563267149 Chr.7: 107203525-107203525 on Build GRCh38
rs542357355 Chr.7: 107203413-107203413 on Build GRCh38
rs1010936552 Chr.7: 107203289-107203289 on Build GRCh38
rs769241945 Chr.7: 107203541-107203541 on Build GRCh38
rs530971455 Chr.7: 107203431-107203431 on Build GRCh38
rs983934927 Chr.7: 107203429-107203429 on Build GRCh38
rs889454863 Chr.7: 107203556-107203556 on Build GRCh38
rs777167502 Chr.7: 107203532-107203533 on Build GRCh38
rs757320773 Chr.7: 107203481-107203481 on Build GRCh38
rs762082725 Chr.7: 107203607-107203607 on Build GRCh38
rs760038268 Chr.7: 107203555-107203555 on Build GRCh38
rs577491628 Chr.7: 107203279-107203279 on Build GRCh38
rs1000375620 Chr.7: 107203354-107203354 on Build GRCh38
rs549788738 Chr.7: 107203521-107203521 on Build GRCh38
rs897091614 Chr.7: 107203579-107203579 on Build GRCh38
rs561925830 Chr.7: 107203422-107203422 on Build GRCh38
rs934884770 Chr.7: 107203150-107203150 on Build GRCh38
rs979420649 Chr.7: 107203384-107203384 on Build GRCh38
rs1050606884 Chr.7: 107203574-107203574 on Build GRCh38
rs533669494 Chr.7: 107203400-107203400 on Build GRCh38
rs760015202 Chr.7: 107203603-107203606 on Build GRCh38
rs916939955 Chr.7: 107203485-107203485 on Build GRCh38
rs1049510631 Chr.7: 107203540-107203540 on Build GRCh38
rs571484508 Chr.7: 107203348-107203348 on Build GRCh38
rs754067805 Chr.7: 107203478-107203478 on Build GRCh38
rs776776780 Chr.7: 107203264-107203264 on Build GRCh38
rs750444305 Chr.7: 107203508-107203508 on Build GRCh38
rs760915377 Chr.7: 107203467-107203467 on Build GRCh38
rs201334786 Chr.7: 107203586-107203586 on Build GRCh38
rs751497917 Chr.7: 107203355-107203356 on Build GRCh38
rs889191758 Chr.7: 107203584-107203584 on Build GRCh38
rs779046049 Chr.7: 107203501-107203501 on Build GRCh38
rs1041885692 Chr.7: 107203309-107203309 on Build GRCh38
rs747809652 Chr.7: 107203527-107203527 on Build GRCh38
rs765579393 Chr.7: 107203314-107203314 on Build GRCh38
rs1027964670 Chr.7: 107203254-107203254 on Build GRCh38
rs771306316 Chr.7: 107203500-107203514 on Build GRCh38
rs141942249 Chr.7: 107203605-107203605 on Build GRCh38
rs1026795823 Chr.7: 107203329-107203330 on Build GRCh38
rs759494116 Chr.7: 107203274-107203274 on Build GRCh38
rs185768753 Chr.7: 107203165-107203165 on Build GRCh38
rs868650147 Chr.7: 107203583-107203583 on Build GRCh38
rs34100214 Chr.7: 107203550-107203550 on Build GRCh38
rs886061872 Chr.7: 107203269-107203269 on Build GRCh38
rs972618505 Chr.7: 107203323-107203323 on Build GRCh38
rs931075544 Chr.7: 107203203-107203203 on Build GRCh38
rs570136021 Chr.7: 107203334-107203338 on Build GRCh38
rs200090407 Chr.7: 107203602-107203602 on Build GRCh38
rs746241629 Chr.7: 107203383-107203383 on Build GRCh38
rs1005257968 Chr.7: 107203557-107203557 on Build GRCh38
rs1008884469 Chr.7: 107203386-107203386 on Build GRCh38
rs758354967 Chr.7: 107203512-107203512 on Build GRCh38
rs927024440 Chr.7: 107203437-107203437 on Build GRCh38
rs961496888 Chr.7: 107203305-107203305 on Build GRCh38
rs918514411 Chr.7: 107203414-107203414 on Build GRCh38
rs373781763 Chr.7: 107203473-107203473 on Build GRCh38
rs559846037 Chr.7: 107203368-107203368 on Build GRCh38
rs927055005 Chr.7: 107203250-107203250 on Build GRCh38
rs763239163 Chr.7: 107203387-107203387 on Build GRCh38
rs992444192 Chr.7: 107203465-107203465 on Build GRCh38
rs901352133 Chr.7: 107203433-107203433 on Build GRCh38
rs1017248688 Chr.7: 107203560-107203560 on Build GRCh38
rs918451375 Chr.7: 107203171-107203171 on Build GRCh38
rs188638963 Chr.7: 107203359-107203359 on Build GRCh38
rs6958494 Chr.7: 107203223-107203223 on Build GRCh38
rs1005434254 Chr.7: 107203270-107203270 on Build GRCh38
rs936981914 Chr.7: 107203261-107203261 on Build GRCh38
rs772525300 Chr.7: 107203565-107203565 on Build GRCh38

View Full Product Details