Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 111736791-111737293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK4
Entrez Gene ID
9732
Gene Name
dedicator of cytokinesis 4
Gene Aliases
-
UniGene
Hs.654652
Gene Chromosome Location
Chr.7: 111726108-112206407 on Build GRCh38
Transcripts
NM_014705.3 XM_006716188.2 XM_006716189.2 XM_017012819.1 XM_017012820.1 XM_017012821.1 XM_017012822.1 XM_017012823.1 XM_017012824.1 XM_017012825.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5406235 c.5255C>T p.S1752F Chr.7: 111736931-111736931 on Build GRCh38
5406236 c.1913C>T p.S638F Chr.7: 111736931-111736931 on Build GRCh38
3877257 c.5209A>G p.N1737D Chr.7: 111736977-111736977 on Build GRCh38
4358869 c.1879G>A p.D627N Chr.7: 111736965-111736965 on Build GRCh38
4964691 c.1882G>A p.G628R Chr.7: 111736962-111736962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776505417 Chr.7: 111737060-111737060 on Build GRCh38
rs751183443 Chr.7: 111736988-111736988 on Build GRCh38
rs984775911 Chr.7: 111737083-111737083 on Build GRCh38
rs1033036628 Chr.7: 111736904-111736904 on Build GRCh38
rs768706438 Chr.7: 111736950-111736950 on Build GRCh38
rs757910835 Chr.7: 111736970-111736970 on Build GRCh38
rs953292573 Chr.7: 111737011-111737011 on Build GRCh38
rs1026145387 Chr.7: 111737269-111737269 on Build GRCh38
rs993272595 Chr.7: 111737262-111737262 on Build GRCh38
rs184672508 Chr.7: 111736877-111736877 on Build GRCh38
rs1054855279 Chr.7: 111737212-111737212 on Build GRCh38
rs776151471 Chr.7: 111737120-111737120 on Build GRCh38
rs763662318 Chr.7: 111736973-111736973 on Build GRCh38
rs375220879 Chr.7: 111737047-111737047 on Build GRCh38
rs189055113 Chr.7: 111737029-111737029 on Build GRCh38
rs535465340 Chr.7: 111736997-111736997 on Build GRCh38
rs889129910 Chr.7: 111737220-111737220 on Build GRCh38
rs149384765 Chr.7: 111736845-111736845 on Build GRCh38
rs1030665813 Chr.7: 111736865-111736865 on Build GRCh38
rs767870061 Chr.7: 111736995-111736996 on Build GRCh38
rs760442905 Chr.7: 111736847-111736847 on Build GRCh38
rs745710848 Chr.7: 111737001-111737001 on Build GRCh38
rs564183702 Chr.7: 111736949-111736949 on Build GRCh38
rs763945143 Chr.7: 111737079-111737079 on Build GRCh38
rs991745576 Chr.7: 111737121-111737121 on Build GRCh38
rs761495439 Chr.7: 111737190-111737190 on Build GRCh38
rs948983081 Chr.7: 111737113-111737113 on Build GRCh38
rs750933826 Chr.7: 111737210-111737210 on Build GRCh38
rs765718205 Chr.7: 111737207-111737207 on Build GRCh38
rs770246903 Chr.7: 111737044-111737044 on Build GRCh38
rs1052641345 Chr.7: 111737243-111737243 on Build GRCh38
rs1013016381 Chr.7: 111736825-111736825 on Build GRCh38
rs1040286885 Chr.7: 111737248-111737248 on Build GRCh38
rs901797525 Chr.7: 111737257-111737257 on Build GRCh38
rs762392933 Chr.7: 111737089-111737089 on Build GRCh38
rs1041935165 Chr.7: 111737193-111737193 on Build GRCh38
rs1046335724 Chr.7: 111736833-111736833 on Build GRCh38
rs1026006181 Chr.7: 111737005-111737005 on Build GRCh38
rs761046979 Chr.7: 111736961-111736961 on Build GRCh38
rs138260886 Chr.7: 111737183-111737183 on Build GRCh38
rs1007850141 Chr.7: 111737245-111737245 on Build GRCh38
rs182404082 Chr.7: 111737059-111737059 on Build GRCh38
rs755942259 Chr.7: 111737006-111737006 on Build GRCh38
rs775396975 Chr.7: 111736924-111736924 on Build GRCh38
rs762240664 Chr.7: 111736917-111736918 on Build GRCh38
rs930669366 Chr.7: 111737072-111737072 on Build GRCh38
rs773731165 Chr.7: 111736959-111736959 on Build GRCh38
rs187011569 Chr.7: 111737101-111737101 on Build GRCh38
rs202207944 Chr.7: 111736963-111736963 on Build GRCh38
rs917401202 Chr.7: 111737182-111737182 on Build GRCh38
rs530320673 Chr.7: 111737185-111737185 on Build GRCh38
rs903154184 Chr.7: 111736871-111736871 on Build GRCh38
rs745783701 Chr.7: 111736883-111736883 on Build GRCh38
rs895996422 Chr.7: 111736823-111736823 on Build GRCh38
rs924670181 Chr.7: 111737200-111737200 on Build GRCh38
rs145638553 Chr.7: 111736848-111736848 on Build GRCh38
rs750781177 Chr.7: 111737005-111737006 on Build GRCh38
rs889027290 Chr.7: 111736937-111736937 on Build GRCh38
rs547741055 Chr.7: 111737108-111737110 on Build GRCh38
rs984861831 Chr.7: 111737069-111737069 on Build GRCh38
rs775902528 Chr.7: 111736895-111736895 on Build GRCh38
rs1045860191 Chr.7: 111737154-111737154 on Build GRCh38
rs550995671 Chr.7: 111737156-111737156 on Build GRCh38
rs201048359 Chr.7: 111737030-111737030 on Build GRCh38
rs759316413 Chr.7: 111736817-111736817 on Build GRCh38
rs926345304 Chr.7: 111737099-111737099 on Build GRCh38
rs766914725 Chr.7: 111736962-111736962 on Build GRCh38
rs377250270 Chr.7: 111736854-111736854 on Build GRCh38

View Full Product Details