Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 111741458-111741965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK4
Entrez Gene ID
9732
Gene Name
dedicator of cytokinesis 4
Gene Aliases
-
UniGene
Hs.654652
Gene Chromosome Location
Chr.7: 111726108-112206407 on Build GRCh38
Transcripts
NM_014705.3 XM_006716188.2 XM_006716189.2 XM_017012819.1 XM_017012820.1 XM_017012821.1 XM_017012822.1 XM_017012823.1 XM_017012824.1 XM_017012825.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5721047 c.1466G>A p.G489E Chr.7: 111741615-111741615 on Build GRCh38
5463581 c.4891A>G p.S1631G Chr.7: 111741541-111741541 on Build GRCh38
5721045 c.4817G>A p.G1606E Chr.7: 111741615-111741615 on Build GRCh38
3631950 c.4821G>T p.V1607V Chr.7: 111741602-111741602 on Build GRCh38
5463582 c.4882A>G p.S1628G Chr.7: 111741541-111741541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185317060 Chr.7: 111741712-111741712 on Build GRCh38
rs747984091 Chr.7: 111741599-111741599 on Build GRCh38
rs771997046 Chr.7: 111741517-111741517 on Build GRCh38
rs765468990 Chr.7: 111741543-111741543 on Build GRCh38
rs559505178 Chr.7: 111741920-111741920 on Build GRCh38
rs746988311 Chr.7: 111741606-111741606 on Build GRCh38
rs752598274 Chr.7: 111741534-111741534 on Build GRCh38
rs778011503 Chr.7: 111741721-111741721 on Build GRCh38
rs755684621 Chr.7: 111741571-111741571 on Build GRCh38
rs992698243 Chr.7: 111741629-111741629 on Build GRCh38
rs549588731 Chr.7: 111741561-111741561 on Build GRCh38
rs919738797 Chr.7: 111741642-111741642 on Build GRCh38
rs888467444 Chr.7: 111741910-111741910 on Build GRCh38
rs565634053 Chr.7: 111741675-111741675 on Build GRCh38
rs769031094 Chr.7: 111741683-111741683 on Build GRCh38
rs760727286 Chr.7: 111741520-111741520 on Build GRCh38
rs1029576813 Chr.7: 111741848-111741848 on Build GRCh38
rs765502490 Chr.7: 111741619-111741619 on Build GRCh38
rs531092850 Chr.7: 111741513-111741513 on Build GRCh38
rs376606033 Chr.7: 111741696-111741696 on Build GRCh38
rs747819161 Chr.7: 111741701-111741701 on Build GRCh38
rs996324795 Chr.7: 111741844-111741844 on Build GRCh38
rs555139274 Chr.7: 111741754-111741754 on Build GRCh38
rs530781849 Chr.7: 111741524-111741524 on Build GRCh38
rs769514683 Chr.7: 111741500-111741500 on Build GRCh38
rs181220597 Chr.7: 111741660-111741660 on Build GRCh38
rs568006198 Chr.7: 111741581-111741581 on Build GRCh38
rs904104103 Chr.7: 111741486-111741486 on Build GRCh38
rs746899129 Chr.7: 111741706-111741706 on Build GRCh38
rs779843119 Chr.7: 111741501-111741501 on Build GRCh38
rs115045507 Chr.7: 111741489-111741489 on Build GRCh38
rs757450490 Chr.7: 111741544-111741544 on Build GRCh38
rs778501945 Chr.7: 111741596-111741596 on Build GRCh38
rs543256843 Chr.7: 111741747-111741747 on Build GRCh38
rs953816659 Chr.7: 111741892-111741892 on Build GRCh38
rs377699698 Chr.7: 111741499-111741499 on Build GRCh38
rs371201135 Chr.7: 111741512-111741512 on Build GRCh38
rs912267971 Chr.7: 111741594-111741594 on Build GRCh38
rs916636320 Chr.7: 111741682-111741682 on Build GRCh38
rs936898487 Chr.7: 111741720-111741720 on Build GRCh38
rs573626290 Chr.7: 111741809-111741809 on Build GRCh38
rs191952903 Chr.7: 111741733-111741733 on Build GRCh38
rs764622040 Chr.7: 111741669-111741669 on Build GRCh38
rs766511218 Chr.7: 111741523-111741523 on Build GRCh38
rs890063374 Chr.7: 111741734-111741734 on Build GRCh38
rs770886821 Chr.7: 111741707-111741707 on Build GRCh38
rs773216903 Chr.7: 111741519-111741519 on Build GRCh38
rs994273686 Chr.7: 111741875-111741875 on Build GRCh38
rs775660558 Chr.7: 111741625-111741625 on Build GRCh38
rs889969337 Chr.7: 111741507-111741507 on Build GRCh38
rs897225926 Chr.7: 111741874-111741874 on Build GRCh38
rs764713923 Chr.7: 111741567-111741567 on Build GRCh38
rs1055382899 Chr.7: 111741676-111741676 on Build GRCh38
rs540913870 Chr.7: 111741867-111741867 on Build GRCh38
rs765761389 Chr.7: 111741686-111741686 on Build GRCh38
rs779753390 Chr.7: 111741573-111741573 on Build GRCh38
rs1032504925 Chr.7: 111741516-111741516 on Build GRCh38
rs1041256152 Chr.7: 111741853-111741853 on Build GRCh38
rs761611515 Chr.7: 111741514-111741514 on Build GRCh38
rs1048387530 Chr.7: 111741900-111741900 on Build GRCh38
rs138174084 Chr.7: 111741703-111741703 on Build GRCh38
rs570156858 Chr.7: 111741498-111741498 on Build GRCh38
rs867557702 Chr.7: 111741622-111741622 on Build GRCh38
rs771910846 Chr.7: 111741602-111741602 on Build GRCh38
rs759637184 Chr.7: 111741616-111741616 on Build GRCh38
rs749038740 Chr.7: 111741502-111741502 on Build GRCh38
rs770974755 Chr.7: 111741607-111741607 on Build GRCh38
rs776794984 Chr.7: 111741609-111741609 on Build GRCh38
rs535311309 Chr.7: 111741591-111741591 on Build GRCh38
rs758156487 Chr.7: 111741702-111741702 on Build GRCh38
rs752171917 Chr.7: 111741569-111741569 on Build GRCh38
rs754526204 Chr.7: 111741699-111741699 on Build GRCh38

View Full Product Details