Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122078777-122079272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASS
Entrez Gene ID
10157
Gene Name
aminoadipate-semialdehyde synthase
Gene Aliases
LKR/SDH, LKRSDH, LORSDH
UniGene
Hs.156738
Gene Chromosome Location
Chr.7: 122073544-122144290 on Build GRCh38
Transcripts
NM_005763.3 XM_011515725.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5036069 c.2421A>C p.Q807H Chr.7: 122078926-122078926 on Build GRCh38
2766866 c.2485G>T p.G829C Chr.7: 122078862-122078862 on Build GRCh38
2766868 c.2436G>A p.E812E Chr.7: 122078911-122078911 on Build GRCh38
4480582 c.2432C>T p.A811V Chr.7: 122078915-122078915 on Build GRCh38
5749912 c.2465T>A p.L822* Chr.7: 122078882-122078882 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879057473 Chr.7: 122078927-122078927 on Build GRCh38
rs374226696 Chr.7: 122078868-122078868 on Build GRCh38
rs558634023 Chr.7: 122078799-122078799 on Build GRCh38
rs1006487502 Chr.7: 122079239-122079241 on Build GRCh38
rs762188159 Chr.7: 122078921-122078921 on Build GRCh38
rs780396059 Chr.7: 122078827-122078827 on Build GRCh38
rs368886192 Chr.7: 122078999-122078999 on Build GRCh38
rs972093374 Chr.7: 122079099-122079099 on Build GRCh38
rs180824698 Chr.7: 122078966-122078966 on Build GRCh38
rs776455006 Chr.7: 122078871-122078871 on Build GRCh38
rs765465312 Chr.7: 122079115-122079115 on Build GRCh38
rs958843516 Chr.7: 122079002-122079002 on Build GRCh38
rs950981265 Chr.7: 122078928-122078928 on Build GRCh38
rs773886567 Chr.7: 122078963-122078963 on Build GRCh38
rs943322786 Chr.7: 122079059-122079059 on Build GRCh38
rs757491991 Chr.7: 122078816-122078816 on Build GRCh38
rs903506271 Chr.7: 122078844-122078845 on Build GRCh38
rs750249378 Chr.7: 122079223-122079223 on Build GRCh38
rs577129335 Chr.7: 122078835-122078835 on Build GRCh38
rs1027015345 Chr.7: 122078919-122078919 on Build GRCh38
rs568131802 Chr.7: 122079219-122079219 on Build GRCh38
rs368777484 Chr.7: 122078910-122078910 on Build GRCh38
rs144721391 Chr.7: 122079233-122079233 on Build GRCh38
rs910736205 Chr.7: 122079210-122079210 on Build GRCh38
rs531319319 Chr.7: 122079206-122079206 on Build GRCh38
rs575593407 Chr.7: 122078800-122078800 on Build GRCh38
rs1001788326 Chr.7: 122079194-122079194 on Build GRCh38
rs73441002 Chr.7: 122078888-122078888 on Build GRCh38
rs574621404 Chr.7: 122078879-122078879 on Build GRCh38
rs750804318 Chr.7: 122078822-122078822 on Build GRCh38
rs777196791 Chr.7: 122078854-122078854 on Build GRCh38
rs892402401 Chr.7: 122079124-122079124 on Build GRCh38
rs762791713 Chr.7: 122079243-122079243 on Build GRCh38
rs190997123 Chr.7: 122079245-122079245 on Build GRCh38
rs750859178 Chr.7: 122078947-122078947 on Build GRCh38
rs17144072 Chr.7: 122078901-122078901 on Build GRCh38
rs746729656 Chr.7: 122078855-122078855 on Build GRCh38
rs764089104 Chr.7: 122078902-122078902 on Build GRCh38
rs767826019 Chr.7: 122078930-122078930 on Build GRCh38
rs745616346 Chr.7: 122078872-122078872 on Build GRCh38
rs532649293 Chr.7: 122079068-122079068 on Build GRCh38
rs1044008061 Chr.7: 122079185-122079185 on Build GRCh38
rs186758179 Chr.7: 122079102-122079102 on Build GRCh38
rs950922807 Chr.7: 122079234-122079234 on Build GRCh38
rs1021943908 Chr.7: 122078834-122078834 on Build GRCh38
rs1049684309 Chr.7: 122079149-122079149 on Build GRCh38
rs997802251 Chr.7: 122079227-122079227 on Build GRCh38
rs991440692 Chr.7: 122079019-122079019 on Build GRCh38
rs770557763 Chr.7: 122078857-122078857 on Build GRCh38
rs544322668 Chr.7: 122078812-122078812 on Build GRCh38
rs774571411 Chr.7: 122078907-122078907 on Build GRCh38
rs917217563 Chr.7: 122079026-122079026 on Build GRCh38
rs751129137 Chr.7: 122078953-122078954 on Build GRCh38
rs372473763 Chr.7: 122078934-122078934 on Build GRCh38
rs755504116 Chr.7: 122078971-122078971 on Build GRCh38
rs768065768 Chr.7: 122078821-122078821 on Build GRCh38
rs903539246 Chr.7: 122079189-122079189 on Build GRCh38
rs757857246 Chr.7: 122078849-122078849 on Build GRCh38
rs971396236 Chr.7: 122079077-122079077 on Build GRCh38
rs563078369 Chr.7: 122079109-122079109 on Build GRCh38
rs746500410 Chr.7: 122078990-122078990 on Build GRCh38
rs777321532 Chr.7: 122078985-122078985 on Build GRCh38
rs548234015 Chr.7: 122079218-122079218 on Build GRCh38
rs541318195 Chr.7: 122079000-122079000 on Build GRCh38

View Full Product Details