Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122081414-122081921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASS
Entrez Gene ID
10157
Gene Name
aminoadipate-semialdehyde synthase
Gene Aliases
LKR/SDH, LKRSDH, LORSDH
UniGene
Hs.156738
Gene Chromosome Location
Chr.7: 122073544-122144290 on Build GRCh38
Transcripts
NM_005763.3 XM_011515725.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5731997 c.2236G>T p.E746* Chr.7: 122081544-122081544 on Build GRCh38
6846011 c.2228T>C p.I743T Chr.7: 122081552-122081552 on Build GRCh38
3366747 c.2219T>A p.L740* Chr.7: 122081561-122081561 on Build GRCh38
7428998 c.2256A>T p.R752S Chr.7: 122081524-122081524 on Build GRCh38
278490 c.2241G>A p.A747A Chr.7: 122081539-122081539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963183956 Chr.7: 122081440-122081440 on Build GRCh38
rs957574173 Chr.7: 122081784-122081784 on Build GRCh38
rs764949013 Chr.7: 122081632-122081632 on Build GRCh38
rs768862108 Chr.7: 122081611-122081611 on Build GRCh38
rs775829568 Chr.7: 122081499-122081499 on Build GRCh38
rs930421858 Chr.7: 122081684-122081684 on Build GRCh38
rs758480052 Chr.7: 122081454-122081454 on Build GRCh38
rs1050372096 Chr.7: 122081706-122081706 on Build GRCh38
rs961642217 Chr.7: 122081878-122081878 on Build GRCh38
rs150922740 Chr.7: 122081504-122081504 on Build GRCh38
rs139475283 Chr.7: 122081521-122081521 on Build GRCh38
rs368322844 Chr.7: 122081605-122081605 on Build GRCh38
rs777793974 Chr.7: 122081568-122081568 on Build GRCh38
rs746871208 Chr.7: 122081572-122081572 on Build GRCh38
rs772638222 Chr.7: 122081510-122081510 on Build GRCh38
rs752353924 Chr.7: 122081645-122081645 on Build GRCh38
rs28405443 Chr.7: 122081829-122081829 on Build GRCh38
rs982079004 Chr.7: 122081823-122081823 on Build GRCh38
rs151124568 Chr.7: 122081625-122081625 on Build GRCh38
rs770424300 Chr.7: 122081609-122081610 on Build GRCh38
rs752515450 Chr.7: 122081550-122081550 on Build GRCh38
rs994771422 Chr.7: 122081451-122081451 on Build GRCh38
rs754612868 Chr.7: 122081539-122081539 on Build GRCh38
rs1015960543 Chr.7: 122081885-122081885 on Build GRCh38
rs1045231896 Chr.7: 122081746-122081746 on Build GRCh38
rs1025893507 Chr.7: 122081820-122081820 on Build GRCh38
rs758445857 Chr.7: 122081556-122081556 on Build GRCh38
rs530294885 Chr.7: 122081621-122081621 on Build GRCh38
rs902824198 Chr.7: 122081737-122081737 on Build GRCh38
rs753561492 Chr.7: 122081522-122081522 on Build GRCh38
rs940140907 Chr.7: 122081666-122081666 on Build GRCh38
rs995902382 Chr.7: 122081887-122081887 on Build GRCh38
rs781181917 Chr.7: 122081875-122081875 on Build GRCh38
rs888785629 Chr.7: 122081475-122081475 on Build GRCh38
rs893868140 Chr.7: 122081634-122081634 on Build GRCh38
rs889028675 Chr.7: 122081711-122081711 on Build GRCh38
rs746153221 Chr.7: 122081588-122081588 on Build GRCh38
rs1206370 Chr.7: 122081801-122081801 on Build GRCh38
rs543148580 Chr.7: 122081439-122081439 on Build GRCh38
rs200806638 Chr.7: 122081617-122081617 on Build GRCh38
rs563437450 Chr.7: 122081487-122081487 on Build GRCh38
rs1016425487 Chr.7: 122081438-122081438 on Build GRCh38
rs548792596 Chr.7: 122081518-122081518 on Build GRCh38
rs550001308 Chr.7: 122081723-122081723 on Build GRCh38
rs35726770 Chr.7: 122081453-122081454 on Build GRCh38
rs372679186 Chr.7: 122081540-122081540 on Build GRCh38
rs1038436298 Chr.7: 122081669-122081669 on Build GRCh38
rs535017358 Chr.7: 122081890-122081890 on Build GRCh38
rs751019547 Chr.7: 122081861-122081861 on Build GRCh38
rs760267836 Chr.7: 122081513-122081513 on Build GRCh38
rs570198982 Chr.7: 122081748-122081748 on Build GRCh38
rs192031195 Chr.7: 122081886-122081886 on Build GRCh38
rs1032683934 Chr.7: 122081765-122081766 on Build GRCh38
rs933216316 Chr.7: 122081489-122081489 on Build GRCh38
rs374884203 Chr.7: 122081481-122081481 on Build GRCh38
rs770875442 Chr.7: 122081480-122081480 on Build GRCh38
rs748161288 Chr.7: 122081449-122081449 on Build GRCh38
rs751633998 Chr.7: 122081731-122081731 on Build GRCh38
rs781064687 Chr.7: 122081583-122081583 on Build GRCh38
rs780765810 Chr.7: 122081627-122081628 on Build GRCh38
rs906701360 Chr.7: 122081755-122081755 on Build GRCh38
rs769991420 Chr.7: 122081597-122081597 on Build GRCh38
rs775882678 Chr.7: 122081599-122081599 on Build GRCh38
rs372939523 Chr.7: 122081627-122081627 on Build GRCh38
rs757370522 Chr.7: 122081578-122081578 on Build GRCh38
rs199606844 Chr.7: 122081801-122081801 on Build GRCh38
rs369222602 Chr.7: 122081503-122081503 on Build GRCh38
rs772120908 Chr.7: 122081453-122081453 on Build GRCh38
rs770146974 Chr.7: 122081490-122081490 on Build GRCh38
rs899004473 Chr.7: 122081670-122081670 on Build GRCh38
rs141097099 Chr.7: 122081626-122081626 on Build GRCh38
rs1023203398 Chr.7: 122081816-122081816 on Build GRCh38

View Full Product Details