Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127582510-127583001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCC1
Entrez Gene ID
79571
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
GCC1P, GCC88
UniGene
Hs.521168
Gene Chromosome Location
Chr.7: 127580628-127585600 on Build GRCh38
Transcripts
NM_024523.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1447846 c.1562A>T p.Q521L Chr.7: 127582780-127582780 on Build GRCh38
395363 c.1395G>T p.M465I Chr.7: 127582947-127582947 on Build GRCh38
6368335 c.1640A>G p.Q547R Chr.7: 127582702-127582702 on Build GRCh38
5447750 c.1570C>T p.L524F Chr.7: 127582772-127582772 on Build GRCh38
2768543 c.1470A>G p.E490E Chr.7: 127582872-127582872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367564453 Chr.7: 127582544-127582544 on Build GRCh38
rs1055813194 Chr.7: 127582577-127582577 on Build GRCh38
rs369520519 Chr.7: 127582913-127582913 on Build GRCh38
rs773169241 Chr.7: 127582533-127582533 on Build GRCh38
rs111491677 Chr.7: 127582960-127582960 on Build GRCh38
rs758362532 Chr.7: 127582967-127582967 on Build GRCh38
rs778809732 Chr.7: 127582703-127582703 on Build GRCh38
rs190536068 Chr.7: 127582767-127582767 on Build GRCh38
rs372791694 Chr.7: 127582592-127582592 on Build GRCh38
rs764589858 Chr.7: 127582779-127582779 on Build GRCh38
rs368859467 Chr.7: 127582946-127582946 on Build GRCh38
rs754967267 Chr.7: 127582684-127582684 on Build GRCh38
rs755684526 Chr.7: 127582669-127582669 on Build GRCh38
rs865891541 Chr.7: 127582682-127582682 on Build GRCh38
rs757501854 Chr.7: 127582853-127582853 on Build GRCh38
rs779765798 Chr.7: 127582670-127582670 on Build GRCh38
rs181796106 Chr.7: 127582905-127582905 on Build GRCh38
rs201718169 Chr.7: 127582578-127582578 on Build GRCh38
rs753035232 Chr.7: 127582652-127582652 on Build GRCh38
rs376149635 Chr.7: 127582637-127582637 on Build GRCh38
rs545815817 Chr.7: 127582566-127582566 on Build GRCh38
rs770942238 Chr.7: 127582730-127582730 on Build GRCh38
rs765916895 Chr.7: 127582791-127582791 on Build GRCh38
rs771038043 Chr.7: 127582635-127582635 on Build GRCh38
rs774780646 Chr.7: 127582938-127582938 on Build GRCh38
rs763757405 Chr.7: 127582973-127582973 on Build GRCh38
rs747315951 Chr.7: 127582729-127582729 on Build GRCh38
rs780111481 Chr.7: 127582589-127582589 on Build GRCh38
rs999024126 Chr.7: 127582758-127582758 on Build GRCh38
rs151126177 Chr.7: 127582727-127582727 on Build GRCh38
rs767571267 Chr.7: 127582530-127582530 on Build GRCh38
rs766893283 Chr.7: 127582541-127582541 on Build GRCh38
rs765520352 Chr.7: 127582651-127582651 on Build GRCh38
rs141040379 Chr.7: 127582749-127582749 on Build GRCh38
rs753426587 Chr.7: 127582806-127582806 on Build GRCh38
rs150257982 Chr.7: 127582903-127582903 on Build GRCh38
rs747917255 Chr.7: 127582710-127582710 on Build GRCh38
rs867976386 Chr.7: 127582693-127582693 on Build GRCh38
rs754913190 Chr.7: 127582810-127582810 on Build GRCh38
rs761482078 Chr.7: 127582654-127582654 on Build GRCh38
rs765062755 Chr.7: 127582818-127582818 on Build GRCh38
rs760647290 Chr.7: 127582629-127582629 on Build GRCh38
rs767058616 Chr.7: 127582656-127582656 on Build GRCh38
rs751154961 Chr.7: 127582568-127582568 on Build GRCh38
rs34469388 Chr.7: 127582741-127582742 on Build GRCh38
rs374524039 Chr.7: 127582612-127582612 on Build GRCh38
rs781200946 Chr.7: 127582854-127582854 on Build GRCh38
rs199928954 Chr.7: 127582959-127582959 on Build GRCh38
rs760571264 Chr.7: 127582947-127582947 on Build GRCh38
rs772723890 Chr.7: 127582786-127582786 on Build GRCh38
rs774860737 Chr.7: 127582551-127582551 on Build GRCh38
rs753767197 Chr.7: 127582683-127582683 on Build GRCh38
rs777638792 Chr.7: 127582845-127582845 on Build GRCh38
rs773293270 Chr.7: 127582619-127582619 on Build GRCh38
rs776697442 Chr.7: 127582732-127582732 on Build GRCh38
rs965536496 Chr.7: 127582793-127582793 on Build GRCh38
rs1007870257 Chr.7: 127582898-127582898 on Build GRCh38
rs764868196 Chr.7: 127582952-127582952 on Build GRCh38
rs35125522 Chr.7: 127582549-127582549 on Build GRCh38
rs201063771 Chr.7: 127582608-127582608 on Build GRCh38
rs756864303 Chr.7: 127582570-127582570 on Build GRCh38
rs371600897 Chr.7: 127582942-127582942 on Build GRCh38
rs147776017 Chr.7: 127582571-127582571 on Build GRCh38
rs745928948 Chr.7: 127582866-127582866 on Build GRCh38
rs549907133 Chr.7: 127582613-127582613 on Build GRCh38
rs759997521 Chr.7: 127582739-127582739 on Build GRCh38
rs766151079 Chr.7: 127582941-127582941 on Build GRCh38
rs760360160 Chr.7: 127582939-127582939 on Build GRCh38
rs903462687 Chr.7: 127582757-127582757 on Build GRCh38
rs138754679 Chr.7: 127582908-127582908 on Build GRCh38
rs147109145 Chr.7: 127582665-127582665 on Build GRCh38
rs141830030 Chr.7: 127582659-127582659 on Build GRCh38
rs760415411 Chr.7: 127582790-127582790 on Build GRCh38
rs1043638831 Chr.7: 127582673-127582673 on Build GRCh38
rs776118131 Chr.7: 127582756-127582756 on Build GRCh38
rs755702113 Chr.7: 127582576-127582576 on Build GRCh38
rs752718782 Chr.7: 127582819-127582819 on Build GRCh38
rs777111161 Chr.7: 127582636-127582636 on Build GRCh38
rs562333991 Chr.7: 127582717-127582717 on Build GRCh38
rs142108786 Chr.7: 127582922-127582922 on Build GRCh38
rs758122643 Chr.7: 127582841-127582841 on Build GRCh38
rs747168639 Chr.7: 127582846-127582846 on Build GRCh38
rs140406043 Chr.7: 127582535-127582535 on Build GRCh38

View Full Product Details