Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127584449-127584955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCC1
Entrez Gene ID
79571
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
GCC1P, GCC88
UniGene
Hs.521168
Gene Chromosome Location
Chr.7: 127580628-127585600 on Build GRCh38
Transcripts
NM_024523.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6319690 c.680C>A p.A227D Chr.7: 127584503-127584503 on Build GRCh38
5406429 c.317G>A p.S106N Chr.7: 127584866-127584866 on Build GRCh38
744681 c.452G>T p.G151V Chr.7: 127584731-127584731 on Build GRCh38
4139328 c.435G>T p.G145G Chr.7: 127584748-127584748 on Build GRCh38
4944711 c.593A>G p.E198G Chr.7: 127584590-127584590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765693208 Chr.7: 127584493-127584493 on Build GRCh38
rs759381858 Chr.7: 127584742-127584742 on Build GRCh38
rs985860954 Chr.7: 127584573-127584573 on Build GRCh38
rs960850309 Chr.7: 127584728-127584728 on Build GRCh38
rs142380869 Chr.7: 127584572-127584572 on Build GRCh38
rs754945493 Chr.7: 127584677-127584677 on Build GRCh38
rs577776580 Chr.7: 127584882-127584882 on Build GRCh38
rs867484826 Chr.7: 127584639-127584639 on Build GRCh38
rs2285348 Chr.7: 127584922-127584922 on Build GRCh38
rs151303849 Chr.7: 127584708-127584708 on Build GRCh38
rs765198057 Chr.7: 127584799-127584799 on Build GRCh38
rs138293120 Chr.7: 127584891-127584892 on Build GRCh38
rs757504084 Chr.7: 127584819-127584819 on Build GRCh38
rs767098451 Chr.7: 127584536-127584536 on Build GRCh38
rs372115883 Chr.7: 127584490-127584490 on Build GRCh38
rs375812885 Chr.7: 127584696-127584696 on Build GRCh38
rs560808114 Chr.7: 127584926-127584926 on Build GRCh38
rs773819474 Chr.7: 127584499-127584499 on Build GRCh38
rs760254098 Chr.7: 127584654-127584654 on Build GRCh38
rs1043875258 Chr.7: 127584814-127584814 on Build GRCh38
rs781338488 Chr.7: 127584732-127584732 on Build GRCh38
rs759341081 Chr.7: 127584790-127584790 on Build GRCh38
rs753658097 Chr.7: 127584674-127584674 on Build GRCh38
rs769081096 Chr.7: 127584885-127584885 on Build GRCh38
rs1044536202 Chr.7: 127584517-127584517 on Build GRCh38
rs771449194 Chr.7: 127584482-127584482 on Build GRCh38
rs767640305 Chr.7: 127584707-127584708 on Build GRCh38
rs755738074 Chr.7: 127584541-127584541 on Build GRCh38
rs765271840 Chr.7: 127584912-127584913 on Build GRCh38
rs771305960 Chr.7: 127584619-127584619 on Build GRCh38
rs763891408 Chr.7: 127584816-127584816 on Build GRCh38
rs779421781 Chr.7: 127584891-127584891 on Build GRCh38
rs781086950 Chr.7: 127584820-127584820 on Build GRCh38
rs374351066 Chr.7: 127584863-127584863 on Build GRCh38
rs762723715 Chr.7: 127584808-127584808 on Build GRCh38
rs760986634 Chr.7: 127584581-127584581 on Build GRCh38
rs1030936147 Chr.7: 127584514-127584514 on Build GRCh38
rs770358676 Chr.7: 127584757-127584757 on Build GRCh38
rs751381399 Chr.7: 127584928-127584928 on Build GRCh38
rs777111047 Chr.7: 127584593-127584593 on Build GRCh38
rs140583068 Chr.7: 127584785-127584785 on Build GRCh38
rs752559290 Chr.7: 127584692-127584692 on Build GRCh38
rs750497246 Chr.7: 127584871-127584871 on Build GRCh38
rs747068348 Chr.7: 127584586-127584586 on Build GRCh38
rs200284347 Chr.7: 127584555-127584555 on Build GRCh38
rs746618070 Chr.7: 127584906-127584906 on Build GRCh38
rs1000402305 Chr.7: 127584548-127584548 on Build GRCh38
rs1008800986 Chr.7: 127584597-127584597 on Build GRCh38
rs764900383 Chr.7: 127584685-127584685 on Build GRCh38
rs760413694 Chr.7: 127584768-127584768 on Build GRCh38
rs35322201 Chr.7: 127584818-127584818 on Build GRCh38
rs765563695 Chr.7: 127584910-127584910 on Build GRCh38
rs367828009 Chr.7: 127584551-127584551 on Build GRCh38
rs746401983 Chr.7: 127584594-127584594 on Build GRCh38
rs776234489 Chr.7: 127584781-127584781 on Build GRCh38
rs752616296 Chr.7: 127584807-127584807 on Build GRCh38
rs769989565 Chr.7: 127584596-127584596 on Build GRCh38
rs375254345 Chr.7: 127584523-127584523 on Build GRCh38
rs773411272 Chr.7: 127584725-127584745 on Build GRCh38
rs757271302 Chr.7: 127584717-127584717 on Build GRCh38
rs17151044 Chr.7: 127584927-127584927 on Build GRCh38
rs771066838 Chr.7: 127584587-127584587 on Build GRCh38
rs761327169 Chr.7: 127584599-127584599 on Build GRCh38
rs916953922 Chr.7: 127584851-127584851 on Build GRCh38
rs749978587 Chr.7: 127584540-127584540 on Build GRCh38
rs776194716 Chr.7: 127584917-127584917 on Build GRCh38
rs775441390 Chr.7: 127584923-127584923 on Build GRCh38
rs1043439610 Chr.7: 127584480-127584480 on Build GRCh38
rs370013935 Chr.7: 127584750-127584750 on Build GRCh38
rs773035632 Chr.7: 127584631-127584631 on Build GRCh38
rs962691156 Chr.7: 127584649-127584649 on Build GRCh38
rs903524738 Chr.7: 127584502-127584502 on Build GRCh38
rs778780245 Chr.7: 127584571-127584571 on Build GRCh38
rs952252130 Chr.7: 127584749-127584749 on Build GRCh38
rs201415172 Chr.7: 127584777-127584777 on Build GRCh38
rs780478029 Chr.7: 127584761-127584761 on Build GRCh38
rs999409947 Chr.7: 127584509-127584509 on Build GRCh38
rs780602930 Chr.7: 127584875-127584875 on Build GRCh38
rs918865874 Chr.7: 127584662-127584662 on Build GRCh38
rs776777500 Chr.7: 127584765-127584765 on Build GRCh38
rs767260147 Chr.7: 127584628-127584628 on Build GRCh38
rs749637521 Chr.7: 127584764-127584764 on Build GRCh38
rs766042503 Chr.7: 127584667-127584667 on Build GRCh38
rs201944678 Chr.7: 127584739-127584739 on Build GRCh38
rs960976185 Chr.7: 127584803-127584803 on Build GRCh38
rs772436091 Chr.7: 127584554-127584554 on Build GRCh38

View Full Product Details