Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151176323-151176742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB10
Entrez Gene ID
136371
Gene Name
ankyrin repeat and SOCS box containing 10
Gene Aliases
GLC1F
UniGene
Hs.647081
Gene Chromosome Location
Chr.7: 151175698-151187832 on Build GRCh38
Transcripts
NM_001142459.1 NM_001142460.1 NM_080871.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3762499 c.1114C>T p.R372C Chr.7: 151176667-151176667 on Build GRCh38
3762501 c.1249C>T p.R417C Chr.7: 151176667-151176667 on Build GRCh38
4004380 c.1267C>T p.R423W Chr.7: 151176649-151176649 on Build GRCh38
4004378 c.1132C>T p.R378W Chr.7: 151176649-151176649 on Build GRCh38
318793 c.1126T>A p.C376S Chr.7: 151176655-151176655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780083830 Chr.7: 151176660-151176660 on Build GRCh38
rs310598 Chr.7: 151176521-151176521 on Build GRCh38
rs973068070 Chr.7: 151176643-151176643 on Build GRCh38
rs912069244 Chr.7: 151176666-151176666 on Build GRCh38
rs539514085 Chr.7: 151176492-151176492 on Build GRCh38
rs1004114604 Chr.7: 151176487-151176487 on Build GRCh38
rs948354271 Chr.7: 151176454-151176454 on Build GRCh38
rs915667502 Chr.7: 151176450-151176450 on Build GRCh38
rs1052427442 Chr.7: 151176504-151176504 on Build GRCh38
rs764867865 Chr.7: 151176366-151176366 on Build GRCh38
rs189183340 Chr.7: 151176570-151176570 on Build GRCh38
rs73478620 Chr.7: 151176537-151176537 on Build GRCh38
rs920855248 Chr.7: 151176416-151176416 on Build GRCh38
rs367834189 Chr.7: 151176686-151176686 on Build GRCh38
rs546982480 Chr.7: 151176704-151176704 on Build GRCh38
rs554773033 Chr.7: 151176388-151176388 on Build GRCh38
rs191838657 Chr.7: 151176347-151176347 on Build GRCh38
rs973687404 Chr.7: 151176414-151176414 on Build GRCh38
rs771203361 Chr.7: 151176565-151176565 on Build GRCh38
rs1018885291 Chr.7: 151176527-151176527 on Build GRCh38
rs770356435 Chr.7: 151176592-151176592 on Build GRCh38
rs533844799 Chr.7: 151176413-151176413 on Build GRCh38
rs753462958 Chr.7: 151176590-151176590 on Build GRCh38
rs919533 Chr.7: 151176577-151176577 on Build GRCh38
rs766958974 Chr.7: 151176648-151176648 on Build GRCh38
rs977669835 Chr.7: 151176389-151176389 on Build GRCh38
rs995367980 Chr.7: 151176509-151176509 on Build GRCh38
rs1034061352 Chr.7: 151176376-151176376 on Build GRCh38
rs1036978559 Chr.7: 151176488-151176488 on Build GRCh38
rs754547437 Chr.7: 151176640-151176640 on Build GRCh38
rs4726004 Chr.7: 151176479-151176479 on Build GRCh38
rs764552895 Chr.7: 151176511-151176511 on Build GRCh38
rs778515347 Chr.7: 151176650-151176650 on Build GRCh38
rs532441181 Chr.7: 151176696-151176696 on Build GRCh38
rs992494697 Chr.7: 151176394-151176394 on Build GRCh38
rs1045395097 Chr.7: 151176455-151176455 on Build GRCh38
rs763403427 Chr.7: 151176510-151176510 on Build GRCh38
rs898302033 Chr.7: 151176497-151176497 on Build GRCh38
rs61628535 Chr.7: 151176649-151176649 on Build GRCh38
rs763697610 Chr.7: 151176615-151176616 on Build GRCh38
rs62489646 Chr.7: 151176667-151176667 on Build GRCh38
rs763709556 Chr.7: 151176619-151176619 on Build GRCh38
rs936188600 Chr.7: 151176403-151176403 on Build GRCh38
rs904449951 Chr.7: 151176526-151176526 on Build GRCh38
rs968553846 Chr.7: 151176358-151176358 on Build GRCh38
rs567980404 Chr.7: 151176395-151176395 on Build GRCh38
rs371108358 Chr.7: 151176601-151176601 on Build GRCh38
rs184479700 Chr.7: 151176380-151176380 on Build GRCh38
rs959800416 Chr.7: 151176613-151176613 on Build GRCh38
rs564809133 Chr.7: 151176641-151176641 on Build GRCh38
rs771290995 Chr.7: 151176714-151176714 on Build GRCh38
rs373724579 Chr.7: 151176657-151176657 on Build GRCh38
rs553307306 Chr.7: 151176449-151176449 on Build GRCh38
rs977615907 Chr.7: 151176549-151176549 on Build GRCh38
rs747074111 Chr.7: 151176554-151176554 on Build GRCh38
rs763412421 Chr.7: 151176602-151176602 on Build GRCh38
rs944815220 Chr.7: 151176493-151176493 on Build GRCh38

View Full Product Details