Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155457695-155458200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EN2
Entrez Gene ID
2020
Gene Name
engrailed homeobox 2
Gene Aliases
-
UniGene
Hs.134989
Gene Chromosome Location
Chr.7: 155458129-155464831 on Build GRCh38
Transcripts
NM_001427.3

SNP Detail

SNP ID Chromosome Location
rs980011875 Chr.7: 155457963-155457963 on Build GRCh38
rs1032453795 Chr.7: 155458152-155458152 on Build GRCh38
rs1052378299 Chr.7: 155457796-155457796 on Build GRCh38
rs925494577 Chr.7: 155457973-155457973 on Build GRCh38
rs1056921676 Chr.7: 155458079-155458079 on Build GRCh38
rs1030379691 Chr.7: 155458158-155458158 on Build GRCh38
rs1032151965 Chr.7: 155457741-155457762 on Build GRCh38
rs906440612 Chr.7: 155457762-155457762 on Build GRCh38
rs897574120 Chr.7: 155457887-155457887 on Build GRCh38
rs933329777 Chr.7: 155458002-155458002 on Build GRCh38
rs377394985 Chr.7: 155458127-155458127 on Build GRCh38
rs1013006363 Chr.7: 155458091-155458091 on Build GRCh38
rs933219450 Chr.7: 155457826-155457826 on Build GRCh38
rs969551164 Chr.7: 155457961-155457961 on Build GRCh38
rs542019421 Chr.7: 155458037-155458037 on Build GRCh38
rs923303185 Chr.7: 155457813-155457813 on Build GRCh38
rs1003909574 Chr.7: 155457825-155457825 on Build GRCh38
rs1037929151 Chr.7: 155458025-155458025 on Build GRCh38
rs957953696 Chr.7: 155457886-155457887 on Build GRCh38
rs530831462 Chr.7: 155458105-155458105 on Build GRCh38
rs988208046 Chr.7: 155457980-155457980 on Build GRCh38
rs1001159337 Chr.7: 155458135-155458135 on Build GRCh38
rs954691981 Chr.7: 155458160-155458160 on Build GRCh38
rs925369018 Chr.7: 155457746-155457746 on Build GRCh38
rs935294410 Chr.7: 155457783-155457783 on Build GRCh38
rs1024321337 Chr.7: 155457938-155457938 on Build GRCh38
rs770375144 Chr.7: 155458153-155458153 on Build GRCh38
rs888924120 Chr.7: 155457855-155457855 on Build GRCh38
rs986534158 Chr.7: 155458173-155458173 on Build GRCh38
rs553512887 Chr.7: 155458012-155458012 on Build GRCh38
rs905442112 Chr.7: 155458111-155458111 on Build GRCh38
rs568597443 Chr.7: 155458035-155458035 on Build GRCh38
rs1013815430 Chr.7: 155457921-155457921 on Build GRCh38
rs956774872 Chr.7: 155457977-155457977 on Build GRCh38
rs895457521 Chr.7: 155458088-155458088 on Build GRCh38
rs563389380 Chr.7: 155458061-155458061 on Build GRCh38
rs1006473702 Chr.7: 155457870-155457870 on Build GRCh38
rs993231100 Chr.7: 155458066-155458066 on Build GRCh38
rs1050790390 Chr.7: 155457837-155457837 on Build GRCh38
rs993282107 Chr.7: 155457897-155457897 on Build GRCh38
rs941994661 Chr.7: 155458019-155458019 on Build GRCh38
rs916737611 Chr.7: 155457727-155457727 on Build GRCh38
rs959718741 Chr.7: 155457900-155457900 on Build GRCh38
rs910585784 Chr.7: 155458178-155458178 on Build GRCh38
rs979513252 Chr.7: 155457744-155457744 on Build GRCh38
rs373375475 Chr.7: 155458013-155458013 on Build GRCh38
rs988155646 Chr.7: 155458154-155458154 on Build GRCh38
rs146853597 Chr.7: 155457983-155457983 on Build GRCh38
rs1023851037 Chr.7: 155458092-155458092 on Build GRCh38
rs992850975 Chr.7: 155457722-155457722 on Build GRCh38

View Full Product Details