Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161542567-161543068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR3A
Entrez Gene ID
2214
Gene Name
Fc fragment of IgG receptor IIIa
Gene Aliases
CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
UniGene
Hs.372679
Gene Chromosome Location
Chr.1: 161541759-161550623 on Build GRCh38
Transcripts
NM_001127593.1 NM_001127595.1 NM_001127596.1 XM_011509293.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873807 c.825C>T p.D275D Chr.1: 161543060-161543060 on Build GRCh38
320389 c.817A>G p.T273A Chr.1: 161543068-161543068 on Build GRCh38
6888970 c.859C>T p.P287S Chr.1: 161543026-161543026 on Build GRCh38
463276 c.841T>C p.F281L Chr.1: 161543044-161543044 on Build GRCh38
4024414 c.839A>C p.K280T Chr.1: 161543046-161543046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565224758 Chr.1: 161542603-161542603 on Build GRCh38
rs533822244 Chr.1: 161542985-161542985 on Build GRCh38
rs762080565 Chr.1: 161543039-161543039 on Build GRCh38
rs903079937 Chr.1: 161543021-161543021 on Build GRCh38
rs929639692 Chr.1: 161542888-161542888 on Build GRCh38
rs892956348 Chr.1: 161542659-161542659 on Build GRCh38
rs542465352 Chr.1: 161543016-161543016 on Build GRCh38
rs547220247 Chr.1: 161542748-161542748 on Build GRCh38
rs567017084 Chr.1: 161542785-161542785 on Build GRCh38
rs966706182 Chr.1: 161542692-161542692 on Build GRCh38
rs3181668 Chr.1: 161542884-161542884 on Build GRCh38
rs555534589 Chr.1: 161543002-161543002 on Build GRCh38
rs753801794 Chr.1: 161542972-161542972 on Build GRCh38
rs745793832 Chr.1: 161542984-161542984 on Build GRCh38
rs138533290 Chr.1: 161542589-161542589 on Build GRCh38
rs911435611 Chr.1: 161542799-161542799 on Build GRCh38
rs550970281 Chr.1: 161542641-161542641 on Build GRCh38
rs756249160 Chr.1: 161542964-161542964 on Build GRCh38
rs768167909 Chr.1: 161543013-161543013 on Build GRCh38
rs1055593011 Chr.1: 161542650-161542650 on Build GRCh38
rs767229743 Chr.1: 161543025-161543025 on Build GRCh38
rs373205302 Chr.1: 161542594-161542594 on Build GRCh38
rs1042214 Chr.1: 161542732-161542732 on Build GRCh38
rs530970781 Chr.1: 161542626-161542626 on Build GRCh38
rs376707460 Chr.1: 161542675-161542675 on Build GRCh38
rs963187885 Chr.1: 161542814-161542814 on Build GRCh38
rs929608794 Chr.1: 161542640-161542641 on Build GRCh38
rs955452821 Chr.1: 161542766-161542766 on Build GRCh38
rs999459540 Chr.1: 161542693-161542693 on Build GRCh38
rs779795990 Chr.1: 161542991-161542991 on Build GRCh38
rs937145025 Chr.1: 161542927-161542927 on Build GRCh38
rs1126552 Chr.1: 161542996-161542996 on Build GRCh38
rs999492281 Chr.1: 161543030-161543030 on Build GRCh38
rs2499445 Chr.1: 161542755-161542755 on Build GRCh38
rs1042209 Chr.1: 161542981-161542981 on Build GRCh38
rs1042208 Chr.1: 161543044-161543044 on Build GRCh38
rs974344311 Chr.1: 161542856-161542856 on Build GRCh38
rs568266752 Chr.1: 161542979-161542979 on Build GRCh38
rs779031010 Chr.1: 161542983-161542983 on Build GRCh38
rs928371796 Chr.1: 161542910-161542910 on Build GRCh38
rs746855462 Chr.1: 161542992-161542992 on Build GRCh38
rs201318652 Chr.1: 161542642-161542642 on Build GRCh38
rs986036401 Chr.1: 161542775-161542775 on Build GRCh38
rs893000298 Chr.1: 161542962-161542962 on Build GRCh38
rs761113992 Chr.1: 161543024-161543024 on Build GRCh38
rs747818657 Chr.1: 161543001-161543001 on Build GRCh38
rs1042215 Chr.1: 161542728-161542728 on Build GRCh38
rs369766210 Chr.1: 161542677-161542677 on Build GRCh38
rs567991867 Chr.1: 161542649-161542649 on Build GRCh38
rs1039025792 Chr.1: 161542635-161542635 on Build GRCh38
rs759815530 Chr.1: 161543028-161543028 on Build GRCh38
rs763344006 Chr.1: 161543005-161543006 on Build GRCh38
rs1045523891 Chr.1: 161542641-161542641 on Build GRCh38
rs773725219 Chr.1: 161542995-161542998 on Build GRCh38
rs772894301 Chr.1: 161543009-161543009 on Build GRCh38
rs538851014 Chr.1: 161542848-161542848 on Build GRCh38
rs906921085 Chr.1: 161542644-161542644 on Build GRCh38
rs114559215 Chr.1: 161542870-161542870 on Build GRCh38
rs2973 Chr.1: 161542868-161542868 on Build GRCh38
rs1045390863 Chr.1: 161542901-161542901 on Build GRCh38
rs767581354 Chr.1: 161542746-161542746 on Build GRCh38
rs918283893 Chr.1: 161542638-161542638 on Build GRCh38
rs754086384 Chr.1: 161543036-161543036 on Build GRCh38
rs768148652 Chr.1: 161542995-161542995 on Build GRCh38
rs1011526477 Chr.1: 161542673-161542673 on Build GRCh38
rs7539036 Chr.1: 161542941-161542941 on Build GRCh38

View Full Product Details