Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165680357-165680791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH9A1
Entrez Gene ID
223
Gene Name
aldehyde dehydrogenase 9 family member A1
Gene Aliases
ALDH4, ALDH7, ALDH9, E3, TMABADH
UniGene
Hs.2533
Gene Chromosome Location
Chr.1: 165662212-165698663 on Build GRCh38
Transcripts
NM_000696.3 XM_011509294.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6392644 c.714G>C p.M238I Chr.1: 165680490-165680490 on Build GRCh38
5844366 c.740C>T p.P247L Chr.1: 165680536-165680536 on Build GRCh38
1689073 c.697G>A p.V233M Chr.1: 165680579-165680579 on Build GRCh38
4954006 c.669C>T p.P223P Chr.1: 165680535-165680535 on Build GRCh38
1689074 c.625G>A p.V209M Chr.1: 165680579-165680579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749430621 Chr.1: 165680660-165680660 on Build GRCh38
rs758621426 Chr.1: 165680500-165680500 on Build GRCh38
rs138240128 Chr.1: 165680623-165680623 on Build GRCh38
rs779617959 Chr.1: 165680498-165680498 on Build GRCh38
rs766037194 Chr.1: 165680544-165680544 on Build GRCh38
rs371323880 Chr.1: 165680535-165680535 on Build GRCh38
rs760374598 Chr.1: 165680712-165680712 on Build GRCh38
rs373284118 Chr.1: 165680526-165680526 on Build GRCh38
rs770756608 Chr.1: 165680663-165680663 on Build GRCh38
rs760415392 Chr.1: 165680608-165680608 on Build GRCh38
rs780492689 Chr.1: 165680502-165680502 on Build GRCh38
rs1033172420 Chr.1: 165680570-165680570 on Build GRCh38
rs747782664 Chr.1: 165680564-165680564 on Build GRCh38
rs922473575 Chr.1: 165680465-165680465 on Build GRCh38
rs746433891 Chr.1: 165680687-165680694 on Build GRCh38
rs372306386 Chr.1: 165680421-165680421 on Build GRCh38
rs189745059 Chr.1: 165680631-165680631 on Build GRCh38
rs758978207 Chr.1: 165680571-165680571 on Build GRCh38
rs988842656 Chr.1: 165680443-165680443 on Build GRCh38
rs554203371 Chr.1: 165680432-165680432 on Build GRCh38
rs1040959840 Chr.1: 165680458-165680458 on Build GRCh38
rs747426195 Chr.1: 165680576-165680576 on Build GRCh38
rs773362229 Chr.1: 165680584-165680584 on Build GRCh38
rs774231534 Chr.1: 165680545-165680545 on Build GRCh38
rs767327379 Chr.1: 165680552-165680552 on Build GRCh38
rs16849374 Chr.1: 165680478-165680478 on Build GRCh38
rs538012018 Chr.1: 165680739-165680739 on Build GRCh38
rs765209056 Chr.1: 165680459-165680459 on Build GRCh38
rs755272735 Chr.1: 165680646-165680646 on Build GRCh38
rs759458345 Chr.1: 165680600-165680600 on Build GRCh38
rs770941005 Chr.1: 165680592-165680592 on Build GRCh38
rs199563024 Chr.1: 165680474-165680474 on Build GRCh38
rs752870482 Chr.1: 165680650-165680650 on Build GRCh38
rs200586555 Chr.1: 165680439-165680439 on Build GRCh38
rs377384593 Chr.1: 165680617-165680617 on Build GRCh38
rs373394230 Chr.1: 165680687-165680687 on Build GRCh38
rs750848698 Chr.1: 165680569-165680569 on Build GRCh38
rs752294825 Chr.1: 165680483-165680483 on Build GRCh38
rs201526858 Chr.1: 165680480-165680480 on Build GRCh38
rs377272939 Chr.1: 165680477-165680477 on Build GRCh38
rs781553471 Chr.1: 165680649-165680649 on Build GRCh38
rs779128532 Chr.1: 165680523-165680524 on Build GRCh38
rs752009450 Chr.1: 165680734-165680734 on Build GRCh38
rs55747724 Chr.1: 165680395-165680396 on Build GRCh38
rs768405399 Chr.1: 165680721-165680721 on Build GRCh38
rs770348091 Chr.1: 165680745-165680745 on Build GRCh38
rs149264874 Chr.1: 165680455-165680455 on Build GRCh38
rs976703538 Chr.1: 165680677-165680677 on Build GRCh38
rs771691687 Chr.1: 165680688-165680688 on Build GRCh38
rs748745835 Chr.1: 165680679-165680679 on Build GRCh38
rs771933140 Chr.1: 165680693-165680693 on Build GRCh38
rs764662044 Chr.1: 165680730-165680730 on Build GRCh38
rs768676900 Chr.1: 165680454-165680454 on Build GRCh38
rs763661988 Chr.1: 165680567-165680567 on Build GRCh38
rs866836592 Chr.1: 165680416-165680416 on Build GRCh38
rs12139219 Chr.1: 165680482-165680482 on Build GRCh38
rs569459164 Chr.1: 165680737-165680737 on Build GRCh38
rs769719769 Chr.1: 165680528-165680528 on Build GRCh38
rs780253329 Chr.1: 165680574-165680574 on Build GRCh38
rs749138817 Chr.1: 165680589-165680589 on Build GRCh38
rs561795333 Chr.1: 165680577-165680577 on Build GRCh38
rs752237585 Chr.1: 165680556-165680556 on Build GRCh38
rs766203435 Chr.1: 165680475-165680475 on Build GRCh38
rs57850528 Chr.1: 165680604-165680604 on Build GRCh38
rs144479289 Chr.1: 165680599-165680599 on Build GRCh38
rs147829872 Chr.1: 165680534-165680534 on Build GRCh38
rs747163146 Chr.1: 165680509-165680509 on Build GRCh38
rs777681082 Chr.1: 165680657-165680657 on Build GRCh38
rs879322619 Chr.1: 165680431-165680431 on Build GRCh38
rs549579859 Chr.1: 165680728-165680728 on Build GRCh38
rs1024946102 Chr.1: 165680560-165680560 on Build GRCh38
rs1065756 Chr.1: 165680614-165680614 on Build GRCh38
rs757811733 Chr.1: 165680495-165680495 on Build GRCh38
rs776674979 Chr.1: 165680517-165680517 on Build GRCh38
rs369406617 Chr.1: 165680694-165680694 on Build GRCh38
rs970256266 Chr.1: 165680561-165680561 on Build GRCh38
rs778974816 Chr.1: 165680676-165680676 on Build GRCh38
rs750220071 Chr.1: 165680468-165680468 on Build GRCh38
rs768910049 Chr.1: 165680512-165680512 on Build GRCh38
rs751921595 Chr.1: 165680632-165680632 on Build GRCh38
rs202016807 Chr.1: 165680547-165680547 on Build GRCh38
rs756387768 Chr.1: 165680655-165680655 on Build GRCh38
rs775638411 Chr.1: 165680450-165680450 on Build GRCh38
rs200799083 Chr.1: 165680579-165680579 on Build GRCh38
rs746236428 Chr.1: 165680499-165680499 on Build GRCh38
rs775213108 Chr.1: 165680606-165680606 on Build GRCh38
rs540669550 Chr.1: 165680425-165680425 on Build GRCh38
rs760967937 Chr.1: 165680451-165680451 on Build GRCh38

View Full Product Details