Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167432936-167433435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD247
Entrez Gene ID
919
Gene Name
CD247 molecule
Gene Aliases
CD3-ZETA, CD3H, CD3Q, CD3Z, IMD25, T3Z, TCRZ
UniGene
Hs.156445
Gene Chromosome Location
Chr.1: 167430640-167518610 on Build GRCh38
Transcripts
NM_000734.3 NM_198053.2 XM_011510144.2 XM_011510145.1 XM_017002800.1 XM_017002801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4546582 c.402G>A p.R134R Chr.1: 167433051-167433051 on Build GRCh38
2185731 c.415G>A p.D139N Chr.1: 167433038-167433038 on Build GRCh38
4546391 c.399G>A p.R133R Chr.1: 167433054-167433054 on Build GRCh38
4435197 c.395G>A p.R132H Chr.1: 167433058-167433058 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372349405 Chr.1: 167432979-167432979 on Build GRCh38
rs753278244 Chr.1: 167433043-167433043 on Build GRCh38
rs770188589 Chr.1: 167433106-167433106 on Build GRCh38
rs906594562 Chr.1: 167433361-167433361 on Build GRCh38
rs542096092 Chr.1: 167433323-167433323 on Build GRCh38
rs759213998 Chr.1: 167433026-167433026 on Build GRCh38
rs771873949 Chr.1: 167433034-167433034 on Build GRCh38
rs916878824 Chr.1: 167433183-167433183 on Build GRCh38
rs1014946214 Chr.1: 167432959-167432959 on Build GRCh38
rs977050363 Chr.1: 167433066-167433066 on Build GRCh38
rs769910123 Chr.1: 167433005-167433005 on Build GRCh38
rs201171777 Chr.1: 167433012-167433012 on Build GRCh38
rs553076257 Chr.1: 167433259-167433259 on Build GRCh38
rs770758796 Chr.1: 167433019-167433019 on Build GRCh38
rs949763381 Chr.1: 167433261-167433261 on Build GRCh38
rs781007114 Chr.1: 167433175-167433175 on Build GRCh38
rs747661888 Chr.1: 167433063-167433065 on Build GRCh38
rs1002239613 Chr.1: 167433378-167433378 on Build GRCh38
rs751981677 Chr.1: 167433056-167433056 on Build GRCh38
rs756323539 Chr.1: 167433077-167433077 on Build GRCh38
rs563902268 Chr.1: 167433349-167433349 on Build GRCh38
rs201594815 Chr.1: 167433058-167433058 on Build GRCh38
rs1025579571 Chr.1: 167433071-167433071 on Build GRCh38
rs957929190 Chr.1: 167433011-167433011 on Build GRCh38
rs150184619 Chr.1: 167433117-167433117 on Build GRCh38
rs762342904 Chr.1: 167433263-167433263 on Build GRCh38
rs541146840 Chr.1: 167433179-167433179 on Build GRCh38
rs369387062 Chr.1: 167433069-167433069 on Build GRCh38
rs776228262 Chr.1: 167433004-167433005 on Build GRCh38
rs985073093 Chr.1: 167433131-167433131 on Build GRCh38
rs778667596 Chr.1: 167433099-167433099 on Build GRCh38
rs770933214 Chr.1: 167433097-167433097 on Build GRCh38
rs937118599 Chr.1: 167433160-167433160 on Build GRCh38
rs768158517 Chr.1: 167433002-167433003 on Build GRCh38
rs371723822 Chr.1: 167433089-167433089 on Build GRCh38
rs919433768 Chr.1: 167433286-167433286 on Build GRCh38
rs903449454 Chr.1: 167433272-167433272 on Build GRCh38
rs775332123 Chr.1: 167433036-167433036 on Build GRCh38
rs930183373 Chr.1: 167433195-167433195 on Build GRCh38
rs942299387 Chr.1: 167433209-167433209 on Build GRCh38
rs375979320 Chr.1: 167433102-167433102 on Build GRCh38
rs909628377 Chr.1: 167433113-167433113 on Build GRCh38
rs781510519 Chr.1: 167433059-167433059 on Build GRCh38
rs369656778 Chr.1: 167433110-167433110 on Build GRCh38
rs746682014 Chr.1: 167432998-167432999 on Build GRCh38
rs1054587078 Chr.1: 167433337-167433337 on Build GRCh38
rs1037894298 Chr.1: 167433271-167433271 on Build GRCh38
rs772849845 Chr.1: 167433257-167433257 on Build GRCh38
rs574574531 Chr.1: 167433311-167433311 on Build GRCh38
rs1042328127 Chr.1: 167433262-167433262 on Build GRCh38
rs763532939 Chr.1: 167433045-167433045 on Build GRCh38
rs148298128 Chr.1: 167433042-167433042 on Build GRCh38
rs766107244 Chr.1: 167433295-167433295 on Build GRCh38
rs748245937 Chr.1: 167432992-167432992 on Build GRCh38
rs777799139 Chr.1: 167433008-167433008 on Build GRCh38
rs184894260 Chr.1: 167433039-167433039 on Build GRCh38
rs769263658 Chr.1: 167433064-167433067 on Build GRCh38
rs762353729 Chr.1: 167433009-167433009 on Build GRCh38
rs147527561 Chr.1: 167433055-167433055 on Build GRCh38
rs868591103 Chr.1: 167433304-167433304 on Build GRCh38
rs569894562 Chr.1: 167433020-167433020 on Build GRCh38
rs1055145045 Chr.1: 167433398-167433398 on Build GRCh38

View Full Product Details