Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167433929-167434439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD247
Entrez Gene ID
919
Gene Name
CD247 molecule
Gene Aliases
CD3-ZETA, CD3H, CD3Q, CD3Z, IMD25, T3Z, TCRZ
UniGene
Hs.156445
Gene Chromosome Location
Chr.1: 167430640-167518610 on Build GRCh38
Transcripts
NM_000734.3 NM_198053.2 XM_011510144.2 XM_011510145.1 XM_017002800.1 XM_017002801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
301897 c.365C>G p.A122G Chr.1: 167434048-167434048 on Build GRCh38
139337 c.361G>A p.E121K Chr.1: 167434052-167434052 on Build GRCh38
1195410 c.359C>T p.A120V Chr.1: 167434054-167434054 on Build GRCh38
5000780 c.391G>A p.E131K Chr.1: 167434022-167434022 on Build GRCh38
1689102 c.337G>A p.E113K Chr.1: 167434076-167434076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34500066 Chr.1: 167434311-167434311 on Build GRCh38
rs150725547 Chr.1: 167434091-167434091 on Build GRCh38
rs946795652 Chr.1: 167434197-167434197 on Build GRCh38
rs775424384 Chr.1: 167433970-167433970 on Build GRCh38
rs990332661 Chr.1: 167433978-167433978 on Build GRCh38
rs753046264 Chr.1: 167434023-167434023 on Build GRCh38
rs959565377 Chr.1: 167434145-167434145 on Build GRCh38
rs1051493040 Chr.1: 167434254-167434254 on Build GRCh38
rs35674953 Chr.1: 167433951-167433951 on Build GRCh38
rs563936741 Chr.1: 167434346-167434346 on Build GRCh38
rs768351916 Chr.1: 167434148-167434148 on Build GRCh38
rs983501937 Chr.1: 167434407-167434407 on Build GRCh38
rs866150338 Chr.1: 167433964-167433964 on Build GRCh38
rs191531479 Chr.1: 167434264-167434264 on Build GRCh38
rs959600008 Chr.1: 167434373-167434373 on Build GRCh38
rs530758655 Chr.1: 167433953-167433953 on Build GRCh38
rs1035702137 Chr.1: 167434361-167434361 on Build GRCh38
rs933972157 Chr.1: 167434249-167434249 on Build GRCh38
rs182972526 Chr.1: 167434397-167434397 on Build GRCh38
rs887751135 Chr.1: 167434117-167434117 on Build GRCh38
rs768383234 Chr.1: 167434087-167434087 on Build GRCh38
rs745871212 Chr.1: 167434054-167434054 on Build GRCh38
rs776212068 Chr.1: 167433984-167433984 on Build GRCh38
rs767138961 Chr.1: 167434293-167434293 on Build GRCh38
rs982782025 Chr.1: 167434127-167434127 on Build GRCh38
rs760005631 Chr.1: 167434009-167434009 on Build GRCh38
rs770304810 Chr.1: 167434161-167434161 on Build GRCh38
rs559429804 Chr.1: 167434246-167434246 on Build GRCh38
rs573579199 Chr.1: 167434219-167434219 on Build GRCh38
rs976642056 Chr.1: 167434377-167434377 on Build GRCh38
rs375668936 Chr.1: 167434090-167434090 on Build GRCh38
rs929474618 Chr.1: 167434395-167434395 on Build GRCh38
rs971050571 Chr.1: 167434125-167434125 on Build GRCh38
rs773202723 Chr.1: 167434040-167434055 on Build GRCh38
rs993882641 Chr.1: 167434292-167434292 on Build GRCh38
rs772774207 Chr.1: 167434111-167434111 on Build GRCh38
rs139030667 Chr.1: 167434120-167434120 on Build GRCh38
rs1024171967 Chr.1: 167434184-167434184 on Build GRCh38
rs889682272 Chr.1: 167434261-167434261 on Build GRCh38
rs774765117 Chr.1: 167434000-167434000 on Build GRCh38
rs915355910 Chr.1: 167434186-167434186 on Build GRCh38
rs910789596 Chr.1: 167434300-167434300 on Build GRCh38
rs372665461 Chr.1: 167434048-167434048 on Build GRCh38
rs933919864 Chr.1: 167433980-167433980 on Build GRCh38
rs764185491 Chr.1: 167434030-167434030 on Build GRCh38
rs946721668 Chr.1: 167433965-167433965 on Build GRCh38
rs113240836 Chr.1: 167434263-167434263 on Build GRCh38
rs923341521 Chr.1: 167434057-167434057 on Build GRCh38
rs773572491 Chr.1: 167434022-167434022 on Build GRCh38
rs36056000 Chr.1: 167434083-167434083 on Build GRCh38
rs756340039 Chr.1: 167434027-167434027 on Build GRCh38
rs75532935 Chr.1: 167434081-167434081 on Build GRCh38
rs528412061 Chr.1: 167434374-167434374 on Build GRCh38
rs35013669 Chr.1: 167434185-167434188 on Build GRCh38
rs952963 Chr.1: 167433975-167433975 on Build GRCh38
rs568106106 Chr.1: 167434053-167434053 on Build GRCh38
rs563414907 Chr.1: 167434266-167434266 on Build GRCh38
rs1035268255 Chr.1: 167434133-167434133 on Build GRCh38
rs138738209 Chr.1: 167433974-167433974 on Build GRCh38
rs199702239 Chr.1: 167433993-167433993 on Build GRCh38
rs769223722 Chr.1: 167434094-167434094 on Build GRCh38
rs952962 Chr.1: 167434267-167434267 on Build GRCh38
rs540922745 Chr.1: 167434236-167434236 on Build GRCh38
rs1002537349 Chr.1: 167434333-167434333 on Build GRCh38
rs200375126 Chr.1: 167433987-167433987 on Build GRCh38
rs953809 Chr.1: 167434388-167434388 on Build GRCh38
rs971538088 Chr.1: 167434202-167434202 on Build GRCh38
rs1038257468 Chr.1: 167434135-167434135 on Build GRCh38
rs535609428 Chr.1: 167434073-167434073 on Build GRCh38
rs760093486 Chr.1: 167434114-167434114 on Build GRCh38

View Full Product Details