Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 75012777-75013306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD1
Entrez Gene ID
83690
Gene Name
cysteine rich secretory protein LCCL domain containing 1
Gene Aliases
CRISP-10, CRISP10, LCRISP1
UniGene
Hs.436542
Gene Chromosome Location
Chr.8: 74984473-75034558 on Build GRCh38
Transcripts
NM_001286777.1 NM_001286778.1 NM_031461.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232099 c.389C>T p.P130L Chr.8: 75012901-75012901 on Build GRCh38
6189361 c.396T>G p.F132L Chr.8: 75012908-75012908 on Build GRCh38
307605 c.463T>A p.Y155N Chr.8: 75012975-75012975 on Build GRCh38
391833 c.413_415delATG p.D139delD Chr.8: 75012925-75012927 on Build GRCh38
4441344 c.420A>C p.E140D Chr.8: 75012932-75012932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757617652 Chr.8: 75012914-75012914 on Build GRCh38
rs909419520 Chr.8: 75013202-75013204 on Build GRCh38
rs758899745 Chr.8: 75012922-75012922 on Build GRCh38
rs928283958 Chr.8: 75013126-75013126 on Build GRCh38
rs773150026 Chr.8: 75012968-75012968 on Build GRCh38
rs942316739 Chr.8: 75013205-75013205 on Build GRCh38
rs771051583 Chr.8: 75013031-75013031 on Build GRCh38
rs933373790 Chr.8: 75013219-75013219 on Build GRCh38
rs142874642 Chr.8: 75012971-75012971 on Build GRCh38
rs771653656 Chr.8: 75012853-75012853 on Build GRCh38
rs200273072 Chr.8: 75012904-75012904 on Build GRCh38
rs1032348502 Chr.8: 75013260-75013260 on Build GRCh38
rs913081166 Chr.8: 75013115-75013115 on Build GRCh38
rs761555951 Chr.8: 75012864-75012864 on Build GRCh38
rs778493533 Chr.8: 75012840-75012840 on Build GRCh38
rs746671711 Chr.8: 75013036-75013036 on Build GRCh38
rs989910884 Chr.8: 75012847-75012847 on Build GRCh38
rs765692351 Chr.8: 75013027-75013027 on Build GRCh38
rs764878772 Chr.8: 75013000-75013000 on Build GRCh38
rs568124456 Chr.8: 75012830-75012830 on Build GRCh38
rs746163483 Chr.8: 75012919-75012919 on Build GRCh38
rs966275109 Chr.8: 75012849-75012849 on Build GRCh38
rs919375704 Chr.8: 75012874-75012874 on Build GRCh38
rs776499281 Chr.8: 75012955-75012955 on Build GRCh38
rs752858747 Chr.8: 75012895-75012895 on Build GRCh38
rs375089740 Chr.8: 75012860-75012860 on Build GRCh38
rs139635677 Chr.8: 75012868-75012868 on Build GRCh38
rs758076492 Chr.8: 75012920-75012920 on Build GRCh38
rs774986499 Chr.8: 75012856-75012856 on Build GRCh38
rs893017580 Chr.8: 75013282-75013282 on Build GRCh38
rs749147587 Chr.8: 75013072-75013072 on Build GRCh38
rs892905371 Chr.8: 75013042-75013042 on Build GRCh38
rs371136231 Chr.8: 75013048-75013048 on Build GRCh38
rs997615098 Chr.8: 75012932-75012932 on Build GRCh38
rs952211532 Chr.8: 75012883-75012883 on Build GRCh38
rs769563591 Chr.8: 75012965-75012965 on Build GRCh38
rs747547689 Chr.8: 75013050-75013050 on Build GRCh38
rs889409701 Chr.8: 75013133-75013133 on Build GRCh38
rs867332166 Chr.8: 75013019-75013019 on Build GRCh38
rs1007730432 Chr.8: 75012816-75012816 on Build GRCh38
rs767877678 Chr.8: 75012973-75012973 on Build GRCh38
rs993701942 Chr.8: 75013190-75013190 on Build GRCh38
rs1026877920 Chr.8: 75013215-75013215 on Build GRCh38
rs745503513 Chr.8: 75012850-75012850 on Build GRCh38
rs369100451 Chr.8: 75012902-75012902 on Build GRCh38
rs1055963692 Chr.8: 75013196-75013196 on Build GRCh38
rs1007844711 Chr.8: 75013138-75013138 on Build GRCh38
rs966680505 Chr.8: 75013180-75013180 on Build GRCh38
rs1036909375 Chr.8: 75013207-75013207 on Build GRCh38
rs377151138 Chr.8: 75012872-75012872 on Build GRCh38
rs746550333 Chr.8: 75012935-75012935 on Build GRCh38
rs556446919 Chr.8: 75013257-75013257 on Build GRCh38
rs928598621 Chr.8: 75013275-75013275 on Build GRCh38
rs750806807 Chr.8: 75013031-75013031 on Build GRCh38
rs201583125 Chr.8: 75013035-75013035 on Build GRCh38
rs552107887 Chr.8: 75012974-75012974 on Build GRCh38
rs773239890 Chr.8: 75013053-75013053 on Build GRCh38
rs1049393298 Chr.8: 75013120-75013120 on Build GRCh38
rs760731631 Chr.8: 75012999-75012999 on Build GRCh38
rs143754470 Chr.8: 75012901-75012901 on Build GRCh38
rs1019187623 Chr.8: 75013154-75013154 on Build GRCh38
rs760136517 Chr.8: 75012857-75012857 on Build GRCh38
rs981185124 Chr.8: 75013125-75013125 on Build GRCh38
rs377728137 Chr.8: 75012972-75012972 on Build GRCh38
rs753799601 Chr.8: 75013025-75013025 on Build GRCh38
rs751578520 Chr.8: 75013150-75013150 on Build GRCh38
rs889114739 Chr.8: 75013255-75013255 on Build GRCh38
rs200298882 Chr.8: 75012893-75012893 on Build GRCh38
rs182818008 Chr.8: 75013235-75013235 on Build GRCh38
rs1011251757 Chr.8: 75013103-75013103 on Build GRCh38
rs780165003 Chr.8: 75013032-75013032 on Build GRCh38
rs748104481 Chr.8: 75012961-75012961 on Build GRCh38
rs1022976422 Chr.8: 75013236-75013236 on Build GRCh38
rs776610689 Chr.8: 75012862-75012862 on Build GRCh38
rs372002946 Chr.8: 75013047-75013047 on Build GRCh38
rs75126481 Chr.8: 75012810-75012810 on Build GRCh38
rs1051890844 Chr.8: 75013220-75013220 on Build GRCh38
rs768182921 Chr.8: 75012954-75012954 on Build GRCh38
rs981433645 Chr.8: 75013259-75013259 on Build GRCh38
rs957472873 Chr.8: 75012812-75012812 on Build GRCh38
rs764216055 Chr.8: 75013016-75013016 on Build GRCh38
rs1045639839 Chr.8: 75012937-75012937 on Build GRCh38
rs952177572 Chr.8: 75013216-75013216 on Build GRCh38
rs138345977 Chr.8: 75012917-75012917 on Build GRCh38
rs372944451 Chr.8: 75012869-75012869 on Build GRCh38
rs76615567 Chr.8: 75013021-75013021 on Build GRCh38
rs758738596 Chr.8: 75013034-75013034 on Build GRCh38
rs879884632 Chr.8: 75012820-75012820 on Build GRCh38

View Full Product Details