Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 75016908-75017408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD1
Entrez Gene ID
83690
Gene Name
cysteine rich secretory protein LCCL domain containing 1
Gene Aliases
CRISP-10, CRISP10, LCRISP1
UniGene
Hs.436542
Gene Chromosome Location
Chr.8: 74984473-75034558 on Build GRCh38
Transcripts
NM_001286777.1 NM_001286778.1 NM_031461.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5640577 c.1082A>T p.K361M Chr.8: 75017405-75017405 on Build GRCh38
5686846 c.934G>T p.E312* Chr.8: 75017051-75017051 on Build GRCh38
300751 c.909C>T p.C303C Chr.8: 75016921-75016921 on Build GRCh38
4674725 c.897delA p.R300fs*46 Chr.8: 75016909-75016909 on Build GRCh38
4674726 c.1010G>A p.C337Y Chr.8: 75017333-75017333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753564953 Chr.8: 75017365-75017367 on Build GRCh38
rs564871516 Chr.8: 75017299-75017299 on Build GRCh38
rs752665381 Chr.8: 75017294-75017294 on Build GRCh38
rs572260524 Chr.8: 75017200-75017200 on Build GRCh38
rs771563684 Chr.8: 75017113-75017113 on Build GRCh38
rs199841663 Chr.8: 75017070-75017070 on Build GRCh38
rs755088060 Chr.8: 75017046-75017046 on Build GRCh38
rs903747747 Chr.8: 75017188-75017188 on Build GRCh38
rs560563736 Chr.8: 75017135-75017135 on Build GRCh38
rs773877874 Chr.8: 75016978-75016978 on Build GRCh38
rs745420208 Chr.8: 75017103-75017103 on Build GRCh38
rs761652703 Chr.8: 75017156-75017156 on Build GRCh38
rs372735125 Chr.8: 75017004-75017004 on Build GRCh38
rs1027079313 Chr.8: 75017336-75017336 on Build GRCh38
rs745330107 Chr.8: 75016982-75016982 on Build GRCh38
rs140591468 Chr.8: 75017380-75017380 on Build GRCh38
rs75402710 Chr.8: 75017136-75017136 on Build GRCh38
rs746462129 Chr.8: 75017356-75017356 on Build GRCh38
rs865976337 Chr.8: 75016935-75016935 on Build GRCh38
rs1024945546 Chr.8: 75017304-75017304 on Build GRCh38
rs777358751 Chr.8: 75016940-75016940 on Build GRCh38
rs756496842 Chr.8: 75017062-75017062 on Build GRCh38
rs757928709 Chr.8: 75017321-75017321 on Build GRCh38
rs149998958 Chr.8: 75017305-75017305 on Build GRCh38
rs754247137 Chr.8: 75017302-75017302 on Build GRCh38
rs563119196 Chr.8: 75016999-75016999 on Build GRCh38
rs766670045 Chr.8: 75017035-75017035 on Build GRCh38
rs921254371 Chr.8: 75017131-75017131 on Build GRCh38
rs368207321 Chr.8: 75017278-75017278 on Build GRCh38
rs368635431 Chr.8: 75017001-75017001 on Build GRCh38
rs542481481 Chr.8: 75017127-75017127 on Build GRCh38
rs1052209217 Chr.8: 75017216-75017216 on Build GRCh38
rs754819399 Chr.8: 75017382-75017383 on Build GRCh38
rs767952215 Chr.8: 75017049-75017049 on Build GRCh38
rs779750749 Chr.8: 75017127-75017128 on Build GRCh38
rs755734190 Chr.8: 75016979-75016982 on Build GRCh38
rs770517082 Chr.8: 75016977-75016977 on Build GRCh38
rs763272270 Chr.8: 75017032-75017032 on Build GRCh38
rs767624404 Chr.8: 75017292-75017292 on Build GRCh38
rs920150717 Chr.8: 75016976-75016976 on Build GRCh38
rs779620373 Chr.8: 75017322-75017322 on Build GRCh38
rs892325371 Chr.8: 75017222-75017222 on Build GRCh38
rs940228959 Chr.8: 75017104-75017104 on Build GRCh38
rs1037212366 Chr.8: 75017129-75017129 on Build GRCh38
rs878857283 Chr.8: 75017051-75017051 on Build GRCh38
rs201994686 Chr.8: 75017308-75017308 on Build GRCh38
rs143020863 Chr.8: 75017320-75017320 on Build GRCh38
rs748512304 Chr.8: 75016939-75016939 on Build GRCh38
rs921699577 Chr.8: 75017352-75017352 on Build GRCh38
rs778511656 Chr.8: 75017094-75017094 on Build GRCh38
rs764455539 Chr.8: 75017300-75017300 on Build GRCh38
rs1013286942 Chr.8: 75017244-75017244 on Build GRCh38
rs775346012 Chr.8: 75017126-75017126 on Build GRCh38
rs759759831 Chr.8: 75017270-75017270 on Build GRCh38
rs368746197 Chr.8: 75017067-75017067 on Build GRCh38
rs374835501 Chr.8: 75016963-75016963 on Build GRCh38
rs1032216175 Chr.8: 75017315-75017315 on Build GRCh38
rs1041172650 Chr.8: 75017120-75017120 on Build GRCh38
rs544907412 Chr.8: 75016938-75016938 on Build GRCh38
rs776231297 Chr.8: 75017019-75017019 on Build GRCh38
rs201783452 Chr.8: 75017266-75017266 on Build GRCh38
rs775464109 Chr.8: 75016987-75016987 on Build GRCh38
rs751712792 Chr.8: 75017039-75017039 on Build GRCh38
rs1055653873 Chr.8: 75017218-75017218 on Build GRCh38
rs768661226 Chr.8: 75017358-75017358 on Build GRCh38
rs753246288 Chr.8: 75017050-75017050 on Build GRCh38
rs766758168 Chr.8: 75017269-75017269 on Build GRCh38
rs1001853353 Chr.8: 75017314-75017314 on Build GRCh38
rs1043455220 Chr.8: 75017142-75017142 on Build GRCh38
rs772043824 Chr.8: 75016984-75016984 on Build GRCh38
rs76339229 Chr.8: 75017108-75017108 on Build GRCh38

View Full Product Details