Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 75017092-75017635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD1
Entrez Gene ID
83690
Gene Name
cysteine rich secretory protein LCCL domain containing 1
Gene Aliases
CRISP-10, CRISP10, LCRISP1
UniGene
Hs.436542
Gene Chromosome Location
Chr.8: 74984473-75034558 on Build GRCh38
Transcripts
NM_001286777.1 NM_001286778.1 NM_031461.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874982 c.1052G>A p.G351D Chr.8: 75017375-75017375 on Build GRCh38
5640577 c.1082A>T p.K361M Chr.8: 75017405-75017405 on Build GRCh38
6635537 c.1024C>A p.H342N Chr.8: 75017347-75017347 on Build GRCh38
4674726 c.1010G>A p.C337Y Chr.8: 75017333-75017333 on Build GRCh38
7362699 c.1118A>T p.Q373L Chr.8: 75017441-75017441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753564953 Chr.8: 75017365-75017367 on Build GRCh38
rs779524759 Chr.8: 75017435-75017435 on Build GRCh38
rs931499863 Chr.8: 75017596-75017596 on Build GRCh38
rs769710930 Chr.8: 75017395-75017395 on Build GRCh38
rs564871516 Chr.8: 75017299-75017299 on Build GRCh38
rs752665381 Chr.8: 75017294-75017294 on Build GRCh38
rs751120378 Chr.8: 75017440-75017440 on Build GRCh38
rs764699142 Chr.8: 75017419-75017419 on Build GRCh38
rs572260524 Chr.8: 75017200-75017200 on Build GRCh38
rs558892566 Chr.8: 75017534-75017534 on Build GRCh38
rs777720965 Chr.8: 75017469-75017469 on Build GRCh38
rs903747747 Chr.8: 75017188-75017188 on Build GRCh38
rs545259926 Chr.8: 75017511-75017512 on Build GRCh38
rs765598042 Chr.8: 75017428-75017428 on Build GRCh38
rs560563736 Chr.8: 75017135-75017135 on Build GRCh38
rs761652703 Chr.8: 75017156-75017156 on Build GRCh38
rs762458786 Chr.8: 75017503-75017504 on Build GRCh38
rs1027079313 Chr.8: 75017336-75017336 on Build GRCh38
rs75402710 Chr.8: 75017136-75017136 on Build GRCh38
rs140591468 Chr.8: 75017380-75017380 on Build GRCh38
rs189810794 Chr.8: 75017426-75017426 on Build GRCh38
rs746462129 Chr.8: 75017356-75017356 on Build GRCh38
rs754422234 Chr.8: 75017446-75017446 on Build GRCh38
rs750718806 Chr.8: 75017432-75017432 on Build GRCh38
rs1024945546 Chr.8: 75017304-75017304 on Build GRCh38
rs757928709 Chr.8: 75017321-75017321 on Build GRCh38
rs772122275 Chr.8: 75017411-75017411 on Build GRCh38
rs373503585 Chr.8: 75017498-75017498 on Build GRCh38
rs149998958 Chr.8: 75017305-75017305 on Build GRCh38
rs973019683 Chr.8: 75017547-75017547 on Build GRCh38
rs754247137 Chr.8: 75017302-75017302 on Build GRCh38
rs921321905 Chr.8: 75017487-75017487 on Build GRCh38
rs921254371 Chr.8: 75017131-75017131 on Build GRCh38
rs368207321 Chr.8: 75017278-75017278 on Build GRCh38
rs1052209217 Chr.8: 75017216-75017216 on Build GRCh38
rs542481481 Chr.8: 75017127-75017127 on Build GRCh38
rs754819399 Chr.8: 75017382-75017383 on Build GRCh38
rs758100678 Chr.8: 75017434-75017434 on Build GRCh38
rs779750749 Chr.8: 75017127-75017128 on Build GRCh38
rs150474883 Chr.8: 75017415-75017415 on Build GRCh38
rs977331518 Chr.8: 75017468-75017468 on Build GRCh38
rs774775508 Chr.8: 75017405-75017405 on Build GRCh38
rs570882648 Chr.8: 75017563-75017563 on Build GRCh38
rs182073632 Chr.8: 75017581-75017581 on Build GRCh38
rs767624404 Chr.8: 75017292-75017292 on Build GRCh38
rs769783972 Chr.8: 75017466-75017466 on Build GRCh38
rs760645153 Chr.8: 75017418-75017418 on Build GRCh38
rs533976647 Chr.8: 75017525-75017525 on Build GRCh38
rs779620373 Chr.8: 75017322-75017322 on Build GRCh38
rs567149765 Chr.8: 75017503-75017503 on Build GRCh38
rs748091694 Chr.8: 75017454-75017454 on Build GRCh38
rs781334099 Chr.8: 75017452-75017452 on Build GRCh38
rs892325371 Chr.8: 75017222-75017222 on Build GRCh38
rs1037212366 Chr.8: 75017129-75017129 on Build GRCh38
rs201994686 Chr.8: 75017308-75017308 on Build GRCh38
rs143020863 Chr.8: 75017320-75017320 on Build GRCh38
rs921699577 Chr.8: 75017352-75017352 on Build GRCh38
rs764455539 Chr.8: 75017300-75017300 on Build GRCh38
rs775346012 Chr.8: 75017126-75017126 on Build GRCh38
rs1013286942 Chr.8: 75017244-75017244 on Build GRCh38
rs759759831 Chr.8: 75017270-75017270 on Build GRCh38
rs1032216175 Chr.8: 75017315-75017315 on Build GRCh38
rs368953488 Chr.8: 75017502-75017502 on Build GRCh38
rs1041172650 Chr.8: 75017120-75017120 on Build GRCh38
rs925985971 Chr.8: 75017512-75017512 on Build GRCh38
rs759855345 Chr.8: 75017408-75017408 on Build GRCh38
rs556755859 Chr.8: 75017600-75017600 on Build GRCh38
rs201783452 Chr.8: 75017266-75017266 on Build GRCh38
rs1055653873 Chr.8: 75017218-75017218 on Build GRCh38
rs768661226 Chr.8: 75017358-75017358 on Build GRCh38
rs766758168 Chr.8: 75017269-75017269 on Build GRCh38
rs1001853353 Chr.8: 75017314-75017314 on Build GRCh38
rs1043455220 Chr.8: 75017142-75017142 on Build GRCh38
rs200771634 Chr.8: 75017391-75017391 on Build GRCh38
rs754337022 Chr.8: 75017425-75017425 on Build GRCh38

View Full Product Details