Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 79766916-79767415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEY1
Entrez Gene ID
23462
Gene Name
hes related family bHLH transcription factor with YRPW motif 1
Gene Aliases
BHLHb31, CHF2, HERP2, HESR1, HRT-1, OAF1, hHRT1
UniGene
Hs.234434
Gene Chromosome Location
Chr.8: 79764010-79767863 on Build GRCh38
Transcripts
NM_001040708.1 NM_012258.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4917699 c.170T>G p.I57S Chr.8: 79767088-79767088 on Build GRCh38
6007453 c.109G>T p.G37C Chr.8: 79767275-79767275 on Build GRCh38
4588209 c.134C>T p.S45F Chr.8: 79767250-79767250 on Build GRCh38
7203797 c.168A>G p.I56M Chr.8: 79767090-79767090 on Build GRCh38
311758 c.178C>A p.R60S Chr.8: 79767080-79767080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764631911 Chr.8: 79766963-79766963 on Build GRCh38
rs756094898 Chr.8: 79767282-79767282 on Build GRCh38
rs1027966515 Chr.8: 79767286-79767286 on Build GRCh38
rs748969294 Chr.8: 79767066-79767066 on Build GRCh38
rs928480792 Chr.8: 79767220-79767220 on Build GRCh38
rs769534721 Chr.8: 79766947-79766947 on Build GRCh38
rs371911243 Chr.8: 79766950-79766950 on Build GRCh38
rs749044330 Chr.8: 79767307-79767310 on Build GRCh38
rs764543834 Chr.8: 79767115-79767115 on Build GRCh38
rs528107512 Chr.8: 79767264-79767264 on Build GRCh38
rs777037397 Chr.8: 79767248-79767248 on Build GRCh38
rs748087926 Chr.8: 79767193-79767193 on Build GRCh38
rs767419342 Chr.8: 79767277-79767277 on Build GRCh38
rs899966250 Chr.8: 79767117-79767117 on Build GRCh38
rs997444809 Chr.8: 79767348-79767348 on Build GRCh38
rs939237381 Chr.8: 79767221-79767221 on Build GRCh38
rs200772885 Chr.8: 79766957-79766957 on Build GRCh38
rs770586080 Chr.8: 79767190-79767193 on Build GRCh38
rs769531892 Chr.8: 79766968-79766968 on Build GRCh38
rs747305349 Chr.8: 79767227-79767227 on Build GRCh38
rs879205886 Chr.8: 79767152-79767153 on Build GRCh38
rs765346979 Chr.8: 79767121-79767121 on Build GRCh38
rs374632267 Chr.8: 79766975-79766975 on Build GRCh38
rs567458611 Chr.8: 79767358-79767358 on Build GRCh38
rs144815296 Chr.8: 79767260-79767260 on Build GRCh38
rs113738053 Chr.8: 79766986-79766986 on Build GRCh38
rs367664484 Chr.8: 79766964-79766964 on Build GRCh38
rs937207190 Chr.8: 79767237-79767237 on Build GRCh38
rs767914832 Chr.8: 79766952-79766952 on Build GRCh38
rs575360045 Chr.8: 79767037-79767037 on Build GRCh38
rs957087334 Chr.8: 79767347-79767347 on Build GRCh38
rs771778263 Chr.8: 79767321-79767321 on Build GRCh38
rs745442086 Chr.8: 79766949-79766949 on Build GRCh38
rs559360995 Chr.8: 79767209-79767209 on Build GRCh38
rs769020386 Chr.8: 79767234-79767234 on Build GRCh38
rs140932864 Chr.8: 79767027-79767027 on Build GRCh38
rs770525659 Chr.8: 79767099-79767099 on Build GRCh38
rs765713886 Chr.8: 79767257-79767257 on Build GRCh38
rs915310023 Chr.8: 79767390-79767390 on Build GRCh38
rs544475862 Chr.8: 79767045-79767045 on Build GRCh38
rs773903559 Chr.8: 79767213-79767213 on Build GRCh38
rs754917036 Chr.8: 79766990-79766990 on Build GRCh38
rs200244794 Chr.8: 79766948-79766949 on Build GRCh38
rs536128981 Chr.8: 79767377-79767377 on Build GRCh38
rs372284042 Chr.8: 79766966-79766966 on Build GRCh38
rs946707801 Chr.8: 79767194-79767194 on Build GRCh38
rs370332199 Chr.8: 79767249-79767249 on Build GRCh38
rs756191310 Chr.8: 79767198-79767198 on Build GRCh38
rs1044986754 Chr.8: 79767200-79767200 on Build GRCh38
rs748794411 Chr.8: 79767205-79767205 on Build GRCh38
rs773703704 Chr.8: 79767354-79767354 on Build GRCh38
rs374460385 Chr.8: 79767118-79767118 on Build GRCh38
rs956816863 Chr.8: 79767151-79767152 on Build GRCh38
rs1041352988 Chr.8: 79767324-79767324 on Build GRCh38
rs777591273 Chr.8: 79767290-79767290 on Build GRCh38
rs777302485 Chr.8: 79767201-79767201 on Build GRCh38
rs376094864 Chr.8: 79767273-79767273 on Build GRCh38
rs773988850 Chr.8: 79767394-79767394 on Build GRCh38
rs368306843 Chr.8: 79767123-79767123 on Build GRCh38
rs758605209 Chr.8: 79766983-79766983 on Build GRCh38
rs1009526761 Chr.8: 79767332-79767332 on Build GRCh38
rs770297237 Chr.8: 79767346-79767346 on Build GRCh38
rs573846021 Chr.8: 79766949-79766949 on Build GRCh38
rs756724011 Chr.8: 79767297-79767297 on Build GRCh38
rs752584773 Chr.8: 79767278-79767278 on Build GRCh38
rs915278876 Chr.8: 79767170-79767170 on Build GRCh38
rs1046777571 Chr.8: 79767298-79767298 on Build GRCh38
rs377204964 Chr.8: 79767120-79767120 on Build GRCh38
rs572924409 Chr.8: 79766978-79766978 on Build GRCh38
rs778265472 Chr.8: 79766946-79766946 on Build GRCh38
rs75113763 Chr.8: 79766972-79766972 on Build GRCh38
rs995120048 Chr.8: 79767063-79767063 on Build GRCh38
rs187014780 Chr.8: 79767313-79767313 on Build GRCh38
rs766557451 Chr.8: 79767269-79767269 on Build GRCh38
rs374953739 Chr.8: 79766951-79766951 on Build GRCh38
rs900082188 Chr.8: 79767341-79767341 on Build GRCh38
rs377726456 Chr.8: 79766984-79766984 on Build GRCh38
rs72684748 Chr.8: 79767150-79767150 on Build GRCh38
rs762552376 Chr.8: 79767300-79767300 on Build GRCh38
rs550465321 Chr.8: 79767317-79767317 on Build GRCh38
rs775295388 Chr.8: 79767178-79767181 on Build GRCh38
rs748536220 Chr.8: 79767343-79767343 on Build GRCh38
rs182968751 Chr.8: 79767294-79767294 on Build GRCh38
rs781514777 Chr.8: 79767328-79767328 on Build GRCh38
rs775899254 Chr.8: 79767101-79767101 on Build GRCh38
rs981286773 Chr.8: 79767216-79767216 on Build GRCh38
rs754827386 Chr.8: 79767138-79767138 on Build GRCh38
rs931065582 Chr.8: 79767292-79767292 on Build GRCh38
rs988215952 Chr.8: 79767378-79767378 on Build GRCh38
rs781311385 Chr.8: 79766992-79766992 on Build GRCh38
rs745485637 Chr.8: 79767241-79767241 on Build GRCh38
rs144046859 Chr.8: 79767323-79767325 on Build GRCh38
rs1020880390 Chr.8: 79767184-79767184 on Build GRCh38
rs967288571 Chr.8: 79767210-79767210 on Build GRCh38
rs769504424 Chr.8: 79767114-79767117 on Build GRCh38
rs751331089 Chr.8: 79767136-79767136 on Build GRCh38
rs547271760 Chr.8: 79767183-79767183 on Build GRCh38
rs374178891 Chr.8: 79767122-79767122 on Build GRCh38
rs761046347 Chr.8: 79767102-79767102 on Build GRCh38
rs768367030 Chr.8: 79767341-79767342 on Build GRCh38
rs759898482 Chr.8: 79767274-79767274 on Build GRCh38

View Full Product Details