Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85245784-85246287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA13
Entrez Gene ID
377677
Gene Name
carbonic anhydrase 13
Gene Aliases
CAXIII
UniGene
Hs.127189
Gene Chromosome Location
Chr.8: 85245487-85284073 on Build GRCh38
Transcripts
NM_198584.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6617095 c.19G>T p.G7* Chr.8: 85245847-85245847 on Build GRCh38
5170078 c.25C>A p.R9S Chr.8: 85245853-85245853 on Build GRCh38
1314241 c.20G>A p.G7E Chr.8: 85245848-85245848 on Build GRCh38
7358828 c.23A>T p.Y8F Chr.8: 85245851-85245851 on Build GRCh38
1755811 c.6G>A p.S2S Chr.8: 85245834-85245834 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758297299 Chr.8: 85245913-85245913 on Build GRCh38
rs996853485 Chr.8: 85246053-85246053 on Build GRCh38
rs902331448 Chr.8: 85245973-85245973 on Build GRCh38
rs1034874397 Chr.8: 85246251-85246251 on Build GRCh38
rs200910539 Chr.8: 85245840-85245840 on Build GRCh38
rs972446878 Chr.8: 85245927-85245927 on Build GRCh38
rs568480544 Chr.8: 85245959-85245959 on Build GRCh38
rs551852757 Chr.8: 85246043-85246043 on Build GRCh38
rs980973777 Chr.8: 85246240-85246240 on Build GRCh38
rs1049342211 Chr.8: 85245949-85245949 on Build GRCh38
rs771516314 Chr.8: 85245819-85245819 on Build GRCh38
rs893727860 Chr.8: 85245926-85245926 on Build GRCh38
rs760123281 Chr.8: 85245854-85245854 on Build GRCh38
rs1031070142 Chr.8: 85246076-85246076 on Build GRCh38
rs142533959 Chr.8: 85245872-85245872 on Build GRCh38
rs765000727 Chr.8: 85245911-85245911 on Build GRCh38
rs775689693 Chr.8: 85245903-85245903 on Build GRCh38
rs1028700589 Chr.8: 85246080-85246080 on Build GRCh38
rs961526371 Chr.8: 85246221-85246221 on Build GRCh38
rs17740658 Chr.8: 85246020-85246020 on Build GRCh38
rs540574779 Chr.8: 85245975-85245975 on Build GRCh38
rs767699457 Chr.8: 85245860-85245860 on Build GRCh38
rs751562410 Chr.8: 85245848-85245849 on Build GRCh38
rs778870555 Chr.8: 85245808-85245808 on Build GRCh38
rs779922342 Chr.8: 85245917-85245917 on Build GRCh38
rs781586323 Chr.8: 85245889-85245890 on Build GRCh38
rs763672760 Chr.8: 85246078-85246078 on Build GRCh38
rs771258100 Chr.8: 85245893-85245893 on Build GRCh38
rs773414651 Chr.8: 85245833-85245833 on Build GRCh38
rs768179962 Chr.8: 85245896-85245896 on Build GRCh38
rs757431331 Chr.8: 85245910-85245910 on Build GRCh38
rs33949682 Chr.8: 85245992-85245996 on Build GRCh38
rs369807031 Chr.8: 85245889-85245889 on Build GRCh38
rs745742507 Chr.8: 85245888-85245888 on Build GRCh38
rs751820737 Chr.8: 85245850-85245851 on Build GRCh38
rs369594146 Chr.8: 85245823-85245823 on Build GRCh38
rs375151255 Chr.8: 85245890-85245890 on Build GRCh38
rs544680746 Chr.8: 85245938-85245938 on Build GRCh38
rs939470337 Chr.8: 85245902-85245902 on Build GRCh38
rs952996817 Chr.8: 85246110-85246110 on Build GRCh38
rs79613391 Chr.8: 85245992-85245992 on Build GRCh38
rs930878447 Chr.8: 85245939-85245939 on Build GRCh38
rs921516691 Chr.8: 85245856-85245856 on Build GRCh38
rs754023052 Chr.8: 85245907-85245907 on Build GRCh38
rs34150174 Chr.8: 85245989-85245993 on Build GRCh38
rs745830476 Chr.8: 85245810-85245810 on Build GRCh38
rs868606075 Chr.8: 85245934-85245934 on Build GRCh38
rs562489285 Chr.8: 85246059-85246059 on Build GRCh38
rs1042214784 Chr.8: 85245963-85245963 on Build GRCh38
rs764110991 Chr.8: 85245906-85245906 on Build GRCh38
rs201498221 Chr.8: 85245912-85245912 on Build GRCh38
rs984350311 Chr.8: 85246147-85246147 on Build GRCh38
rs200216745 Chr.8: 85245853-85245853 on Build GRCh38
rs552303154 Chr.8: 85245830-85245830 on Build GRCh38
rs372593888 Chr.8: 85245891-85245891 on Build GRCh38
rs553239931 Chr.8: 85245895-85245895 on Build GRCh38
rs376366264 Chr.8: 85245820-85245820 on Build GRCh38
rs919293885 Chr.8: 85245928-85245928 on Build GRCh38
rs937719447 Chr.8: 85246255-85246255 on Build GRCh38
rs560449703 Chr.8: 85246017-85246017 on Build GRCh38
rs752881532 Chr.8: 85245865-85245865 on Build GRCh38
rs969301669 Chr.8: 85246222-85246222 on Build GRCh38
rs1022255059 Chr.8: 85246188-85246188 on Build GRCh38
rs756267813 Chr.8: 85245871-85245871 on Build GRCh38
rs900894398 Chr.8: 85246044-85246044 on Build GRCh38
rs200284637 Chr.8: 85245988-85245992 on Build GRCh38
rs764097820 Chr.8: 85245988-85245989 on Build GRCh38
rs559033511 Chr.8: 85245918-85245918 on Build GRCh38
rs532136857 Chr.8: 85245825-85245825 on Build GRCh38
rs531333809 Chr.8: 85246185-85246185 on Build GRCh38
rs768417692 Chr.8: 85245828-85245828 on Build GRCh38
rs1010826013 Chr.8: 85246092-85246092 on Build GRCh38
rs202222221 Chr.8: 85245904-85245904 on Build GRCh38
rs574681472 Chr.8: 85245970-85245970 on Build GRCh38
rs575618795 Chr.8: 85245924-85245924 on Build GRCh38
rs757587958 Chr.8: 85245888-85245889 on Build GRCh38

View Full Product Details