Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85250848-85251354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA13
Entrez Gene ID
377677
Gene Name
carbonic anhydrase 13
Gene Aliases
CAXIII
UniGene
Hs.127189
Gene Chromosome Location
Chr.8: 85245487-85284073 on Build GRCh38
Transcripts
NM_198584.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432406 c.210C>T p.D70D Chr.8: 85250912-85250912 on Build GRCh38
5134028 c.188G>A p.S63N Chr.8: 85250890-85250890 on Build GRCh38
7141878 c.177C>A p.I59I Chr.8: 85250879-85250879 on Build GRCh38
606653 c.201C>T p.F67F Chr.8: 85250903-85250903 on Build GRCh38
5912748 c.235+2T>C p.? Chr.8: 85250939-85250939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111772479 Chr.8: 85251058-85251058 on Build GRCh38
rs907086594 Chr.8: 85251038-85251038 on Build GRCh38
rs7011001 Chr.8: 85251258-85251258 on Build GRCh38
rs773340402 Chr.8: 85250916-85250916 on Build GRCh38
rs556026501 Chr.8: 85251218-85251218 on Build GRCh38
rs34303043 Chr.8: 85251002-85251002 on Build GRCh38
rs1045966687 Chr.8: 85251088-85251088 on Build GRCh38
rs535154003 Chr.8: 85251002-85251003 on Build GRCh38
rs749781343 Chr.8: 85251097-85251097 on Build GRCh38
rs759203734 Chr.8: 85250937-85250937 on Build GRCh38
rs1045551823 Chr.8: 85251034-85251034 on Build GRCh38
rs1009957814 Chr.8: 85251172-85251172 on Build GRCh38
rs974866246 Chr.8: 85251020-85251020 on Build GRCh38
rs373392158 Chr.8: 85250967-85250967 on Build GRCh38
rs567334607 Chr.8: 85251146-85251146 on Build GRCh38
rs906072935 Chr.8: 85251089-85251089 on Build GRCh38
rs370677125 Chr.8: 85250915-85250915 on Build GRCh38
rs982137812 Chr.8: 85251322-85251322 on Build GRCh38
rs1049731588 Chr.8: 85251291-85251291 on Build GRCh38
rs760700227 Chr.8: 85250960-85250960 on Build GRCh38
rs548734922 Chr.8: 85250891-85250891 on Build GRCh38
rs748662901 Chr.8: 85250908-85250908 on Build GRCh38
rs924317262 Chr.8: 85251188-85251188 on Build GRCh38
rs781515388 Chr.8: 85250906-85250906 on Build GRCh38
rs543427419 Chr.8: 85251263-85251263 on Build GRCh38
rs35230947 Chr.8: 85251291-85251292 on Build GRCh38
rs184523200 Chr.8: 85251253-85251253 on Build GRCh38
rs781177535 Chr.8: 85251002-85251004 on Build GRCh38
rs4740046 Chr.8: 85250905-85250905 on Build GRCh38
rs957362546 Chr.8: 85251229-85251229 on Build GRCh38
rs747839649 Chr.8: 85250949-85250949 on Build GRCh38
rs1004248267 Chr.8: 85251042-85251042 on Build GRCh38
rs770338833 Chr.8: 85251003-85251004 on Build GRCh38
rs563822912 Chr.8: 85251265-85251265 on Build GRCh38
rs183655266 Chr.8: 85251127-85251127 on Build GRCh38
rs141188243 Chr.8: 85251126-85251126 on Build GRCh38
rs536596867 Chr.8: 85251101-85251101 on Build GRCh38
rs1045660983 Chr.8: 85251245-85251245 on Build GRCh38
rs1036841999 Chr.8: 85251281-85251281 on Build GRCh38
rs71564210 Chr.8: 85251297-85251298 on Build GRCh38
rs1026792717 Chr.8: 85251254-85251254 on Build GRCh38
rs571228084 Chr.8: 85251065-85251065 on Build GRCh38
rs778663716 Chr.8: 85250948-85250948 on Build GRCh38
rs75498059 Chr.8: 85251003-85251003 on Build GRCh38
rs767376724 Chr.8: 85250939-85250939 on Build GRCh38
rs948675431 Chr.8: 85250978-85250978 on Build GRCh38
rs192326014 Chr.8: 85251244-85251244 on Build GRCh38
rs768148470 Chr.8: 85250913-85250913 on Build GRCh38
rs188706164 Chr.8: 85251238-85251238 on Build GRCh38
rs1020395311 Chr.8: 85251201-85251201 on Build GRCh38
rs752509732 Chr.8: 85250954-85250954 on Build GRCh38
rs978262720 Chr.8: 85251147-85251147 on Build GRCh38
rs1019789391 Chr.8: 85251117-85251117 on Build GRCh38
rs890785534 Chr.8: 85251221-85251221 on Build GRCh38
rs753463298 Chr.8: 85250985-85250985 on Build GRCh38
rs995494154 Chr.8: 85251062-85251062 on Build GRCh38
rs1018202593 Chr.8: 85251237-85251237 on Build GRCh38
rs543721386 Chr.8: 85251019-85251020 on Build GRCh38
rs187543069 Chr.8: 85251047-85251047 on Build GRCh38
rs771183558 Chr.8: 85250934-85250934 on Build GRCh38
rs373427052 Chr.8: 85250892-85250892 on Build GRCh38
rs898549629 Chr.8: 85251282-85251282 on Build GRCh38
rs989819475 Chr.8: 85251234-85251234 on Build GRCh38
rs950070593 Chr.8: 85251314-85251314 on Build GRCh38
rs963866057 Chr.8: 85250922-85250922 on Build GRCh38
rs892894571 Chr.8: 85251169-85251169 on Build GRCh38
rs937508460 Chr.8: 85251116-85251116 on Build GRCh38
rs939820767 Chr.8: 85251279-85251279 on Build GRCh38
rs763159977 Chr.8: 85250920-85250920 on Build GRCh38
rs536335932 Chr.8: 85251151-85251151 on Build GRCh38
rs139111525 Chr.8: 85251284-85251284 on Build GRCh38
rs747806417 Chr.8: 85251048-85251048 on Build GRCh38
rs192392136 Chr.8: 85251064-85251064 on Build GRCh38
rs774174679 Chr.8: 85250936-85250936 on Build GRCh38
rs771733381 Chr.8: 85251292-85251292 on Build GRCh38

View Full Product Details