Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169124795-169125309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B1
Entrez Gene ID
481
Gene Name
ATPase Na+/K+ transporting subunit beta 1
Gene Aliases
ATP1B
UniGene
Hs.291196
Gene Chromosome Location
Chr.1: 169106709-169132722 on Build GRCh38
Transcripts
NM_001677.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899211 c.360C>T p.D120D Chr.1: 169125017-169125017 on Build GRCh38
1689122 c.266C>T p.S89F Chr.1: 169124923-169124923 on Build GRCh38
1683806 c.270delT p.R91fs*13 Chr.1: 169124927-169124927 on Build GRCh38
5010652 c.344C>G p.S115* Chr.1: 169125001-169125001 on Build GRCh38
424512 c.228_229insT p.L77fs*11 Chr.1: 169124885-169124886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967875330 Chr.1: 169125206-169125207 on Build GRCh38
rs1025048888 Chr.1: 169124824-169124824 on Build GRCh38
rs932264213 Chr.1: 169125017-169125017 on Build GRCh38
rs539021907 Chr.1: 169125122-169125122 on Build GRCh38
rs988405768 Chr.1: 169124936-169124936 on Build GRCh38
rs879061760 Chr.1: 169124881-169124881 on Build GRCh38
rs199737584 Chr.1: 169124962-169124962 on Build GRCh38
rs958499891 Chr.1: 169124930-169124930 on Build GRCh38
rs953704341 Chr.1: 169125246-169125246 on Build GRCh38
rs150539946 Chr.1: 169125102-169125102 on Build GRCh38
rs920936371 Chr.1: 169125224-169125224 on Build GRCh38
rs909645017 Chr.1: 169125272-169125272 on Build GRCh38
rs533859904 Chr.1: 169125183-169125183 on Build GRCh38
rs777001068 Chr.1: 169124857-169124862 on Build GRCh38
rs372210037 Chr.1: 169125005-169125005 on Build GRCh38
rs879140975 Chr.1: 169124877-169124877 on Build GRCh38
rs367624941 Chr.1: 169124957-169124957 on Build GRCh38
rs764876165 Chr.1: 169124903-169124903 on Build GRCh38
rs878980414 Chr.1: 169125040-169125040 on Build GRCh38
rs776637135 Chr.1: 169125065-169125065 on Build GRCh38
rs149479006 Chr.1: 169124945-169124945 on Build GRCh38
rs769851426 Chr.1: 169125039-169125039 on Build GRCh38
rs746026374 Chr.1: 169124854-169124854 on Build GRCh38
rs762780258 Chr.1: 169125133-169125133 on Build GRCh38
rs772932534 Chr.1: 169124928-169124928 on Build GRCh38
rs7418916 Chr.1: 169125281-169125281 on Build GRCh38
rs939855669 Chr.1: 169125273-169125273 on Build GRCh38
rs866447952 Chr.1: 169124923-169124923 on Build GRCh38
rs965615019 Chr.1: 169125202-169125202 on Build GRCh38
rs369731734 Chr.1: 169125089-169125089 on Build GRCh38
rs762297318 Chr.1: 169125064-169125065 on Build GRCh38
rs770551991 Chr.1: 169125077-169125077 on Build GRCh38
rs779198300 Chr.1: 169124840-169124840 on Build GRCh38
rs376777964 Chr.1: 169125071-169125071 on Build GRCh38
rs112951922 Chr.1: 169125157-169125157 on Build GRCh38
rs879175226 Chr.1: 169124876-169124876 on Build GRCh38
rs752902404 Chr.1: 169125208-169125208 on Build GRCh38
rs560362746 Chr.1: 169124954-169124954 on Build GRCh38
rs981326343 Chr.1: 169124841-169124841 on Build GRCh38
rs61742560 Chr.1: 169124978-169124978 on Build GRCh38
rs986491406 Chr.1: 169125248-169125248 on Build GRCh38
rs566462232 Chr.1: 169125176-169125176 on Build GRCh38
rs983884930 Chr.1: 169125252-169125252 on Build GRCh38
rs762637917 Chr.1: 169125073-169125073 on Build GRCh38
rs757517281 Chr.1: 169124837-169124837 on Build GRCh38
rs776381919 Chr.1: 169124867-169124867 on Build GRCh38
rs768577639 Chr.1: 169125059-169125059 on Build GRCh38
rs772879342 Chr.1: 169125072-169125072 on Build GRCh38
rs146260113 Chr.1: 169125038-169125038 on Build GRCh38
rs920996589 Chr.1: 169125243-169125243 on Build GRCh38
rs761696878 Chr.1: 169124868-169124868 on Build GRCh38
rs529160002 Chr.1: 169125078-169125078 on Build GRCh38
rs550448239 Chr.1: 169125083-169125083 on Build GRCh38
rs746155696 Chr.1: 169125014-169125014 on Build GRCh38
rs968304 Chr.1: 169124862-169124862 on Build GRCh38
rs1048669015 Chr.1: 169124993-169124993 on Build GRCh38
rs888278486 Chr.1: 169124999-169124999 on Build GRCh38
rs765670633 Chr.1: 169125090-169125090 on Build GRCh38
rs773146131 Chr.1: 169125091-169125091 on Build GRCh38
rs969423073 Chr.1: 169125124-169125124 on Build GRCh38
rs879190761 Chr.1: 169124879-169124879 on Build GRCh38
rs879012016 Chr.1: 169124879-169124879 on Build GRCh38
rs765530612 Chr.1: 169125070-169125071 on Build GRCh38
rs1034877108 Chr.1: 169124836-169124836 on Build GRCh38
rs779129566 Chr.1: 169125012-169125012 on Build GRCh38
rs754423635 Chr.1: 169125086-169125086 on Build GRCh38
rs1892094 Chr.1: 169125221-169125221 on Build GRCh38
rs772333448 Chr.1: 169124855-169124855 on Build GRCh38
rs142696549 Chr.1: 169125184-169125184 on Build GRCh38
rs746983798 Chr.1: 169124858-169124858 on Build GRCh38
rs184720505 Chr.1: 169124827-169124827 on Build GRCh38
rs759081669 Chr.1: 169125079-169125079 on Build GRCh38
rs751220188 Chr.1: 169125082-169125082 on Build GRCh38
rs1002133333 Chr.1: 169125176-169125177 on Build GRCh38
rs7418915 Chr.1: 169125278-169125278 on Build GRCh38
rs897990101 Chr.1: 169125145-169125145 on Build GRCh38
rs932342972 Chr.1: 169125229-169125229 on Build GRCh38
rs758661489 Chr.1: 169125037-169125037 on Build GRCh38
rs762580778 Chr.1: 169124943-169124943 on Build GRCh38
rs944979208 Chr.1: 169125267-169125267 on Build GRCh38
rs1013562207 Chr.1: 169125081-169125081 on Build GRCh38
rs932420338 Chr.1: 169124989-169124989 on Build GRCh38
rs767810489 Chr.1: 169125151-169125151 on Build GRCh38

View Full Product Details