Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169512757-169513303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
UniGene
Hs.30054
Gene Chromosome Location
Chr.1: 169511954-169586632 on Build GRCh38
Transcripts
NM_000130.4 XM_017000660.1

SNP Detail

SNP ID Chromosome Location
rs1053316903 Chr.1: 169512796-169512796 on Build GRCh38
rs777095170 Chr.1: 169512960-169512960 on Build GRCh38
rs114322489 Chr.1: 169513242-169513242 on Build GRCh38
rs1009033117 Chr.1: 169512859-169512859 on Build GRCh38
rs549775870 Chr.1: 169513163-169513163 on Build GRCh38
rs773220210 Chr.1: 169513097-169513097 on Build GRCh38
rs960421178 Chr.1: 169513249-169513249 on Build GRCh38
rs886103412 Chr.1: 169513214-169513214 on Build GRCh38
rs921344321 Chr.1: 169512841-169512841 on Build GRCh38
rs543442430 Chr.1: 169512892-169512892 on Build GRCh38
rs760001405 Chr.1: 169513224-169513224 on Build GRCh38
rs1034855835 Chr.1: 169512963-169512963 on Build GRCh38
rs951313531 Chr.1: 169513077-169513077 on Build GRCh38
rs544989242 Chr.1: 169512989-169512989 on Build GRCh38
rs960530541 Chr.1: 169512980-169512980 on Build GRCh38
rs995324094 Chr.1: 169513057-169513057 on Build GRCh38
rs975809603 Chr.1: 169513149-169513149 on Build GRCh38
rs1057407526 Chr.1: 169513250-169513250 on Build GRCh38
rs770031746 Chr.1: 169513084-169513084 on Build GRCh38
rs562419214 Chr.1: 169512908-169512908 on Build GRCh38
rs994928768 Chr.1: 169512881-169512881 on Build GRCh38
rs527497923 Chr.1: 169513056-169513056 on Build GRCh38
rs188534160 Chr.1: 169513212-169513212 on Build GRCh38
rs991084730 Chr.1: 169513270-169513270 on Build GRCh38
rs2040444 Chr.1: 169513198-169513198 on Build GRCh38
rs983205384 Chr.1: 169512784-169512784 on Build GRCh38
rs1004465875 Chr.1: 169513235-169513235 on Build GRCh38
rs9332677 Chr.1: 169513023-169513023 on Build GRCh38
rs755056508 Chr.1: 169512970-169512979 on Build GRCh38
rs886045536 Chr.1: 169512881-169512881 on Build GRCh38
rs578023501 Chr.1: 169512873-169512873 on Build GRCh38
rs533224248 Chr.1: 169512918-169512918 on Build GRCh38
rs770927759 Chr.1: 169513173-169513173 on Build GRCh38
rs554692992 Chr.1: 169512839-169512839 on Build GRCh38
rs950549168 Chr.1: 169512909-169512909 on Build GRCh38
rs1004098750 Chr.1: 169513104-169513104 on Build GRCh38
rs536489141 Chr.1: 169513271-169513271 on Build GRCh38
rs912534869 Chr.1: 169512790-169512790 on Build GRCh38
rs995086245 Chr.1: 169513204-169513204 on Build GRCh38
rs1052541127 Chr.1: 169512940-169512940 on Build GRCh38
rs944867236 Chr.1: 169513191-169513191 on Build GRCh38
rs185213313 Chr.1: 169513181-169513181 on Build GRCh38
rs191866237 Chr.1: 169513143-169513143 on Build GRCh38
rs562332594 Chr.1: 169513144-169513144 on Build GRCh38
rs1020034399 Chr.1: 169513132-169513132 on Build GRCh38
rs9332675 Chr.1: 169513195-169513195 on Build GRCh38
rs567404075 Chr.1: 169513113-169513113 on Build GRCh38
rs9332676 Chr.1: 169513067-169513067 on Build GRCh38
rs965444910 Chr.1: 169513138-169513138 on Build GRCh38
rs934872183 Chr.1: 169512936-169512936 on Build GRCh38
rs75764442 Chr.1: 169512825-169512825 on Build GRCh38
rs1027302805 Chr.1: 169512895-169512895 on Build GRCh38
rs979200861 Chr.1: 169513076-169513076 on Build GRCh38
rs9332678 Chr.1: 169512877-169512877 on Build GRCh38
rs1044855495 Chr.1: 169513004-169513004 on Build GRCh38
rs965852797 Chr.1: 169512799-169512799 on Build GRCh38
rs572562882 Chr.1: 169512788-169512788 on Build GRCh38

View Full Product Details