Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169523698-169524222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
UniGene
Hs.30054
Gene Chromosome Location
Chr.1: 169511954-169586632 on Build GRCh38
Transcripts
NM_000130.4 XM_017000660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5930522 c.5828G>A p.G1943E Chr.1: 169523865-169523865 on Build GRCh38
2073526 c.5847T>G p.S1949R Chr.1: 169523846-169523846 on Build GRCh38
6059169 c.5835T>A p.Y1945* Chr.1: 169523858-169523858 on Build GRCh38
6657746 c.5876C>A p.S1959Y Chr.1: 169523817-169523817 on Build GRCh38
1162853 c.5882C>T p.P1961L Chr.1: 169523811-169523811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761177380 Chr.1: 169523847-169523847 on Build GRCh38
rs991306620 Chr.1: 169523754-169523754 on Build GRCh38
rs377182698 Chr.1: 169523904-169523904 on Build GRCh38
rs893963202 Chr.1: 169524001-169524001 on Build GRCh38
rs541674012 Chr.1: 169523722-169523722 on Build GRCh38
rs556874827 Chr.1: 169524010-169524010 on Build GRCh38
rs557869574 Chr.1: 169523846-169523846 on Build GRCh38
rs762931699 Chr.1: 169523879-169523879 on Build GRCh38
rs535212435 Chr.1: 169524009-169524009 on Build GRCh38
rs752507138 Chr.1: 169523916-169523916 on Build GRCh38
rs955614785 Chr.1: 169523934-169523934 on Build GRCh38
rs918321117 Chr.1: 169524047-169524047 on Build GRCh38
rs766428271 Chr.1: 169523893-169523893 on Build GRCh38
rs1017733019 Chr.1: 169523999-169523999 on Build GRCh38
rs747821500 Chr.1: 169523775-169523775 on Build GRCh38
rs967038173 Chr.1: 169524062-169524062 on Build GRCh38
rs1008154044 Chr.1: 169523965-169523965 on Build GRCh38
rs372998280 Chr.1: 169523830-169523830 on Build GRCh38
rs977031849 Chr.1: 169524082-169524082 on Build GRCh38
rs747492560 Chr.1: 169523953-169523953 on Build GRCh38
rs778447032 Chr.1: 169523952-169523952 on Build GRCh38
rs748911956 Chr.1: 169523939-169523939 on Build GRCh38
rs781090521 Chr.1: 169523768-169523768 on Build GRCh38
rs755895612 Chr.1: 169523919-169523919 on Build GRCh38
rs765415797 Chr.1: 169523868-169523868 on Build GRCh38
rs754746857 Chr.1: 169523767-169523767 on Build GRCh38
rs922886168 Chr.1: 169523845-169523845 on Build GRCh38
rs781178931 Chr.1: 169523915-169523915 on Build GRCh38
rs776042028 Chr.1: 169523921-169523922 on Build GRCh38
rs991736975 Chr.1: 169524186-169524186 on Build GRCh38
rs770351842 Chr.1: 169523951-169523951 on Build GRCh38
rs748741328 Chr.1: 169523812-169523812 on Build GRCh38
rs938530386 Chr.1: 169523731-169523731 on Build GRCh38
rs897863606 Chr.1: 169523926-169523926 on Build GRCh38
rs577827052 Chr.1: 169523862-169523862 on Build GRCh38
rs1035859011 Chr.1: 169524107-169524107 on Build GRCh38
rs967298104 Chr.1: 169523806-169523806 on Build GRCh38
rs373701795 Chr.1: 169523954-169523954 on Build GRCh38
rs140426662 Chr.1: 169523790-169523790 on Build GRCh38
rs978211268 Chr.1: 169523821-169523821 on Build GRCh38
rs149988964 Chr.1: 169524086-169524086 on Build GRCh38
rs1038543167 Chr.1: 169524030-169524030 on Build GRCh38
rs941004434 Chr.1: 169524016-169524016 on Build GRCh38
rs762072342 Chr.1: 169523865-169523865 on Build GRCh38
rs779854404 Chr.1: 169523764-169523764 on Build GRCh38
rs764474317 Chr.1: 169523858-169523858 on Build GRCh38
rs910890622 Chr.1: 169523994-169523994 on Build GRCh38
rs1011146905 Chr.1: 169524027-169524027 on Build GRCh38
rs754891246 Chr.1: 169523909-169523909 on Build GRCh38
rs758403579 Chr.1: 169523756-169523756 on Build GRCh38
rs865867876 Chr.1: 169524017-169524017 on Build GRCh38
rs777021256 Chr.1: 169523955-169523955 on Build GRCh38
rs769546453 Chr.1: 169523781-169523781 on Build GRCh38
rs1045581766 Chr.1: 169524109-169524109 on Build GRCh38
rs776933032 Chr.1: 169523863-169523863 on Build GRCh38
rs907156853 Chr.1: 169524112-169524112 on Build GRCh38
rs746882392 Chr.1: 169523765-169523765 on Build GRCh38
rs777553363 Chr.1: 169523932-169523932 on Build GRCh38
rs750570307 Chr.1: 169523877-169523877 on Build GRCh38

View Full Product Details