Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171107997-171108501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO3
Entrez Gene ID
2328
Gene Name
flavin containing monooxygenase 3
Gene Aliases
FMOII, TMAU, dJ127D3.1
UniGene
Hs.445350
Gene Chromosome Location
Chr.1: 171090873-171117819 on Build GRCh38
Transcripts
NM_001002294.2 NM_001319173.1 NM_001319174.1 NM_006894.5 XM_011509345.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665624 c.573C>T p.G191G Chr.1: 171108167-171108167 on Build GRCh38
899479 c.533A>T p.E178V Chr.1: 171108127-171108127 on Build GRCh38
7164222 c.546C>T p.F182F Chr.1: 171108140-171108140 on Build GRCh38
1159366 c.585G>A p.S195S Chr.1: 171108179-171108179 on Build GRCh38
5990863 c.626+1delG p.? Chr.1: 171108221-171108221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3832024 Chr.1: 171108185-171108186 on Build GRCh38
rs370429033 Chr.1: 171108044-171108044 on Build GRCh38
rs780449159 Chr.1: 171108214-171108214 on Build GRCh38
rs531808372 Chr.1: 171108321-171108321 on Build GRCh38
rs773652235 Chr.1: 171108249-171108249 on Build GRCh38
rs200165797 Chr.1: 171108103-171108103 on Build GRCh38
rs2066529 Chr.1: 171108024-171108024 on Build GRCh38
rs1010461803 Chr.1: 171108422-171108422 on Build GRCh38
rs1041647720 Chr.1: 171108423-171108423 on Build GRCh38
rs762717250 Chr.1: 171108266-171108266 on Build GRCh38
rs1920149 Chr.1: 171108057-171108057 on Build GRCh38
rs548360460 Chr.1: 171108293-171108293 on Build GRCh38
rs116425430 Chr.1: 171108172-171108172 on Build GRCh38
rs758324579 Chr.1: 171108121-171108121 on Build GRCh38
rs72549327 Chr.1: 171108190-171108190 on Build GRCh38
rs371891579 Chr.1: 171108174-171108174 on Build GRCh38
rs933630464 Chr.1: 171108065-171108065 on Build GRCh38
rs202077178 Chr.1: 171108234-171108234 on Build GRCh38
rs28363550 Chr.1: 171108362-171108362 on Build GRCh38
rs778745952 Chr.1: 171108115-171108115 on Build GRCh38
rs753378707 Chr.1: 171108111-171108111 on Build GRCh38
rs764574402 Chr.1: 171108102-171108102 on Build GRCh38
rs529940450 Chr.1: 171108188-171108188 on Build GRCh38
rs146771691 Chr.1: 171108367-171108367 on Build GRCh38
rs539058927 Chr.1: 171108150-171108150 on Build GRCh38
rs773423537 Chr.1: 171108307-171108307 on Build GRCh38
rs200758892 Chr.1: 171108366-171108366 on Build GRCh38
rs201459120 Chr.1: 171108114-171108114 on Build GRCh38
rs780358952 Chr.1: 171108183-171108184 on Build GRCh38
rs980442718 Chr.1: 171108021-171108021 on Build GRCh38
rs889501001 Chr.1: 171108097-171108097 on Build GRCh38
rs774316645 Chr.1: 171108432-171108432 on Build GRCh38
rs906000525 Chr.1: 171108439-171108439 on Build GRCh38
rs752341553 Chr.1: 171108259-171108259 on Build GRCh38
rs777725103 Chr.1: 171108227-171108227 on Build GRCh38
rs866675988 Chr.1: 171108144-171108144 on Build GRCh38
rs765930962 Chr.1: 171108109-171108109 on Build GRCh38
rs758617515 Chr.1: 171108173-171108174 on Build GRCh38
rs764845113 Chr.1: 171108253-171108253 on Build GRCh38
rs148029783 Chr.1: 171108193-171108193 on Build GRCh38
rs768956786 Chr.1: 171108236-171108236 on Build GRCh38
rs2066534 Chr.1: 171108231-171108231 on Build GRCh38
rs918689663 Chr.1: 171108389-171108389 on Build GRCh38
rs559643079 Chr.1: 171108216-171108216 on Build GRCh38
rs752515583 Chr.1: 171108116-171108116 on Build GRCh38
rs28363549 Chr.1: 171108207-171108207 on Build GRCh38
rs765771190 Chr.1: 171108196-171108196 on Build GRCh38
rs72549328 Chr.1: 171108198-171108199 on Build GRCh38
rs751413078 Chr.1: 171108230-171108230 on Build GRCh38
rs766154092 Chr.1: 171108402-171108402 on Build GRCh38
rs764972414 Chr.1: 171108208-171108208 on Build GRCh38
rs549999748 Chr.1: 171108473-171108473 on Build GRCh38
rs750840253 Chr.1: 171108452-171108452 on Build GRCh38
rs79952472 Chr.1: 171108252-171108252 on Build GRCh38
rs144283823 Chr.1: 171108179-171108179 on Build GRCh38
rs907080450 Chr.1: 171108445-171108445 on Build GRCh38
rs765365523 Chr.1: 171108447-171108451 on Build GRCh38
rs397844257 Chr.1: 171108365-171108365 on Build GRCh38
rs745663963 Chr.1: 171108146-171108146 on Build GRCh38
rs944050600 Chr.1: 171108362-171108362 on Build GRCh38
rs775749562 Chr.1: 171108178-171108178 on Build GRCh38
rs531988135 Chr.1: 171108233-171108233 on Build GRCh38
rs200924412 Chr.1: 171108060-171108060 on Build GRCh38
rs910885777 Chr.1: 171108302-171108302 on Build GRCh38
rs966753510 Chr.1: 171108161-171108161 on Build GRCh38
rs143406401 Chr.1: 171108151-171108151 on Build GRCh38
rs143703134 Chr.1: 171108294-171108294 on Build GRCh38
rs759826525 Chr.1: 171108056-171108056 on Build GRCh38
rs75904274 Chr.1: 171108133-171108133 on Build GRCh38
rs769983049 Chr.1: 171108154-171108154 on Build GRCh38
rs368194307 Chr.1: 171108143-171108143 on Build GRCh38
rs28363551 Chr.1: 171108465-171108465 on Build GRCh38
rs932535169 Chr.1: 171108131-171108131 on Build GRCh38
rs774813678 Chr.1: 171108241-171108241 on Build GRCh38
rs530013323 Chr.1: 171108285-171108285 on Build GRCh38
rs1050815673 Chr.1: 171108088-171108088 on Build GRCh38
rs758201900 Chr.1: 171108222-171108222 on Build GRCh38
rs985332444 Chr.1: 171108300-171108300 on Build GRCh38
rs753360820 Chr.1: 171108206-171108206 on Build GRCh38
rs763358926 Chr.1: 171108068-171108068 on Build GRCh38
rs150042882 Chr.1: 171108041-171108041 on Build GRCh38
rs79233811 Chr.1: 171108400-171108400 on Build GRCh38
rs910127258 Chr.1: 171108038-171108038 on Build GRCh38

View Full Product Details