Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171114272-171114780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO3
Entrez Gene ID
2328
Gene Name
flavin containing monooxygenase 3
Gene Aliases
FMOII, TMAU, dJ127D3.1
UniGene
Hs.445350
Gene Chromosome Location
Chr.1: 171090873-171117819 on Build GRCh38
Transcripts
NM_001002294.2 NM_001319173.1 NM_001319174.1 NM_006894.5 XM_011509345.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4025045 c.1130C>A p.A377D Chr.1: 171114309-171114309 on Build GRCh38
7401065 c.1127G>T p.G376V Chr.1: 171114306-171114306 on Build GRCh38
3478249 c.1122C>T p.S374S Chr.1: 171114301-171114301 on Build GRCh38
352854 c.1132G>C p.A378P Chr.1: 171114311-171114311 on Build GRCh38
6665637 c.1094C>T p.T365I Chr.1: 171114273-171114273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750002345 Chr.1: 171114343-171114343 on Build GRCh38
rs909531 Chr.1: 171114397-171114397 on Build GRCh38
rs959922161 Chr.1: 171114645-171114645 on Build GRCh38
rs1054357877 Chr.1: 171114750-171114750 on Build GRCh38
rs563268847 Chr.1: 171114432-171114432 on Build GRCh38
rs966877714 Chr.1: 171114579-171114579 on Build GRCh38
rs918301146 Chr.1: 171114715-171114715 on Build GRCh38
rs377549018 Chr.1: 171114401-171114401 on Build GRCh38
rs932781687 Chr.1: 171114425-171114425 on Build GRCh38
rs145526965 Chr.1: 171114338-171114338 on Build GRCh38
rs74854600 Chr.1: 171114742-171114742 on Build GRCh38
rs750504636 Chr.1: 171114584-171114584 on Build GRCh38
rs1033841334 Chr.1: 171114641-171114641 on Build GRCh38
rs982648128 Chr.1: 171114585-171114585 on Build GRCh38
rs755888463 Chr.1: 171114749-171114749 on Build GRCh38
rs28363584 Chr.1: 171114748-171114748 on Build GRCh38
rs139647910 Chr.1: 171114673-171114673 on Build GRCh38
rs760983079 Chr.1: 171114309-171114309 on Build GRCh38
rs775064534 Chr.1: 171114317-171114318 on Build GRCh38
rs1048747040 Chr.1: 171114550-171114550 on Build GRCh38
rs185565199 Chr.1: 171114560-171114560 on Build GRCh38
rs944195309 Chr.1: 171114491-171114491 on Build GRCh38
rs28363583 Chr.1: 171114707-171114707 on Build GRCh38
rs754884355 Chr.1: 171114372-171114372 on Build GRCh38
rs767079173 Chr.1: 171114296-171114297 on Build GRCh38
rs771164149 Chr.1: 171114398-171114398 on Build GRCh38
rs531835715 Chr.1: 171114559-171114559 on Build GRCh38
rs1018051029 Chr.1: 171114485-171114485 on Build GRCh38
rs909532 Chr.1: 171114574-171114574 on Build GRCh38
rs779619155 Chr.1: 171114362-171114362 on Build GRCh38
rs912763847 Chr.1: 171114479-171114479 on Build GRCh38
rs373403267 Chr.1: 171114400-171114400 on Build GRCh38
rs1037161827 Chr.1: 171114493-171114493 on Build GRCh38
rs991072456 Chr.1: 171114652-171114652 on Build GRCh38
rs925246548 Chr.1: 171114404-171114404 on Build GRCh38
rs529912807 Chr.1: 171114391-171114391 on Build GRCh38
rs199975586 Chr.1: 171114342-171114342 on Build GRCh38
rs775927424 Chr.1: 171114410-171114410 on Build GRCh38
rs28363582 Chr.1: 171114390-171114390 on Build GRCh38
rs889742130 Chr.1: 171114435-171114435 on Build GRCh38
rs887420032 Chr.1: 171114567-171114567 on Build GRCh38
rs764326621 Chr.1: 171114392-171114392 on Build GRCh38
rs897428950 Chr.1: 171114512-171114512 on Build GRCh38
rs941007156 Chr.1: 171114690-171114690 on Build GRCh38
rs1055235589 Chr.1: 171114330-171114330 on Build GRCh38
rs770157102 Chr.1: 171114407-171114407 on Build GRCh38
rs201860024 Chr.1: 171114324-171114324 on Build GRCh38
rs771161406 Chr.1: 171114312-171114312 on Build GRCh38
rs561326379 Chr.1: 171114481-171114481 on Build GRCh38
rs1006799840 Chr.1: 171114457-171114457 on Build GRCh38
rs754992629 Chr.1: 171114361-171114361 on Build GRCh38
rs751174584 Chr.1: 171114539-171114539 on Build GRCh38
rs867914992 Chr.1: 171114301-171114301 on Build GRCh38
rs755757453 Chr.1: 171114353-171114353 on Build GRCh38
rs72549331 Chr.1: 171114339-171114339 on Build GRCh38
rs753668825 Chr.1: 171114367-171114367 on Build GRCh38
rs1011017166 Chr.1: 171114551-171114551 on Build GRCh38

View Full Product Details