Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171117409-171117914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO3
Entrez Gene ID
2328
Gene Name
flavin containing monooxygenase 3
Gene Aliases
FMOII, TMAU, dJ127D3.1
UniGene
Hs.445350
Gene Chromosome Location
Chr.1: 171090873-171117819 on Build GRCh38
Transcripts
NM_001002294.2 NM_001319173.1 NM_001319174.1 NM_006894.5 XM_011509345.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
677197 c.1568T>C p.L523P Chr.1: 171117411-171117411 on Build GRCh38
5378400 c.1584C>T p.F528F Chr.1: 171117427-171117427 on Build GRCh38
6334470 c.1576G>T p.A526S Chr.1: 171117419-171117419 on Build GRCh38
2074365 c.1567C>A p.L523M Chr.1: 171117410-171117410 on Build GRCh38
7189774 c.1586T>G p.L529R Chr.1: 171117429-171117429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983202884 Chr.1: 171117657-171117657 on Build GRCh38
rs898251442 Chr.1: 171117871-171117871 on Build GRCh38
rs933598609 Chr.1: 171117781-171117781 on Build GRCh38
rs1012605194 Chr.1: 171117876-171117876 on Build GRCh38
rs956211752 Chr.1: 171117526-171117526 on Build GRCh38
rs35949891 Chr.1: 171117863-171117864 on Build GRCh38
rs990133512 Chr.1: 171117694-171117694 on Build GRCh38
rs964803205 Chr.1: 171117703-171117703 on Build GRCh38
rs143079250 Chr.1: 171117483-171117483 on Build GRCh38
rs969696261 Chr.1: 171117557-171117557 on Build GRCh38
rs762333220 Chr.1: 171117815-171117819 on Build GRCh38
rs771349442 Chr.1: 171117806-171117806 on Build GRCh38
rs931879543 Chr.1: 171117854-171117854 on Build GRCh38
rs1014931444 Chr.1: 171117528-171117528 on Build GRCh38
rs547257101 Chr.1: 171117653-171117653 on Build GRCh38
rs542162053 Chr.1: 171117440-171117440 on Build GRCh38
rs1024523917 Chr.1: 171117555-171117555 on Build GRCh38
rs921884854 Chr.1: 171117818-171117818 on Build GRCh38
rs536146962 Chr.1: 171117697-171117697 on Build GRCh38
rs1000270017 Chr.1: 171117478-171117478 on Build GRCh38
rs777464034 Chr.1: 171117494-171117494 on Build GRCh38
rs904644047 Chr.1: 171117458-171117458 on Build GRCh38
rs1038014897 Chr.1: 171117865-171117865 on Build GRCh38
rs569734212 Chr.1: 171117822-171117822 on Build GRCh38
rs199556953 Chr.1: 171117449-171117449 on Build GRCh38
rs764458425 Chr.1: 171117438-171117438 on Build GRCh38
rs971318248 Chr.1: 171117595-171117595 on Build GRCh38
rs951827144 Chr.1: 171117637-171117637 on Build GRCh38
rs374847347 Chr.1: 171117604-171117604 on Build GRCh38
rs182196901 Chr.1: 171117619-171117619 on Build GRCh38
rs923414253 Chr.1: 171117714-171117714 on Build GRCh38
rs529018130 Chr.1: 171117858-171117858 on Build GRCh38
rs151134994 Chr.1: 171117879-171117879 on Build GRCh38
rs201986708 Chr.1: 171117446-171117446 on Build GRCh38
rs747699677 Chr.1: 171117473-171117473 on Build GRCh38
rs41272509 Chr.1: 171117692-171117692 on Build GRCh38
rs199938001 Chr.1: 171117500-171117500 on Build GRCh38
rs192665764 Chr.1: 171117883-171117883 on Build GRCh38
rs974921816 Chr.1: 171117710-171117710 on Build GRCh38
rs1032282290 Chr.1: 171117503-171117503 on Build GRCh38
rs528695978 Chr.1: 171117632-171117632 on Build GRCh38
rs575861495 Chr.1: 171117459-171117459 on Build GRCh38
rs187172674 Chr.1: 171117816-171117816 on Build GRCh38
rs1002728837 Chr.1: 171117598-171117598 on Build GRCh38
rs774866490 Chr.1: 171117847-171117847 on Build GRCh38
rs907719155 Chr.1: 171117762-171117762 on Build GRCh38
rs1000301316 Chr.1: 171117878-171117878 on Build GRCh38
rs778423685 Chr.1: 171117460-171117460 on Build GRCh38
rs371727643 Chr.1: 171117455-171117455 on Build GRCh38

View Full Product Details