Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94495524-94496064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
UniGene
Hs.202238
Gene Chromosome Location
Chr.8: 94487777-94553518 on Build GRCh38
Transcripts
NM_015496.4 XM_017013306.1 XM_017013307.1 XM_017013308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5049601 c.4384-2A>G p.? Chr.8: 94495893-94495893 on Build GRCh38
1314274 c.4479C>T p.L1493L Chr.8: 94495796-94495796 on Build GRCh38
7107624 c.4425C>T p.D1475D Chr.8: 94495850-94495850 on Build GRCh38
82212 c.4419G>A p.L1473L Chr.8: 94495856-94495856 on Build GRCh38
7161073 c.4399G>A p.D1467N Chr.8: 94495876-94495876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550923988 Chr.8: 94495959-94495959 on Build GRCh38
rs966463140 Chr.8: 94495662-94495662 on Build GRCh38
rs959246119 Chr.8: 94495608-94495608 on Build GRCh38
rs766989822 Chr.8: 94495684-94495684 on Build GRCh38
rs781392977 Chr.8: 94495758-94495758 on Build GRCh38
rs910808286 Chr.8: 94495721-94495721 on Build GRCh38
rs377326310 Chr.8: 94495672-94495672 on Build GRCh38
rs770722200 Chr.8: 94495812-94495812 on Build GRCh38
rs981375822 Chr.8: 94495558-94495558 on Build GRCh38
rs781483224 Chr.8: 94495905-94495905 on Build GRCh38
rs566504056 Chr.8: 94495791-94495791 on Build GRCh38
rs397724816 Chr.8: 94495575-94495576 on Build GRCh38
rs889140472 Chr.8: 94495618-94495618 on Build GRCh38
rs199555299 Chr.8: 94495830-94495830 on Build GRCh38
rs774179315 Chr.8: 94495814-94495814 on Build GRCh38
rs370520816 Chr.8: 94495924-94495924 on Build GRCh38
rs1022109714 Chr.8: 94496022-94496022 on Build GRCh38
rs1033593669 Chr.8: 94495606-94495606 on Build GRCh38
rs946826617 Chr.8: 94495605-94495605 on Build GRCh38
rs564021786 Chr.8: 94495665-94495665 on Build GRCh38
rs546605218 Chr.8: 94495771-94495771 on Build GRCh38
rs34942724 Chr.8: 94495557-94495558 on Build GRCh38
rs1006410133 Chr.8: 94495667-94495667 on Build GRCh38
rs766747251 Chr.8: 94495884-94495884 on Build GRCh38
rs11382498 Chr.8: 94495558-94495559 on Build GRCh38
rs777890228 Chr.8: 94495927-94495927 on Build GRCh38
rs1010747884 Chr.8: 94495996-94495996 on Build GRCh38
rs758718932 Chr.8: 94495728-94495728 on Build GRCh38
rs774968127 Chr.8: 94495838-94495838 on Build GRCh38
rs767079433 Chr.8: 94495834-94495834 on Build GRCh38
rs1044215155 Chr.8: 94495962-94495962 on Build GRCh38
rs943579629 Chr.8: 94495722-94495722 on Build GRCh38
rs929835813 Chr.8: 94495952-94495952 on Build GRCh38
rs760120023 Chr.8: 94495851-94495851 on Build GRCh38
rs750762864 Chr.8: 94495723-94495723 on Build GRCh38
rs752838511 Chr.8: 94495912-94495912 on Build GRCh38
rs774170277 Chr.8: 94495961-94495961 on Build GRCh38
rs763737840 Chr.8: 94495855-94495855 on Build GRCh38
rs1003909596 Chr.8: 94495598-94495598 on Build GRCh38
rs755542446 Chr.8: 94495896-94495896 on Build GRCh38
rs755161314 Chr.8: 94495756-94495756 on Build GRCh38
rs752384436 Chr.8: 94495692-94495692 on Build GRCh38
rs1022059309 Chr.8: 94495655-94495655 on Build GRCh38
rs2289457 Chr.8: 94496033-94496033 on Build GRCh38
rs200621602 Chr.8: 94495806-94495806 on Build GRCh38
rs910522124 Chr.8: 94495974-94495974 on Build GRCh38
rs993627013 Chr.8: 94495643-94495643 on Build GRCh38
rs946804930 Chr.8: 94495929-94495929 on Build GRCh38
rs955981011 Chr.8: 94495774-94495774 on Build GRCh38
rs755758929 Chr.8: 94495693-94495693 on Build GRCh38
rs943378909 Chr.8: 94495998-94495998 on Build GRCh38
rs988656417 Chr.8: 94495823-94495823 on Build GRCh38
rs201843078 Chr.8: 94495937-94495937 on Build GRCh38
rs1026761342 Chr.8: 94495569-94495569 on Build GRCh38
rs747497631 Chr.8: 94495746-94495746 on Build GRCh38
rs901982636 Chr.8: 94495638-94495638 on Build GRCh38
rs1007740165 Chr.8: 94495619-94495619 on Build GRCh38
rs752049792 Chr.8: 94495889-94495889 on Build GRCh38
rs76642335 Chr.8: 94495621-94495621 on Build GRCh38
rs765647210 Chr.8: 94495697-94495697 on Build GRCh38
rs770130410 Chr.8: 94495773-94495773 on Build GRCh38
rs372944194 Chr.8: 94495744-94495744 on Build GRCh38
rs934489646 Chr.8: 94495587-94495587 on Build GRCh38
rs570567345 Chr.8: 94495647-94495648 on Build GRCh38
rs770670787 Chr.8: 94495938-94495938 on Build GRCh38
rs200793962 Chr.8: 94495558-94495558 on Build GRCh38
rs369765494 Chr.8: 94495681-94495681 on Build GRCh38
rs906515760 Chr.8: 94495594-94495594 on Build GRCh38
rs367623928 Chr.8: 94495794-94495794 on Build GRCh38
rs142534601 Chr.8: 94495675-94495675 on Build GRCh38
rs552364335 Chr.8: 94495555-94495556 on Build GRCh38
rs551953235 Chr.8: 94495646-94495646 on Build GRCh38
rs753487426 Chr.8: 94495863-94495863 on Build GRCh38
rs928425372 Chr.8: 94495559-94495559 on Build GRCh38
rs1052909836 Chr.8: 94495589-94495589 on Build GRCh38
rs763246506 Chr.8: 94495869-94495869 on Build GRCh38
rs989274803 Chr.8: 94495649-94495649 on Build GRCh38
rs969949241 Chr.8: 94495557-94495558 on Build GRCh38

View Full Product Details