Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 129839391-129839889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM49B
Entrez Gene ID
51571
Gene Name
family with sequence similarity 49 member B
Gene Aliases
BM-009, L1
UniGene
Hs.126941
Gene Chromosome Location
Chr.8: 129839593-130016697 on Build GRCh38
Transcripts
NM_001256763.1 NM_016623.4 NR_046359.1 NR_046360.1 NR_046361.1

SNP Detail

SNP ID Chromosome Location
rs572745100 Chr.8: 129839563-129839566 on Build GRCh38
rs984275009 Chr.8: 129839669-129839671 on Build GRCh38
rs775739199 Chr.8: 129839471-129839471 on Build GRCh38
rs948482128 Chr.8: 129839714-129839714 on Build GRCh38
rs968923874 Chr.8: 129839466-129839466 on Build GRCh38
rs191062044 Chr.8: 129839754-129839754 on Build GRCh38
rs541572209 Chr.8: 129839733-129839733 on Build GRCh38
rs762034470 Chr.8: 129839595-129839595 on Build GRCh38
rs1041811799 Chr.8: 129839480-129839480 on Build GRCh38
rs1055552081 Chr.8: 129839766-129839766 on Build GRCh38
rs763150102 Chr.8: 129839484-129839484 on Build GRCh38
rs942961323 Chr.8: 129839411-129839411 on Build GRCh38
rs117458905 Chr.8: 129839692-129839692 on Build GRCh38
rs570746049 Chr.8: 129839452-129839452 on Build GRCh38
rs946079887 Chr.8: 129839735-129839735 on Build GRCh38
rs866559642 Chr.8: 129839737-129839737 on Build GRCh38
rs553443944 Chr.8: 129839736-129839736 on Build GRCh38
rs115949023 Chr.8: 129839834-129839834 on Build GRCh38
rs146943965 Chr.8: 129839800-129839800 on Build GRCh38
rs1007622551 Chr.8: 129839802-129839802 on Build GRCh38
rs1055583775 Chr.8: 129839517-129839517 on Build GRCh38
rs758130596 Chr.8: 129839826-129839826 on Build GRCh38
rs977034202 Chr.8: 129839723-129839723 on Build GRCh38
rs961745668 Chr.8: 129839581-129839581 on Build GRCh38
rs145529695 Chr.8: 129839444-129839444 on Build GRCh38
rs1051403158 Chr.8: 129839777-129839777 on Build GRCh38
rs964286952 Chr.8: 129839709-129839709 on Build GRCh38
rs1027424810 Chr.8: 129839635-129839635 on Build GRCh38
rs1031921742 Chr.8: 129839828-129839828 on Build GRCh38
rs1014166263 Chr.8: 129839552-129839552 on Build GRCh38
rs894392097 Chr.8: 129839548-129839548 on Build GRCh38
rs993445172 Chr.8: 129839671-129839671 on Build GRCh38
rs774708993 Chr.8: 129839562-129839562 on Build GRCh38
rs148822232 Chr.8: 129839645-129839645 on Build GRCh38
rs951767397 Chr.8: 129839853-129839853 on Build GRCh38
rs1024353906 Chr.8: 129839561-129839561 on Build GRCh38
rs1042168957 Chr.8: 129839748-129839748 on Build GRCh38
rs1041057308 Chr.8: 129839432-129839432 on Build GRCh38
rs554907665 Chr.8: 129839463-129839463 on Build GRCh38
rs773557965 Chr.8: 129839734-129839734 on Build GRCh38
rs894318432 Chr.8: 129839768-129839768 on Build GRCh38
rs1036914172 Chr.8: 129839738-129839738 on Build GRCh38
rs182772311 Chr.8: 129839771-129839771 on Build GRCh38
rs1007506580 Chr.8: 129839440-129839440 on Build GRCh38
rs574900241 Chr.8: 129839496-129839496 on Build GRCh38
rs533986610 Chr.8: 129839434-129839434 on Build GRCh38
rs1017587594 Chr.8: 129839456-129839456 on Build GRCh38
rs916999359 Chr.8: 129839680-129839680 on Build GRCh38
rs998025855 Chr.8: 129839758-129839758 on Build GRCh38
rs768099193 Chr.8: 129839719-129839719 on Build GRCh38
rs993626222 Chr.8: 129839597-129839597 on Build GRCh38
rs993000401 Chr.8: 129839842-129839842 on Build GRCh38
rs556159074 Chr.8: 129839672-129839672 on Build GRCh38
rs769077460 Chr.8: 129839491-129839491 on Build GRCh38
rs759228022 Chr.8: 129839810-129839810 on Build GRCh38
rs35118626 Chr.8: 129839553-129839553 on Build GRCh38
rs1017656384 Chr.8: 129839803-129839803 on Build GRCh38

View Full Product Details