Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 129855533-129856039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM49B
Entrez Gene ID
51571
Gene Name
family with sequence similarity 49 member B
Gene Aliases
BM-009, L1
UniGene
Hs.126941
Gene Chromosome Location
Chr.8: 129839593-130016697 on Build GRCh38
Transcripts
NM_001256763.1 NM_016623.4 NR_046359.1 NR_046360.1 NR_046361.1 XM_005250966.3 XM_011517108.2 XM_011517109.2 XM_011517110.2 XM_011517111.2 XM_011517112.2 XM_011517113.2 XM_011517114.2 XM_011517115.2 XM_011517116.2 XM_011517117.2 XM_011517118.2 XM_011517120.2 XM_011517121.2 XM_011517122.2 XM_011517123.2 XM_011517124.2 XM_011517125.2 XM_017013539.1 XM_017013540.1 XM_017013541.1 XM_017013542.1 XM_017013543.1 XM_017013544.1 XM_017013545.1 XM_017013546.1 XM_017013547.1 XM_017013548.1 XM_017013549.1 XM_017013550.1 XM_017013551.1 XM_017013552.1 XM_017013553.1 XM_017013554.1 XM_017013555.1 XM_017013556.1 XM_017013557.1 XM_017013558.1 XM_017013559.1 XM_017013560.1 XM_017013561.1 XM_017013562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
454013 c.429T>G p.D143E Chr.8: 129855620-129855620 on Build GRCh38
1674174 c.421C>T p.R141W Chr.8: 129855628-129855628 on Build GRCh38
375137 c.352C>A p.P118T Chr.8: 129855697-129855697 on Build GRCh38
6409572 c.367G>T p.E123* Chr.8: 129855682-129855682 on Build GRCh38
5497930 c.432A>G p.E144E Chr.8: 129855617-129855617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760828993 Chr.8: 129855560-129855560 on Build GRCh38
rs11783021 Chr.8: 129855966-129855966 on Build GRCh38
rs199602343 Chr.8: 129855603-129855603 on Build GRCh38
rs923720910 Chr.8: 129855958-129855958 on Build GRCh38
rs947930483 Chr.8: 129856001-129856001 on Build GRCh38
rs896093257 Chr.8: 129855910-129855910 on Build GRCh38
rs777109088 Chr.8: 129855774-129855774 on Build GRCh38
rs113763286 Chr.8: 129855884-129855884 on Build GRCh38
rs774915607 Chr.8: 129855798-129855801 on Build GRCh38
rs899226263 Chr.8: 129855979-129855979 on Build GRCh38
rs771046659 Chr.8: 129855771-129855771 on Build GRCh38
rs202014391 Chr.8: 129855704-129855704 on Build GRCh38
rs565251834 Chr.8: 129855990-129855990 on Build GRCh38
rs185511127 Chr.8: 129855825-129855825 on Build GRCh38
rs773513361 Chr.8: 129855766-129855766 on Build GRCh38
rs760103915 Chr.8: 129855780-129855780 on Build GRCh38
rs747028505 Chr.8: 129855768-129855768 on Build GRCh38
rs79936991 Chr.8: 129856003-129856003 on Build GRCh38
rs374178710 Chr.8: 129855598-129855598 on Build GRCh38
rs554434179 Chr.8: 129855965-129855965 on Build GRCh38
rs368698797 Chr.8: 129855580-129855580 on Build GRCh38
rs765145827 Chr.8: 129855773-129855777 on Build GRCh38
rs756794671 Chr.8: 129855601-129855601 on Build GRCh38
rs1056032897 Chr.8: 129855980-129855980 on Build GRCh38
rs374255443 Chr.8: 129855608-129855608 on Build GRCh38
rs780111793 Chr.8: 129855677-129855677 on Build GRCh38
rs755685679 Chr.8: 129855627-129855627 on Build GRCh38
rs894815952 Chr.8: 129855985-129855985 on Build GRCh38
rs528680290 Chr.8: 129855904-129855904 on Build GRCh38
rs766831950 Chr.8: 129855564-129855564 on Build GRCh38
rs1004800309 Chr.8: 129855943-129855943 on Build GRCh38
rs772134588 Chr.8: 129855764-129855764 on Build GRCh38
rs781603079 Chr.8: 129855605-129855605 on Build GRCh38
rs561575649 Chr.8: 129855893-129855893 on Build GRCh38
rs367790590 Chr.8: 129855824-129855824 on Build GRCh38
rs746375420 Chr.8: 129855796-129855796 on Build GRCh38
rs1043220635 Chr.8: 129855790-129855790 on Build GRCh38
rs754811169 Chr.8: 129855721-129855721 on Build GRCh38
rs765739159 Chr.8: 129855781-129855781 on Build GRCh38
rs556972913 Chr.8: 129855736-129855736 on Build GRCh38
rs2306528 Chr.8: 129855815-129855815 on Build GRCh38
rs778781080 Chr.8: 129855733-129855733 on Build GRCh38
rs540015860 Chr.8: 129855978-129855978 on Build GRCh38
rs754816185 Chr.8: 129855753-129855754 on Build GRCh38
rs200833467 Chr.8: 129855581-129855581 on Build GRCh38
rs771739361 Chr.8: 129856012-129856012 on Build GRCh38

View Full Product Details