Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 140511863-140512355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRAC1
Entrez Gene ID
54108
Gene Name
chromatin accessibility complex 1
Gene Aliases
CHARC1, CHARC15, CHRAC-1, CHRAC-15, CHRAC15, YCL1
UniGene
Hs.279704
Gene Chromosome Location
Chr.8: 140511298-140517153 on Build GRCh38
Transcripts
NM_017444.5 NR_023360.2 NR_040712.1

SNP Detail

SNP ID Chromosome Location
rs951133811 Chr.8: 140512076-140512076 on Build GRCh38
rs115258008 Chr.8: 140512317-140512317 on Build GRCh38
rs183879873 Chr.8: 140512087-140512087 on Build GRCh38
rs527893343 Chr.8: 140512113-140512113 on Build GRCh38
rs762891262 Chr.8: 140511893-140511893 on Build GRCh38
rs967165956 Chr.8: 140512201-140512201 on Build GRCh38
rs1025144451 Chr.8: 140511941-140511941 on Build GRCh38
rs548133706 Chr.8: 140511990-140511990 on Build GRCh38
rs1011298137 Chr.8: 140511939-140511939 on Build GRCh38
rs894076411 Chr.8: 140512189-140512189 on Build GRCh38
rs368202310 Chr.8: 140512302-140512302 on Build GRCh38
rs13439063 Chr.8: 140512181-140512181 on Build GRCh38
rs986033708 Chr.8: 140512282-140512282 on Build GRCh38
rs1050055871 Chr.8: 140511904-140511904 on Build GRCh38
rs980980730 Chr.8: 140512116-140512116 on Build GRCh38
rs979534497 Chr.8: 140512326-140512326 on Build GRCh38
rs764109292 Chr.8: 140511901-140511901 on Build GRCh38
rs529773033 Chr.8: 140511907-140511907 on Build GRCh38
rs909551876 Chr.8: 140511917-140511917 on Build GRCh38
rs552494245 Chr.8: 140512080-140512080 on Build GRCh38
rs757354333 Chr.8: 140511945-140511945 on Build GRCh38
rs966839808 Chr.8: 140512019-140512019 on Build GRCh38
rs1039630536 Chr.8: 140512197-140512200 on Build GRCh38
rs777736346 Chr.8: 140512035-140512035 on Build GRCh38
rs897586091 Chr.8: 140511926-140511926 on Build GRCh38
rs546501804 Chr.8: 140512118-140512118 on Build GRCh38
rs992264427 Chr.8: 140512141-140512141 on Build GRCh38
rs1020958723 Chr.8: 140512193-140512193 on Build GRCh38
rs772130804 Chr.8: 140512007-140512007 on Build GRCh38
rs945900965 Chr.8: 140511888-140511888 on Build GRCh38
rs557786873 Chr.8: 140512051-140512051 on Build GRCh38
rs998315431 Chr.8: 140512242-140512242 on Build GRCh38
rs777050908 Chr.8: 140512061-140512061 on Build GRCh38
rs910202261 Chr.8: 140512069-140512069 on Build GRCh38
rs899726195 Chr.8: 140512205-140512205 on Build GRCh38
rs906748155 Chr.8: 140512138-140512138 on Build GRCh38
rs779719980 Chr.8: 140511962-140511962 on Build GRCh38
rs1037880215 Chr.8: 140511913-140511913 on Build GRCh38
rs189000781 Chr.8: 140512246-140512246 on Build GRCh38
rs994047625 Chr.8: 140511927-140511927 on Build GRCh38
rs925388743 Chr.8: 140512328-140512328 on Build GRCh38
rs1029518947 Chr.8: 140512074-140512074 on Build GRCh38
rs1033636981 Chr.8: 140511963-140511963 on Build GRCh38
rs998041837 Chr.8: 140512072-140512072 on Build GRCh38
rs954603152 Chr.8: 140512056-140512056 on Build GRCh38
rs766952343 Chr.8: 140511920-140511920 on Build GRCh38
rs933229620 Chr.8: 140511896-140511896 on Build GRCh38
rs981431407 Chr.8: 140511956-140511956 on Build GRCh38
rs747154578 Chr.8: 140512050-140512050 on Build GRCh38
rs926904234 Chr.8: 140512129-140512129 on Build GRCh38
rs998115465 Chr.8: 140512208-140512208 on Build GRCh38
rs929043070 Chr.8: 140512309-140512309 on Build GRCh38
rs535564579 Chr.8: 140512148-140512148 on Build GRCh38
rs563120782 Chr.8: 140512036-140512036 on Build GRCh38
rs1000079419 Chr.8: 140512063-140512063 on Build GRCh38
rs540792362 Chr.8: 140512307-140512307 on Build GRCh38
rs1021430443 Chr.8: 140512010-140512010 on Build GRCh38
rs13439065 Chr.8: 140512287-140512287 on Build GRCh38
rs115878356 Chr.8: 140512133-140512133 on Build GRCh38
rs779469354 Chr.8: 140511988-140511988 on Build GRCh38
rs192824182 Chr.8: 140512332-140512332 on Build GRCh38
rs945473561 Chr.8: 140511991-140511991 on Build GRCh38
rs1029466728 Chr.8: 140512212-140512212 on Build GRCh38
rs1003961212 Chr.8: 140512277-140512277 on Build GRCh38
rs992380378 Chr.8: 140512314-140512314 on Build GRCh38
rs76790724 Chr.8: 140512065-140512065 on Build GRCh38
rs939554682 Chr.8: 140511967-140511967 on Build GRCh38
rs761324390 Chr.8: 140511909-140511909 on Build GRCh38
rs1044202136 Chr.8: 140512166-140512166 on Build GRCh38
rs569465037 Chr.8: 140512168-140512168 on Build GRCh38
rs559353647 Chr.8: 140511886-140511886 on Build GRCh38
rs954551660 Chr.8: 140512257-140512257 on Build GRCh38
rs754175710 Chr.8: 140511966-140511966 on Build GRCh38
rs914087906 Chr.8: 140512144-140512144 on Build GRCh38
rs144047429 Chr.8: 140512296-140512296 on Build GRCh38
rs958053450 Chr.8: 140511979-140511979 on Build GRCh38
rs923162182 Chr.8: 140511894-140511894 on Build GRCh38
rs747646847 Chr.8: 140512119-140512119 on Build GRCh38
rs755791633 Chr.8: 140511958-140511958 on Build GRCh38
rs918963640 Chr.8: 140512300-140512300 on Build GRCh38
rs1011303086 Chr.8: 140512191-140512191 on Build GRCh38
rs750656764 Chr.8: 140512275-140512275 on Build GRCh38
rs1002390423 Chr.8: 140512171-140512171 on Build GRCh38
rs1043705248 Chr.8: 140512039-140512039 on Build GRCh38
rs138537283 Chr.8: 140512291-140512291 on Build GRCh38
rs914034525 Chr.8: 140512316-140512316 on Build GRCh38
rs867133840 Chr.8: 140511969-140511969 on Build GRCh38
rs945448690 Chr.8: 140512160-140512160 on Build GRCh38
rs772464244 Chr.8: 140512031-140512032 on Build GRCh38
rs750046415 Chr.8: 140511955-140511955 on Build GRCh38
rs929078027 Chr.8: 140512124-140512124 on Build GRCh38
rs941662010 Chr.8: 140512075-140512075 on Build GRCh38
rs928122041 Chr.8: 140512330-140512330 on Build GRCh38
rs755389607 Chr.8: 140511977-140511977 on Build GRCh38

View Full Product Details