Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14735656-14736169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
UniGene
Hs.50850
Gene Chromosome Location
Chr.9: 14734666-14911653 on Build GRCh38
Transcripts
NM_001177704.1 NM_144966.5 XM_005251382.3 XM_005251384.4 XM_006716729.3 XM_011517758.2 XM_017014316.1 XM_017014317.1 XM_017014318.1 XM_017014319.1 XM_017014320.1 XM_017014321.1 XM_017014322.1 XM_017014323.1 XM_017014324.1 XM_017014325.1 XM_017014326.1 XM_017014327.1

SNP Detail

SNP ID Chromosome Location
rs560326767 Chr.9: 14735790-14735790 on Build GRCh38
rs993645131 Chr.9: 14736125-14736125 on Build GRCh38
rs1009490857 Chr.9: 14735746-14735746 on Build GRCh38
rs1054346763 Chr.9: 14735775-14735775 on Build GRCh38
rs1039731712 Chr.9: 14735903-14735903 on Build GRCh38
rs937261905 Chr.9: 14735767-14735767 on Build GRCh38
rs981158603 Chr.9: 14735739-14735739 on Build GRCh38
rs886063756 Chr.9: 14736002-14736002 on Build GRCh38
rs926961249 Chr.9: 14736048-14736048 on Build GRCh38
rs926252497 Chr.9: 14735935-14735935 on Build GRCh38
rs752951576 Chr.9: 14735943-14735943 on Build GRCh38
rs962263007 Chr.9: 14736111-14736111 on Build GRCh38
rs939121683 Chr.9: 14735940-14735940 on Build GRCh38
rs369753007 Chr.9: 14735866-14735866 on Build GRCh38
rs987933485 Chr.9: 14735863-14735863 on Build GRCh38
rs11789504 Chr.9: 14736140-14736140 on Build GRCh38
rs953508841 Chr.9: 14735845-14735845 on Build GRCh38
rs752777576 Chr.9: 14735869-14735869 on Build GRCh38
rs150510806 Chr.9: 14736020-14736020 on Build GRCh38
rs1004416560 Chr.9: 14736014-14736014 on Build GRCh38
rs892339060 Chr.9: 14735953-14735953 on Build GRCh38
rs537080439 Chr.9: 14735794-14735794 on Build GRCh38
rs1056578361 Chr.9: 14735951-14735951 on Build GRCh38
rs995722266 Chr.9: 14735923-14735923 on Build GRCh38
rs907654735 Chr.9: 14735716-14735716 on Build GRCh38
rs113410147 Chr.9: 14735928-14735931 on Build GRCh38
rs892259614 Chr.9: 14735729-14735729 on Build GRCh38
rs886063755 Chr.9: 14735838-14735838 on Build GRCh38
rs149242359 Chr.9: 14736097-14736097 on Build GRCh38
rs528248069 Chr.9: 14735730-14735730 on Build GRCh38
rs1027801058 Chr.9: 14736138-14736138 on Build GRCh38
rs958328283 Chr.9: 14736092-14736092 on Build GRCh38
rs751524710 Chr.9: 14735727-14735727 on Build GRCh38
rs549260371 Chr.9: 14735692-14735692 on Build GRCh38
rs972340747 Chr.9: 14735889-14735889 on Build GRCh38
rs116356673 Chr.9: 14736094-14736094 on Build GRCh38
rs1041939526 Chr.9: 14735838-14735838 on Build GRCh38
rs143690507 Chr.9: 14735760-14735760 on Build GRCh38
rs1044862953 Chr.9: 14735684-14735684 on Build GRCh38
rs78207703 Chr.9: 14735931-14735931 on Build GRCh38
rs996697825 Chr.9: 14736055-14736055 on Build GRCh38
rs981577026 Chr.9: 14736034-14736034 on Build GRCh38
rs899815446 Chr.9: 14735904-14735904 on Build GRCh38
rs1050400478 Chr.9: 14736077-14736077 on Build GRCh38
rs1494343 Chr.9: 14735696-14735696 on Build GRCh38
rs971192593 Chr.9: 14736023-14736023 on Build GRCh38
rs962517927 Chr.9: 14735883-14735883 on Build GRCh38
rs555610646 Chr.9: 14735892-14735892 on Build GRCh38
rs534888810 Chr.9: 14735976-14735976 on Build GRCh38
rs1056312185 Chr.9: 14735728-14735728 on Build GRCh38
rs566879689 Chr.9: 14735844-14735844 on Build GRCh38
rs914532518 Chr.9: 14736096-14736096 on Build GRCh38
rs886063757 Chr.9: 14736138-14736138 on Build GRCh38
rs754838735 Chr.9: 14735759-14735759 on Build GRCh38
rs35992592 Chr.9: 14736007-14736008 on Build GRCh38
rs1041581778 Chr.9: 14736132-14736132 on Build GRCh38
rs903764849 Chr.9: 14736031-14736031 on Build GRCh38
rs535746918 Chr.9: 14735857-14735857 on Build GRCh38
rs550149566 Chr.9: 14735816-14735816 on Build GRCh38

View Full Product Details