Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14746781-14747277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
UniGene
Hs.50850
Gene Chromosome Location
Chr.9: 14734666-14911653 on Build GRCh38
Transcripts
NM_001177704.1 NM_144966.5 XM_005251382.3 XM_005251384.4 XM_006716729.3 XM_011517758.2 XM_017014316.1 XM_017014317.1 XM_017014318.1 XM_017014319.1 XM_017014320.1 XM_017014321.1 XM_017014322.1 XM_017014323.1 XM_017014324.1 XM_017014325.1 XM_017014326.1 XM_017014327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5410857 c.6010G>A p.D2004N Chr.9: 14747051-14747051 on Build GRCh38
3656191 c.6105G>A p.G2035G Chr.9: 14746956-14746956 on Build GRCh38
5968093 c.1662G>T p.K554N Chr.9: 14747007-14747007 on Build GRCh38
6429159 c.1733G>T p.S578I Chr.9: 14746936-14746936 on Build GRCh38
7268582 c.1726G>A p.A576T Chr.9: 14746943-14746943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762234673 Chr.9: 14747100-14747100 on Build GRCh38
rs771752655 Chr.9: 14746905-14746905 on Build GRCh38
rs780095104 Chr.9: 14746886-14746886 on Build GRCh38
rs771531342 Chr.9: 14746957-14746957 on Build GRCh38
rs751665114 Chr.9: 14746874-14746874 on Build GRCh38
rs774868755 Chr.9: 14747096-14747096 on Build GRCh38
rs754058350 Chr.9: 14747233-14747233 on Build GRCh38
rs550955683 Chr.9: 14746859-14746859 on Build GRCh38
rs75125669 Chr.9: 14746993-14746993 on Build GRCh38
rs777270214 Chr.9: 14747075-14747075 on Build GRCh38
rs373182919 Chr.9: 14746918-14746918 on Build GRCh38
rs2270530 Chr.9: 14747159-14747159 on Build GRCh38
rs748058742 Chr.9: 14747064-14747064 on Build GRCh38
rs572603801 Chr.9: 14747209-14747209 on Build GRCh38
rs17290109 Chr.9: 14746848-14746848 on Build GRCh38
rs558194682 Chr.9: 14747073-14747073 on Build GRCh38
rs761290172 Chr.9: 14746902-14746902 on Build GRCh38
rs947280592 Chr.9: 14746822-14746822 on Build GRCh38
rs934520223 Chr.9: 14747033-14747033 on Build GRCh38
rs1007530556 Chr.9: 14746830-14746830 on Build GRCh38
rs757513458 Chr.9: 14746877-14746877 on Build GRCh38
rs764655202 Chr.9: 14746992-14746992 on Build GRCh38
rs763196871 Chr.9: 14747234-14747237 on Build GRCh38
rs778888575 Chr.9: 14747062-14747062 on Build GRCh38
rs756420258 Chr.9: 14747031-14747031 on Build GRCh38
rs908910462 Chr.9: 14746867-14746867 on Build GRCh38
rs775932714 Chr.9: 14746969-14746969 on Build GRCh38
rs1041550995 Chr.9: 14746872-14746872 on Build GRCh38
rs990899832 Chr.9: 14747076-14747076 on Build GRCh38
rs373394661 Chr.9: 14746973-14746973 on Build GRCh38
rs1053579583 Chr.9: 14747170-14747170 on Build GRCh38
rs767740329 Chr.9: 14747021-14747021 on Build GRCh38
rs2270529 Chr.9: 14747135-14747135 on Build GRCh38
rs919419142 Chr.9: 14746852-14746852 on Build GRCh38
rs1016487209 Chr.9: 14747067-14747067 on Build GRCh38
rs768628308 Chr.9: 14746953-14746953 on Build GRCh38
rs979128726 Chr.9: 14746975-14746975 on Build GRCh38
rs750720207 Chr.9: 14746944-14746944 on Build GRCh38
rs886063765 Chr.9: 14746975-14746975 on Build GRCh38
rs770077384 Chr.9: 14746965-14746965 on Build GRCh38
rs917583009 Chr.9: 14747082-14747082 on Build GRCh38
rs924536193 Chr.9: 14747002-14747002 on Build GRCh38
rs888482142 Chr.9: 14747128-14747128 on Build GRCh38
rs749631410 Chr.9: 14747091-14747091 on Build GRCh38
rs371517454 Chr.9: 14746958-14746958 on Build GRCh38
rs961883154 Chr.9: 14746919-14746919 on Build GRCh38
rs145607985 Chr.9: 14747136-14747136 on Build GRCh38
rs375371745 Chr.9: 14747216-14747216 on Build GRCh38
rs781440682 Chr.9: 14746943-14746943 on Build GRCh38
rs542384549 Chr.9: 14746827-14746827 on Build GRCh38
rs757721445 Chr.9: 14747188-14747188 on Build GRCh38
rs373482754 Chr.9: 14746908-14746908 on Build GRCh38
rs1005547572 Chr.9: 14747132-14747132 on Build GRCh38
rs775735324 Chr.9: 14746969-14746970 on Build GRCh38
rs774758614 Chr.9: 14747015-14747015 on Build GRCh38
rs528114123 Chr.9: 14746858-14746858 on Build GRCh38
rs200987019 Chr.9: 14747040-14747040 on Build GRCh38
rs747665116 Chr.9: 14746954-14746954 on Build GRCh38
rs377085995 Chr.9: 14747028-14747028 on Build GRCh38
rs776329304 Chr.9: 14747090-14747090 on Build GRCh38
rs544661735 Chr.9: 14747219-14747219 on Build GRCh38
rs889711285 Chr.9: 14747174-14747174 on Build GRCh38
rs374903635 Chr.9: 14746909-14746909 on Build GRCh38
rs769747031 Chr.9: 14746882-14746882 on Build GRCh38
rs752880221 Chr.9: 14746929-14746929 on Build GRCh38
rs191713464 Chr.9: 14747054-14747054 on Build GRCh38
rs73644845 Chr.9: 14747180-14747180 on Build GRCh38
rs201717559 Chr.9: 14746934-14746934 on Build GRCh38
rs746694579 Chr.9: 14746964-14746964 on Build GRCh38
rs755936692 Chr.9: 14746948-14746948 on Build GRCh38
rs564553123 Chr.9: 14747234-14747234 on Build GRCh38
rs566175340 Chr.9: 14747052-14747052 on Build GRCh38
rs369645771 Chr.9: 14746910-14746910 on Build GRCh38
rs766678522 Chr.9: 14747230-14747230 on Build GRCh38
rs774185315 Chr.9: 14746820-14746820 on Build GRCh38
rs559569540 Chr.9: 14746887-14746887 on Build GRCh38
rs932684585 Chr.9: 14747104-14747104 on Build GRCh38
rs746604828 Chr.9: 14747078-14747078 on Build GRCh38
rs959751928 Chr.9: 14746941-14746941 on Build GRCh38
rs1026367640 Chr.9: 14747238-14747238 on Build GRCh38
rs199776467 Chr.9: 14746879-14746879 on Build GRCh38
rs1049766490 Chr.9: 14747106-14747106 on Build GRCh38
rs578046556 Chr.9: 14747086-14747086 on Build GRCh38
rs200035180 Chr.9: 14747235-14747235 on Build GRCh38
rs949196536 Chr.9: 14747087-14747087 on Build GRCh38
rs763283407 Chr.9: 14746980-14746980 on Build GRCh38
rs757345586 Chr.9: 14746942-14746942 on Build GRCh38
rs761927519 Chr.9: 14747017-14747017 on Build GRCh38
rs752612632 Chr.9: 14747245-14747245 on Build GRCh38
rs758344694 Chr.9: 14747253-14747253 on Build GRCh38
rs542797406 Chr.9: 14747165-14747165 on Build GRCh38
rs201502030 Chr.9: 14746952-14746952 on Build GRCh38
rs1039486407 Chr.9: 14747203-14747203 on Build GRCh38
rs536842666 Chr.9: 14746927-14746927 on Build GRCh38
rs1018124604 Chr.9: 14746835-14746835 on Build GRCh38
rs1004132383 Chr.9: 14747138-14747138 on Build GRCh38
rs1035874003 Chr.9: 14746937-14746937 on Build GRCh38

View Full Product Details