Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14747542-14748040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
UniGene
Hs.50850
Gene Chromosome Location
Chr.9: 14734666-14911653 on Build GRCh38
Transcripts
NM_001177704.1 NM_144966.5 XM_005251382.3 XM_005251384.4 XM_006716729.3 XM_011517758.2 XM_017014316.1 XM_017014317.1 XM_017014318.1 XM_017014319.1 XM_017014320.1 XM_017014321.1 XM_017014322.1 XM_017014323.1 XM_017014324.1 XM_017014325.1 XM_017014326.1 XM_017014327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5595808 c.657C>T p.I219I Chr.9: 14747718-14747718 on Build GRCh38
5595807 c.5807C>T p.S1936F Chr.9: 14747718-14747718 on Build GRCh38
6516966 c.1440C>T p.D480D Chr.9: 14747693-14747693 on Build GRCh38
3952581 c.5808C>G p.S1936S Chr.9: 14747717-14747717 on Build GRCh38
6516967 c.5832C>T p.D1944D Chr.9: 14747693-14747693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548335559 Chr.9: 14747709-14747709 on Build GRCh38
rs565918778 Chr.9: 14747895-14747895 on Build GRCh38
rs774136112 Chr.9: 14747962-14747962 on Build GRCh38
rs970930849 Chr.9: 14747649-14747649 on Build GRCh38
rs369305667 Chr.9: 14747636-14747636 on Build GRCh38
rs922508668 Chr.9: 14747818-14747818 on Build GRCh38
rs534542048 Chr.9: 14748000-14748001 on Build GRCh38
rs757680975 Chr.9: 14747868-14747868 on Build GRCh38
rs551878540 Chr.9: 14747624-14747624 on Build GRCh38
rs976965743 Chr.9: 14747796-14747796 on Build GRCh38
rs765923904 Chr.9: 14748002-14748003 on Build GRCh38
rs999778961 Chr.9: 14747897-14747897 on Build GRCh38
rs112071417 Chr.9: 14747735-14747735 on Build GRCh38
rs1037441758 Chr.9: 14747888-14747888 on Build GRCh38
rs918560218 Chr.9: 14747909-14747909 on Build GRCh38
rs757689559 Chr.9: 14747765-14747765 on Build GRCh38
rs112910368 Chr.9: 14747871-14747871 on Build GRCh38
rs201497795 Chr.9: 14747725-14747725 on Build GRCh38
rs369775410 Chr.9: 14747861-14747861 on Build GRCh38
rs999916017 Chr.9: 14747997-14747997 on Build GRCh38
rs539710620 Chr.9: 14747952-14747952 on Build GRCh38
rs192549106 Chr.9: 14747826-14747826 on Build GRCh38
rs140829838 Chr.9: 14747651-14747651 on Build GRCh38
rs753567691 Chr.9: 14747694-14747694 on Build GRCh38
rs895389737 Chr.9: 14747934-14747934 on Build GRCh38
rs1030697701 Chr.9: 14747906-14747906 on Build GRCh38
rs1038313913 Chr.9: 14747598-14747598 on Build GRCh38
rs751699199 Chr.9: 14747864-14747865 on Build GRCh38
rs188153650 Chr.9: 14747730-14747730 on Build GRCh38
rs933338991 Chr.9: 14747575-14747575 on Build GRCh38
rs1022413310 Chr.9: 14747623-14747623 on Build GRCh38
rs143991406 Chr.9: 14748001-14748002 on Build GRCh38
rs922523794 Chr.9: 14747814-14747814 on Build GRCh38
rs529022197 Chr.9: 14747860-14747860 on Build GRCh38
rs778288375 Chr.9: 14747724-14747724 on Build GRCh38
rs929859569 Chr.9: 14747820-14747820 on Build GRCh38
rs895358724 Chr.9: 14747632-14747632 on Build GRCh38
rs1031831492 Chr.9: 14748002-14748002 on Build GRCh38
rs891498233 Chr.9: 14748014-14748014 on Build GRCh38
rs749899966 Chr.9: 14747639-14747639 on Build GRCh38
rs1047679463 Chr.9: 14747576-14747576 on Build GRCh38
rs114909107 Chr.9: 14747711-14747711 on Build GRCh38
rs939347117 Chr.9: 14747916-14747916 on Build GRCh38
rs779551648 Chr.9: 14747685-14747685 on Build GRCh38
rs879022216 Chr.9: 14747799-14747799 on Build GRCh38
rs1022699507 Chr.9: 14747647-14747647 on Build GRCh38
rs557802851 Chr.9: 14748015-14748015 on Build GRCh38
rs866115618 Chr.9: 14747707-14747707 on Build GRCh38
rs745636845 Chr.9: 14747965-14747965 on Build GRCh38
rs115715965 Chr.9: 14747831-14747831 on Build GRCh38
rs535140027 Chr.9: 14747594-14747594 on Build GRCh38
rs369579969 Chr.9: 14747767-14747767 on Build GRCh38
rs1056666714 Chr.9: 14747924-14747924 on Build GRCh38
rs957815287 Chr.9: 14747687-14747689 on Build GRCh38
rs953881670 Chr.9: 14747828-14747828 on Build GRCh38
rs1025244302 Chr.9: 14747643-14747643 on Build GRCh38
rs963922650 Chr.9: 14747787-14747787 on Build GRCh38
rs75798907 Chr.9: 14747986-14747986 on Build GRCh38
rs564624823 Chr.9: 14747985-14747985 on Build GRCh38
rs886532015 Chr.9: 14747581-14747581 on Build GRCh38
rs1037777139 Chr.9: 14747836-14747836 on Build GRCh38

View Full Product Details