Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 17235518-17236047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTLN
Entrez Gene ID
54875
Gene Name
centlein
Gene Aliases
C9orf101, C9orf39, bA340N12.1
UniGene
Hs.435381
Gene Chromosome Location
Chr.9: 17134991-17510019 on Build GRCh38
Transcripts
NM_001114395.2 NM_017738.3 XM_005251492.1 XM_006716793.3 XM_011517941.2 XM_017014839.1 XM_017014840.1 XM_017014841.1 XM_017014842.1 XM_017014843.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3433041 c.661G>T p.E221* Chr.9: 17235784-17235784 on Build GRCh38
5201451 c.648A>T p.K216N Chr.9: 17235771-17235771 on Build GRCh38
7282727 c.669+6T>C p.? Chr.9: 17235798-17235798 on Build GRCh38
5826499 c.661G>T p.E221* Chr.9: 17235784-17235784 on Build GRCh38
3906384 c.577C>T p.R193W Chr.9: 17235700-17235700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753299970 Chr.9: 17235818-17235818 on Build GRCh38
rs778558860 Chr.9: 17235716-17235716 on Build GRCh38
rs564149063 Chr.9: 17235669-17235669 on Build GRCh38
rs79772556 Chr.9: 17235645-17235645 on Build GRCh38
rs201402782 Chr.9: 17235757-17235757 on Build GRCh38
rs771206364 Chr.9: 17235730-17235730 on Build GRCh38
rs368343858 Chr.9: 17235666-17235666 on Build GRCh38
rs1050798750 Chr.9: 17235858-17235858 on Build GRCh38
rs748912376 Chr.9: 17235705-17235705 on Build GRCh38
rs753175528 Chr.9: 17235656-17235657 on Build GRCh38
rs376504700 Chr.9: 17235808-17235808 on Build GRCh38
rs1051937082 Chr.9: 17235622-17235622 on Build GRCh38
rs767419795 Chr.9: 17235691-17235691 on Build GRCh38
rs773610353 Chr.9: 17235607-17235607 on Build GRCh38
rs890367718 Chr.9: 17235889-17235889 on Build GRCh38
rs980280803 Chr.9: 17235994-17235994 on Build GRCh38
rs752307741 Chr.9: 17235830-17235830 on Build GRCh38
rs563283677 Chr.9: 17235590-17235590 on Build GRCh38
rs550576537 Chr.9: 17235820-17235820 on Build GRCh38
rs777035862 Chr.9: 17235623-17235623 on Build GRCh38
rs67017076 Chr.9: 17235645-17235645 on Build GRCh38
rs200513513 Chr.9: 17235772-17235772 on Build GRCh38
rs3814137 Chr.9: 17235565-17235565 on Build GRCh38
rs763146549 Chr.9: 17235753-17235753 on Build GRCh38
rs768956416 Chr.9: 17235762-17235762 on Build GRCh38
rs1028881724 Chr.9: 17235583-17235583 on Build GRCh38
rs751346501 Chr.9: 17235672-17235672 on Build GRCh38
rs1008681140 Chr.9: 17235921-17235921 on Build GRCh38
rs765246731 Chr.9: 17235636-17235636 on Build GRCh38
rs953014142 Chr.9: 17235986-17235986 on Build GRCh38
rs765857631 Chr.9: 17235815-17235815 on Build GRCh38
rs200971583 Chr.9: 17235642-17235642 on Build GRCh38
rs936913966 Chr.9: 17235543-17235543 on Build GRCh38
rs995210630 Chr.9: 17235744-17235744 on Build GRCh38
rs767796555 Chr.9: 17235804-17235804 on Build GRCh38
rs371692098 Chr.9: 17235797-17235797 on Build GRCh38
rs961324385 Chr.9: 17235947-17235950 on Build GRCh38
rs528248591 Chr.9: 17235700-17235700 on Build GRCh38
rs752105062 Chr.9: 17235869-17235869 on Build GRCh38
rs775761421 Chr.9: 17235748-17235748 on Build GRCh38
rs747496020 Chr.9: 17235618-17235618 on Build GRCh38
rs981282123 Chr.9: 17235892-17235892 on Build GRCh38
rs750338969 Chr.9: 17235694-17235694 on Build GRCh38
rs554157525 Chr.9: 17235823-17235823 on Build GRCh38
rs761608773 Chr.9: 17235789-17235789 on Build GRCh38
rs1032449985 Chr.9: 17235962-17235962 on Build GRCh38
rs994224132 Chr.9: 17235948-17235948 on Build GRCh38
rs781682940 Chr.9: 17235732-17235732 on Build GRCh38
rs753049767 Chr.9: 17235839-17235839 on Build GRCh38
rs201832695 Chr.9: 17235642-17235642 on Build GRCh38
rs1019572158 Chr.9: 17235923-17235923 on Build GRCh38
rs767271088 Chr.9: 17235790-17235790 on Build GRCh38
rs397708736 Chr.9: 17235651-17235651 on Build GRCh38
rs914191665 Chr.9: 17235572-17235572 on Build GRCh38
rs763307145 Chr.9: 17235661-17235661 on Build GRCh38
rs898138601 Chr.9: 17235742-17235742 on Build GRCh38
rs750373402 Chr.9: 17235795-17235795 on Build GRCh38
rs778410612 Chr.9: 17236024-17236024 on Build GRCh38
rs769961579 Chr.9: 17235739-17235739 on Build GRCh38
rs756584500 Chr.9: 17235973-17235973 on Build GRCh38
rs771421394 Chr.9: 17235621-17235621 on Build GRCh38
rs146167182 Chr.9: 17235712-17235712 on Build GRCh38
rs759595975 Chr.9: 17235628-17235628 on Build GRCh38
rs202134714 Chr.9: 17235727-17235727 on Build GRCh38
rs757029709 Chr.9: 17235689-17235689 on Build GRCh38
rs200386421 Chr.9: 17235638-17235638 on Build GRCh38
rs374609860 Chr.9: 17235755-17235755 on Build GRCh38

View Full Product Details