Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19054926-19055469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS6
Entrez Gene ID
54801
Gene Name
HAUS augmin like complex subunit 6
Gene Aliases
Dgt6, FAM29A
UniGene
Hs.533468
Gene Chromosome Location
Chr.9: 19053137-19102942 on Build GRCh38
Transcripts
NM_001270890.1 NM_017645.4 XM_011517935.2 XM_017014830.1

SNP Detail

SNP ID Chromosome Location
rs988152208 Chr.9: 19055090-19055090 on Build GRCh38
rs181846510 Chr.9: 19055386-19055386 on Build GRCh38
rs1029640635 Chr.9: 19055001-19055001 on Build GRCh38
rs981234460 Chr.9: 19055428-19055428 on Build GRCh38
rs540283189 Chr.9: 19055112-19055112 on Build GRCh38
rs1032466095 Chr.9: 19055063-19055063 on Build GRCh38
rs997670999 Chr.9: 19055252-19055252 on Build GRCh38
rs1015106944 Chr.9: 19054976-19054976 on Build GRCh38
rs575025108 Chr.9: 19055165-19055165 on Build GRCh38
rs184940855 Chr.9: 19055071-19055071 on Build GRCh38
rs949864044 Chr.9: 19055118-19055118 on Build GRCh38
rs766682528 Chr.9: 19055097-19055097 on Build GRCh38
rs113119868 Chr.9: 19055142-19055142 on Build GRCh38
rs902035371 Chr.9: 19055249-19055249 on Build GRCh38
rs1029280743 Chr.9: 19055267-19055267 on Build GRCh38
rs531199539 Chr.9: 19055342-19055342 on Build GRCh38
rs762997858 Chr.9: 19055095-19055095 on Build GRCh38
rs765162659 Chr.9: 19054974-19054974 on Build GRCh38
rs763737307 Chr.9: 19055137-19055137 on Build GRCh38
rs901981166 Chr.9: 19054995-19054995 on Build GRCh38
rs971234856 Chr.9: 19055424-19055424 on Build GRCh38
rs575176681 Chr.9: 19055031-19055031 on Build GRCh38
rs139842699 Chr.9: 19055002-19055003 on Build GRCh38
rs189785405 Chr.9: 19055375-19055375 on Build GRCh38
rs35849076 Chr.9: 19055323-19055324 on Build GRCh38
rs564007160 Chr.9: 19055314-19055314 on Build GRCh38
rs927191888 Chr.9: 19055136-19055137 on Build GRCh38
rs773333855 Chr.9: 19055004-19055004 on Build GRCh38
rs887415444 Chr.9: 19055226-19055226 on Build GRCh38
rs979832450 Chr.9: 19055046-19055046 on Build GRCh38
rs1049203977 Chr.9: 19055143-19055143 on Build GRCh38
rs34022991 Chr.9: 19055359-19055360 on Build GRCh38
rs889362828 Chr.9: 19055296-19055296 on Build GRCh38
rs762810808 Chr.9: 19055078-19055078 on Build GRCh38
rs940432323 Chr.9: 19055238-19055238 on Build GRCh38
rs35635494 Chr.9: 19055326-19055326 on Build GRCh38
rs542421865 Chr.9: 19055201-19055201 on Build GRCh38
rs572855859 Chr.9: 19055092-19055092 on Build GRCh38
rs749950261 Chr.9: 19054970-19054970 on Build GRCh38
rs1032765893 Chr.9: 19055371-19055371 on Build GRCh38
rs981350371 Chr.9: 19055126-19055126 on Build GRCh38
rs997783320 Chr.9: 19054998-19054998 on Build GRCh38
rs149388889 Chr.9: 19055068-19055068 on Build GRCh38
rs576425049 Chr.9: 19055423-19055423 on Build GRCh38
rs7020235 Chr.9: 19054957-19054957 on Build GRCh38
rs1009625862 Chr.9: 19055392-19055392 on Build GRCh38

View Full Product Details