Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19056146-19056599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS6
Entrez Gene ID
54801
Gene Name
HAUS augmin like complex subunit 6
Gene Aliases
Dgt6, FAM29A
UniGene
Hs.533468
Gene Chromosome Location
Chr.9: 19053137-19102942 on Build GRCh38
Transcripts
NM_001270890.1 NM_017645.4 XM_011517935.2 XM_017014830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5518884 c.2811_2813delAGA p.E937delE Chr.9: 19056398-19056400 on Build GRCh38
6570708 c.2837C>G p.A946G Chr.9: 19056374-19056374 on Build GRCh38
3699297 c.2850G>A p.P950P Chr.9: 19056361-19056361 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753529067 Chr.9: 19056361-19056361 on Build GRCh38
rs749927397 Chr.9: 19056453-19056456 on Build GRCh38
rs1033749088 Chr.9: 19056185-19056185 on Build GRCh38
rs772720948 Chr.9: 19056344-19056344 on Build GRCh38
rs1055652236 Chr.9: 19056498-19056499 on Build GRCh38
rs778717205 Chr.9: 19056453-19056453 on Build GRCh38
rs144178724 Chr.9: 19056210-19056210 on Build GRCh38
rs766966694 Chr.9: 19056338-19056338 on Build GRCh38
rs746096313 Chr.9: 19056301-19056301 on Build GRCh38
rs745890823 Chr.9: 19056382-19056382 on Build GRCh38
rs749869179 Chr.9: 19056341-19056341 on Build GRCh38
rs1006044908 Chr.9: 19056217-19056217 on Build GRCh38
rs764525945 Chr.9: 19056362-19056362 on Build GRCh38
rs546514405 Chr.9: 19056297-19056297 on Build GRCh38
rs938011690 Chr.9: 19056488-19056488 on Build GRCh38
rs528514229 Chr.9: 19056350-19056350 on Build GRCh38
rs776460106 Chr.9: 19056425-19056425 on Build GRCh38
rs957816589 Chr.9: 19056569-19056569 on Build GRCh38
rs755821951 Chr.9: 19056398-19056400 on Build GRCh38
rs756237604 Chr.9: 19056439-19056439 on Build GRCh38
rs760288459 Chr.9: 19056416-19056416 on Build GRCh38
rs778203483 Chr.9: 19056247-19056247 on Build GRCh38
rs529319617 Chr.9: 19056397-19056397 on Build GRCh38
rs993433967 Chr.9: 19056535-19056535 on Build GRCh38
rs750883539 Chr.9: 19056435-19056435 on Build GRCh38
rs186429271 Chr.9: 19056484-19056484 on Build GRCh38
rs749487544 Chr.9: 19056399-19056399 on Build GRCh38
rs140286860 Chr.9: 19056387-19056387 on Build GRCh38
rs961957451 Chr.9: 19056321-19056321 on Build GRCh38
rs372584901 Chr.9: 19056426-19056426 on Build GRCh38
rs954556233 Chr.9: 19056552-19056552 on Build GRCh38
rs780693662 Chr.9: 19056322-19056322 on Build GRCh38
rs768139920 Chr.9: 19056431-19056431 on Build GRCh38
rs769996829 Chr.9: 19056305-19056305 on Build GRCh38
rs897634663 Chr.9: 19056534-19056536 on Build GRCh38
rs142212509 Chr.9: 19056325-19056325 on Build GRCh38
rs142907947 Chr.9: 19056367-19056367 on Build GRCh38
rs749119861 Chr.9: 19056449-19056452 on Build GRCh38
rs775265277 Chr.9: 19056313-19056313 on Build GRCh38
rs139954288 Chr.9: 19056279-19056279 on Build GRCh38
rs796606479 Chr.9: 19056520-19056523 on Build GRCh38
rs773017843 Chr.9: 19056415-19056415 on Build GRCh38
rs374328391 Chr.9: 19056333-19056333 on Build GRCh38
rs112485743 Chr.9: 19056538-19056538 on Build GRCh38
rs151203048 Chr.9: 19056376-19056376 on Build GRCh38
rs377100314 Chr.9: 19056429-19056429 on Build GRCh38
rs779658653 Chr.9: 19056441-19056441 on Build GRCh38
rs62563589 Chr.9: 19056501-19056501 on Build GRCh38
rs550364315 Chr.9: 19056420-19056420 on Build GRCh38
rs573767459 Chr.9: 19056194-19056194 on Build GRCh38
rs780077552 Chr.9: 19056444-19056444 on Build GRCh38
rs752147454 Chr.9: 19056366-19056366 on Build GRCh38
rs372844925 Chr.9: 19056357-19056357 on Build GRCh38
rs371418229 Chr.9: 19056412-19056412 on Build GRCh38
rs888536604 Chr.9: 19056500-19056500 on Build GRCh38
rs115504877 Chr.9: 19056508-19056508 on Build GRCh38
rs369306335 Chr.9: 19056409-19056409 on Build GRCh38
rs557564656 Chr.9: 19056252-19056252 on Build GRCh38
rs1017813062 Chr.9: 19056559-19056559 on Build GRCh38
rs989663907 Chr.9: 19056419-19056420 on Build GRCh38
rs985807978 Chr.9: 19056558-19056558 on Build GRCh38
rs749146311 Chr.9: 19056319-19056319 on Build GRCh38
rs201056474 Chr.9: 19056400-19056400 on Build GRCh38
rs989158637 Chr.9: 19056573-19056573 on Build GRCh38
rs1030236077 Chr.9: 19056544-19056544 on Build GRCh38

View Full Product Details