Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19408776-19409279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACER2
Entrez Gene ID
340485
Gene Name
alkaline ceramidase 2
Gene Aliases
ALKCDase2, ASAH3L
UniGene
Hs.41379
Gene Chromosome Location
Chr.9: 19408927-19455173 on Build GRCh38
Transcripts
NM_001010887.2 XM_005251448.3 XM_011517858.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3942991 c.11C>T p.P4L Chr.9: 19409095-19409095 on Build GRCh38
7353942 c.41G>C p.S14T Chr.9: 19409125-19409125 on Build GRCh38
4515165 c.9C>T p.A3A Chr.9: 19409093-19409093 on Build GRCh38
4163685 c.21G>T p.W7C Chr.9: 19409105-19409105 on Build GRCh38
3395732 c.108+8G>T p.? Chr.9: 19409200-19409200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755282852 Chr.9: 19409170-19409170 on Build GRCh38
rs547339284 Chr.9: 19409008-19409008 on Build GRCh38
rs759360813 Chr.9: 19409134-19409134 on Build GRCh38
rs751129108 Chr.9: 19409073-19409073 on Build GRCh38
rs974091000 Chr.9: 19408921-19408921 on Build GRCh38
rs780328845 Chr.9: 19409089-19409089 on Build GRCh38
rs916605749 Chr.9: 19408801-19408801 on Build GRCh38
rs921714672 Chr.9: 19408807-19408807 on Build GRCh38
rs144513346 Chr.9: 19409096-19409096 on Build GRCh38
rs1036286187 Chr.9: 19408843-19408843 on Build GRCh38
rs992768458 Chr.9: 19408821-19408821 on Build GRCh38
rs377429359 Chr.9: 19408988-19408988 on Build GRCh38
rs573879549 Chr.9: 19408827-19408827 on Build GRCh38
rs1012754673 Chr.9: 19409044-19409044 on Build GRCh38
rs767197220 Chr.9: 19409157-19409157 on Build GRCh38
rs895675122 Chr.9: 19409024-19409024 on Build GRCh38
rs1055744119 Chr.9: 19409006-19409006 on Build GRCh38
rs577445038 Chr.9: 19408909-19408910 on Build GRCh38
rs991405864 Chr.9: 19408911-19408911 on Build GRCh38
rs538774152 Chr.9: 19409138-19409138 on Build GRCh38
rs543593006 Chr.9: 19409221-19409221 on Build GRCh38
rs930164714 Chr.9: 19408839-19408839 on Build GRCh38
rs576417429 Chr.9: 19409212-19409212 on Build GRCh38
rs556236139 Chr.9: 19408884-19408884 on Build GRCh38
rs771164024 Chr.9: 19409125-19409125 on Build GRCh38
rs1005220697 Chr.9: 19408837-19408837 on Build GRCh38
rs947646593 Chr.9: 19409167-19409167 on Build GRCh38
rs747105650 Chr.9: 19409199-19409199 on Build GRCh38
rs899173813 Chr.9: 19408854-19408854 on Build GRCh38
rs113245552 Chr.9: 19409087-19409087 on Build GRCh38
rs944267534 Chr.9: 19408954-19408954 on Build GRCh38
rs931822562 Chr.9: 19408819-19408819 on Build GRCh38
rs911721722 Chr.9: 19408947-19408947 on Build GRCh38
rs550044724 Chr.9: 19408972-19408972 on Build GRCh38
rs765135971 Chr.9: 19408926-19408926 on Build GRCh38
rs1024909869 Chr.9: 19408895-19408895 on Build GRCh38
rs565095485 Chr.9: 19409242-19409242 on Build GRCh38
rs549242757 Chr.9: 19409041-19409041 on Build GRCh38
rs10122075 Chr.9: 19409109-19409109 on Build GRCh38
rs903349476 Chr.9: 19409106-19409106 on Build GRCh38
rs560309171 Chr.9: 19408903-19408903 on Build GRCh38
rs940578964 Chr.9: 19408950-19408950 on Build GRCh38
rs1033250668 Chr.9: 19409128-19409128 on Build GRCh38
rs182872766 Chr.9: 19408828-19408828 on Build GRCh38
rs758496444 Chr.9: 19409191-19409191 on Build GRCh38
rs142611269 Chr.9: 19409162-19409162 on Build GRCh38
rs761794922 Chr.9: 19408914-19408914 on Build GRCh38
rs187863827 Chr.9: 19408935-19408935 on Build GRCh38
rs1039367826 Chr.9: 19408874-19408874 on Build GRCh38
rs532197555 Chr.9: 19409247-19409247 on Build GRCh38
rs536277643 Chr.9: 19409040-19409040 on Build GRCh38
rs748661462 Chr.9: 19408996-19408996 on Build GRCh38
rs531534022 Chr.9: 19408946-19408946 on Build GRCh38
rs957633533 Chr.9: 19409137-19409137 on Build GRCh38
rs767723984 Chr.9: 19409086-19409086 on Build GRCh38
rs931743037 Chr.9: 19408941-19408941 on Build GRCh38
rs760980206 Chr.9: 19409115-19409115 on Build GRCh38
rs755548497 Chr.9: 19409097-19409097 on Build GRCh38
rs773515513 Chr.9: 19409114-19409114 on Build GRCh38
rs779382872 Chr.9: 19409102-19409102 on Build GRCh38
rs763644748 Chr.9: 19409148-19409148 on Build GRCh38
rs987292997 Chr.9: 19408944-19408944 on Build GRCh38
rs112136424 Chr.9: 19408938-19408938 on Build GRCh38
rs769611020 Chr.9: 19409178-19409182 on Build GRCh38
rs561197497 Chr.9: 19408940-19408940 on Build GRCh38
rs779397326 Chr.9: 19409185-19409185 on Build GRCh38
rs532542185 Chr.9: 19408971-19408971 on Build GRCh38
rs994836148 Chr.9: 19408863-19408863 on Build GRCh38
rs747152832 Chr.9: 19409120-19409120 on Build GRCh38
rs891388576 Chr.9: 19408872-19408872 on Build GRCh38
rs1042710824 Chr.9: 19409169-19409169 on Build GRCh38
rs775501749 Chr.9: 19409145-19409145 on Build GRCh38
rs762579917 Chr.9: 19409147-19409147 on Build GRCh38
rs201611502 Chr.9: 19409201-19409201 on Build GRCh38
rs757755326 Chr.9: 19409248-19409248 on Build GRCh38
rs866543372 Chr.9: 19408967-19408967 on Build GRCh38
rs538796930 Chr.9: 19408866-19408867 on Build GRCh38
rs967624782 Chr.9: 19408896-19408896 on Build GRCh38
rs920896371 Chr.9: 19408982-19408982 on Build GRCh38
rs748615867 Chr.9: 19409188-19409188 on Build GRCh38
rs1040011933 Chr.9: 19408959-19408959 on Build GRCh38
rs759095859 Chr.9: 19409046-19409057 on Build GRCh38
rs745662574 Chr.9: 19409209-19409209 on Build GRCh38
rs1034387894 Chr.9: 19408878-19408878 on Build GRCh38
rs969339144 Chr.9: 19409063-19409063 on Build GRCh38
rs567666035 Chr.9: 19409072-19409072 on Build GRCh38
rs11792865 Chr.9: 19409208-19409208 on Build GRCh38
rs899537911 Chr.9: 19408883-19408883 on Build GRCh38
rs565552974 Chr.9: 19409011-19409011 on Build GRCh38
rs894471694 Chr.9: 19409243-19409243 on Build GRCh38
rs144402037 Chr.9: 19409005-19409005 on Build GRCh38
rs894534946 Chr.9: 19409007-19409007 on Build GRCh38
rs115574837 Chr.9: 19408899-19408899 on Build GRCh38
rs564929964 Chr.9: 19408989-19408989 on Build GRCh38
rs769668275 Chr.9: 19409210-19409210 on Build GRCh38
rs983309624 Chr.9: 19409104-19409104 on Build GRCh38
rs201941464 Chr.9: 19409075-19409075 on Build GRCh38
rs572271466 Chr.9: 19408918-19408918 on Build GRCh38
rs899121374 Chr.9: 19409001-19409001 on Build GRCh38
rs757923845 Chr.9: 19409055-19409055 on Build GRCh38
rs764542576 Chr.9: 19409152-19409152 on Build GRCh38
rs554276695 Chr.9: 19409156-19409156 on Build GRCh38
rs138659556 Chr.9: 19408873-19408874 on Build GRCh38
rs777766936 Chr.9: 19409192-19409192 on Build GRCh38
rs577725338 Chr.9: 19408886-19408886 on Build GRCh38

View Full Product Details