Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19451703-19452204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACER2
Entrez Gene ID
340485
Gene Name
alkaline ceramidase 2
Gene Aliases
ALKCDase2, ASAH3L
UniGene
Hs.41379
Gene Chromosome Location
Chr.9: 19408927-19455173 on Build GRCh38
Transcripts
NM_001010887.2 XM_005251448.3 XM_011517859.2 XM_017014694.1

SNP Detail

SNP ID Chromosome Location
rs1052772532 Chr.9: 19452070-19452070 on Build GRCh38
rs1041671817 Chr.9: 19451814-19451814 on Build GRCh38
rs1049847963 Chr.9: 19452054-19452054 on Build GRCh38
rs55751242 Chr.9: 19451952-19451953 on Build GRCh38
rs547398049 Chr.9: 19451734-19451734 on Build GRCh38
rs777434555 Chr.9: 19452120-19452120 on Build GRCh38
rs1014466218 Chr.9: 19452182-19452182 on Build GRCh38
rs564864322 Chr.9: 19451729-19451729 on Build GRCh38
rs1031446035 Chr.9: 19451909-19451909 on Build GRCh38
rs999928576 Chr.9: 19451905-19451905 on Build GRCh38
rs202212878 Chr.9: 19451955-19451955 on Build GRCh38
rs554170293 Chr.9: 19451888-19451888 on Build GRCh38
rs1014099782 Chr.9: 19451940-19451940 on Build GRCh38
rs970004586 Chr.9: 19451947-19451947 on Build GRCh38
rs940143703 Chr.9: 19451996-19451996 on Build GRCh38
rs112164706 Chr.9: 19451951-19451952 on Build GRCh38
rs538632380 Chr.9: 19451841-19451841 on Build GRCh38
rs955847850 Chr.9: 19451920-19451920 on Build GRCh38
rs770252614 Chr.9: 19451792-19451792 on Build GRCh38
rs1024610779 Chr.9: 19451946-19451946 on Build GRCh38
rs550218500 Chr.9: 19451981-19451981 on Build GRCh38
rs996545554 Chr.9: 19451727-19451727 on Build GRCh38
rs752010810 Chr.9: 19451896-19451896 on Build GRCh38
rs374175884 Chr.9: 19451748-19451748 on Build GRCh38
rs932952175 Chr.9: 19452049-19452049 on Build GRCh38
rs554296522 Chr.9: 19451980-19451980 on Build GRCh38
rs759084396 Chr.9: 19451970-19451970 on Build GRCh38
rs17565631 Chr.9: 19451953-19451953 on Build GRCh38
rs767075390 Chr.9: 19452050-19452050 on Build GRCh38
rs931426466 Chr.9: 19451797-19451797 on Build GRCh38
rs150022470 Chr.9: 19451731-19451731 on Build GRCh38
rs955941444 Chr.9: 19451754-19451754 on Build GRCh38
rs12343276 Chr.9: 19451809-19451809 on Build GRCh38
rs1041219014 Chr.9: 19452007-19452007 on Build GRCh38
rs191846430 Chr.9: 19452061-19452061 on Build GRCh38
rs996875416 Chr.9: 19451853-19451853 on Build GRCh38
rs755433629 Chr.9: 19452098-19452098 on Build GRCh38
rs867265231 Chr.9: 19451895-19451895 on Build GRCh38
rs752328743 Chr.9: 19452055-19452055 on Build GRCh38
rs901432102 Chr.9: 19451843-19451843 on Build GRCh38
rs187689495 Chr.9: 19451773-19451773 on Build GRCh38
rs1004478424 Chr.9: 19452166-19452166 on Build GRCh38
rs7032360 Chr.9: 19451789-19451789 on Build GRCh38
rs926663013 Chr.9: 19451795-19451795 on Build GRCh38
rs571586278 Chr.9: 19451838-19451838 on Build GRCh38
rs528034517 Chr.9: 19451784-19451784 on Build GRCh38
rs371633826 Chr.9: 19451995-19451995 on Build GRCh38
rs949383203 Chr.9: 19451790-19451790 on Build GRCh38
rs908647096 Chr.9: 19451969-19451969 on Build GRCh38
rs891553351 Chr.9: 19452136-19452136 on Build GRCh38
rs564807335 Chr.9: 19452111-19452111 on Build GRCh38
rs567865371 Chr.9: 19451800-19451800 on Build GRCh38
rs543086924 Chr.9: 19452079-19452079 on Build GRCh38
rs374126446 Chr.9: 19452010-19452010 on Build GRCh38
rs1049445224 Chr.9: 19451884-19451884 on Build GRCh38
rs756613169 Chr.9: 19451953-19451954 on Build GRCh38
rs766847726 Chr.9: 19451817-19451817 on Build GRCh38
rs532268513 Chr.9: 19452164-19452164 on Build GRCh38
rs41270117 Chr.9: 19451811-19451811 on Build GRCh38
rs1031500044 Chr.9: 19451738-19451740 on Build GRCh38
rs373060253 Chr.9: 19451953-19451953 on Build GRCh38

View Full Product Details