Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33030303-33030846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA1
Entrez Gene ID
3301
Gene Name
DnaJ heat shock protein family (Hsp40) member A1
Gene Aliases
DJ-2, DjA1, HDJ2, HSDJ, HSJ-2, HSJ2, HSPF4, NEDD7, hDJ-2
UniGene
Hs.445203
Gene Chromosome Location
Chr.9: 33025203-33039907 on Build GRCh38
Transcripts
NM_001314039.1 NM_001539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7174627 c.575G>T p.R192I Chr.9: 33030599-33030599 on Build GRCh38
252477 c.427A>G p.K143E Chr.9: 33030451-33030451 on Build GRCh38
1108284 c.460C>T p.R154* Chr.9: 33030484-33030484 on Build GRCh38
3664303 c.643+10G>C p.? Chr.9: 33030677-33030677 on Build GRCh38
6987916 c.592G>T p.G198* Chr.9: 33030616-33030616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040089082 Chr.9: 33030808-33030808 on Build GRCh38
rs768358364 Chr.9: 33030701-33030701 on Build GRCh38
rs769775792 Chr.9: 33030686-33030686 on Build GRCh38
rs141054448 Chr.9: 33030816-33030816 on Build GRCh38
rs752728344 Chr.9: 33030327-33030327 on Build GRCh38
rs920430078 Chr.9: 33030628-33030628 on Build GRCh38
rs146416874 Chr.9: 33030580-33030580 on Build GRCh38
rs567909525 Chr.9: 33030328-33030328 on Build GRCh38
rs368602920 Chr.9: 33030573-33030573 on Build GRCh38
rs762414764 Chr.9: 33030413-33030413 on Build GRCh38
rs781323878 Chr.9: 33030679-33030679 on Build GRCh38
rs571401003 Chr.9: 33030755-33030755 on Build GRCh38
rs906205217 Chr.9: 33030704-33030704 on Build GRCh38
rs191263229 Chr.9: 33030354-33030354 on Build GRCh38
rs754180020 Chr.9: 33030597-33030597 on Build GRCh38
rs1055196805 Chr.9: 33030773-33030773 on Build GRCh38
rs758663767 Chr.9: 33030626-33030626 on Build GRCh38
rs765016656 Chr.9: 33030409-33030409 on Build GRCh38
rs748332900 Chr.9: 33030412-33030412 on Build GRCh38
rs768443026 Chr.9: 33030775-33030775 on Build GRCh38
rs752922535 Chr.9: 33030624-33030624 on Build GRCh38
rs769545051 Chr.9: 33030546-33030546 on Build GRCh38
rs766840420 Chr.9: 33030716-33030716 on Build GRCh38
rs999678458 Chr.9: 33030329-33030329 on Build GRCh38
rs776582793 Chr.9: 33030404-33030404 on Build GRCh38
rs961948955 Chr.9: 33030571-33030571 on Build GRCh38
rs779155858 Chr.9: 33030465-33030465 on Build GRCh38
rs78775007 Chr.9: 33030778-33030778 on Build GRCh38
rs745676787 Chr.9: 33030502-33030502 on Build GRCh38
rs201027662 Chr.9: 33030638-33030638 on Build GRCh38
rs969414946 Chr.9: 33030370-33030370 on Build GRCh38
rs959533358 Chr.9: 33030343-33030343 on Build GRCh38
rs746866732 Chr.9: 33030484-33030484 on Build GRCh38
rs773922554 Chr.9: 33030581-33030581 on Build GRCh38
rs776531290 Chr.9: 33030492-33030492 on Build GRCh38
rs374784271 Chr.9: 33030414-33030414 on Build GRCh38
rs374236433 Chr.9: 33030687-33030687 on Build GRCh38
rs112587848 Chr.9: 33030669-33030669 on Build GRCh38
rs893846857 Chr.9: 33030348-33030348 on Build GRCh38
rs749208836 Chr.9: 33030712-33030712 on Build GRCh38
rs752202634 Chr.9: 33030412-33030412 on Build GRCh38
rs748833230 Chr.9: 33030689-33030689 on Build GRCh38
rs756350679 Chr.9: 33030644-33030644 on Build GRCh38
rs770691518 Chr.9: 33030402-33030402 on Build GRCh38
rs369149295 Chr.9: 33030436-33030436 on Build GRCh38
rs201185136 Chr.9: 33030625-33030625 on Build GRCh38
rs935574676 Chr.9: 33030816-33030816 on Build GRCh38
rs985889012 Chr.9: 33030733-33030733 on Build GRCh38
rs768035452 Chr.9: 33030422-33030422 on Build GRCh38
rs764237868 Chr.9: 33030715-33030717 on Build GRCh38
rs1031195534 Chr.9: 33030331-33030331 on Build GRCh38
rs530024543 Chr.9: 33030353-33030353 on Build GRCh38
rs758475036 Chr.9: 33030471-33030471 on Build GRCh38
rs750955525 Chr.9: 33030429-33030429 on Build GRCh38
rs553707415 Chr.9: 33030390-33030390 on Build GRCh38
rs112441039 Chr.9: 33030793-33030793 on Build GRCh38
rs922828653 Chr.9: 33030812-33030812 on Build GRCh38
rs369090653 Chr.9: 33030579-33030579 on Build GRCh38
rs764436093 Chr.9: 33030744-33030744 on Build GRCh38
rs763191340 Chr.9: 33030418-33030421 on Build GRCh38
rs982102335 Chr.9: 33030398-33030398 on Build GRCh38
rs761020508 Chr.9: 33030587-33030587 on Build GRCh38
rs757216481 Chr.9: 33030676-33030676 on Build GRCh38
rs761334340 Chr.9: 33030713-33030713 on Build GRCh38
rs376930856 Chr.9: 33030528-33030528 on Build GRCh38
rs200424525 Chr.9: 33030629-33030629 on Build GRCh38
rs755376906 Chr.9: 33030615-33030615 on Build GRCh38
rs112314263 Chr.9: 33030670-33030670 on Build GRCh38
rs766901754 Chr.9: 33030589-33030589 on Build GRCh38
rs1033572518 Chr.9: 33030337-33030337 on Build GRCh38
rs139108229 Chr.9: 33030616-33030616 on Build GRCh38
rs780336128 Chr.9: 33030432-33030432 on Build GRCh38
rs748229438 Chr.9: 33030468-33030468 on Build GRCh38
rs202246880 Chr.9: 33030635-33030635 on Build GRCh38
rs759202732 Chr.9: 33030407-33030407 on Build GRCh38
rs747148431 Chr.9: 33030665-33030665 on Build GRCh38
rs977146872 Chr.9: 33030448-33030448 on Build GRCh38
rs754206372 Chr.9: 33030433-33030433 on Build GRCh38
rs771000953 Chr.9: 33030485-33030485 on Build GRCh38
rs566601668 Chr.9: 33030381-33030381 on Build GRCh38
rs910015688 Chr.9: 33030631-33030631 on Build GRCh38
rs372998530 Chr.9: 33030475-33030475 on Build GRCh38
rs968783130 Chr.9: 33030359-33030359 on Build GRCh38
rs777650491 Chr.9: 33030657-33030657 on Build GRCh38
rs57062539 Chr.9: 33030746-33030746 on Build GRCh38
rs1024175595 Chr.9: 33030366-33030366 on Build GRCh38
rs745623646 Chr.9: 33030683-33030683 on Build GRCh38

View Full Product Details