Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33038585-33039083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA1
Entrez Gene ID
3301
Gene Name
DnaJ heat shock protein family (Hsp40) member A1
Gene Aliases
DJ-2, DjA1, HDJ2, HSDJ, HSJ-2, HSJ2, HSPF4, NEDD7, hDJ-2
UniGene
Hs.445203
Gene Chromosome Location
Chr.9: 33025203-33039907 on Build GRCh38
Transcripts
NM_001314039.1 NM_001539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232194 c.985C>T p.P329S Chr.9: 33038694-33038694 on Build GRCh38
3906869 c.1063A>G p.T355A Chr.9: 33038772-33038772 on Build GRCh38
5177912 c.1173_1174insGTTCAGTG p.C394fs*>7 Chr.9: 33038882-33038883 on Build GRCh38
5507159 c.1051G>A p.E351K Chr.9: 33038760-33038760 on Build GRCh38
3906870 c.1127A>G p.Y376C Chr.9: 33038836-33038836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748280590 Chr.9: 33038868-33038868 on Build GRCh38
rs756513491 Chr.9: 33038827-33038827 on Build GRCh38
rs935817109 Chr.9: 33038628-33038628 on Build GRCh38
rs761015443 Chr.9: 33038974-33038974 on Build GRCh38
rs766703382 Chr.9: 33038796-33038797 on Build GRCh38
rs568893066 Chr.9: 33038865-33038865 on Build GRCh38
rs781143779 Chr.9: 33038912-33038912 on Build GRCh38
rs911925877 Chr.9: 33038630-33038630 on Build GRCh38
rs760362507 Chr.9: 33038655-33038655 on Build GRCh38
rs758245297 Chr.9: 33038872-33038872 on Build GRCh38
rs766672756 Chr.9: 33038975-33038975 on Build GRCh38
rs775074991 Chr.9: 33038796-33038796 on Build GRCh38
rs181836256 Chr.9: 33038631-33038631 on Build GRCh38
rs141132012 Chr.9: 33038829-33038829 on Build GRCh38
rs780305363 Chr.9: 33038706-33038706 on Build GRCh38
rs765543456 Chr.9: 33038982-33038982 on Build GRCh38
rs773775893 Chr.9: 33038958-33038958 on Build GRCh38
rs760589124 Chr.9: 33038679-33038679 on Build GRCh38
rs777895077 Chr.9: 33038885-33038885 on Build GRCh38
rs1004514057 Chr.9: 33038936-33038936 on Build GRCh38
rs7296 Chr.9: 33039026-33039026 on Build GRCh38
rs925938396 Chr.9: 33038660-33038660 on Build GRCh38
rs770904664 Chr.9: 33038904-33038904 on Build GRCh38
rs768094883 Chr.9: 33038800-33038800 on Build GRCh38
rs766150865 Chr.9: 33038663-33038663 on Build GRCh38
rs746793842 Chr.9: 33038894-33038894 on Build GRCh38
rs987675090 Chr.9: 33038620-33038620 on Build GRCh38
rs868488974 Chr.9: 33038786-33038786 on Build GRCh38
rs774816290 Chr.9: 33038922-33038922 on Build GRCh38
rs990381230 Chr.9: 33039024-33039024 on Build GRCh38
rs776768067 Chr.9: 33038909-33038909 on Build GRCh38
rs757800298 Chr.9: 33038693-33038693 on Build GRCh38
rs756571976 Chr.9: 33038673-33038673 on Build GRCh38
rs759099963 Chr.9: 33038789-33038789 on Build GRCh38
rs758568855 Chr.9: 33038995-33038995 on Build GRCh38
rs771821386 Chr.9: 33038640-33038640 on Build GRCh38
rs776256677 Chr.9: 33038669-33038669 on Build GRCh38
rs867748833 Chr.9: 33038751-33038751 on Build GRCh38
rs767710743 Chr.9: 33038697-33038697 on Build GRCh38
rs1041824142 Chr.9: 33038645-33038645 on Build GRCh38
rs755505219 Chr.9: 33038965-33038966 on Build GRCh38
rs745449469 Chr.9: 33038914-33038914 on Build GRCh38
rs1039624400 Chr.9: 33038765-33038765 on Build GRCh38
rs750767685 Chr.9: 33038823-33038823 on Build GRCh38
rs374710198 Chr.9: 33038830-33038830 on Build GRCh38
rs754394456 Chr.9: 33038863-33038865 on Build GRCh38
rs114496815 Chr.9: 33039043-33039043 on Build GRCh38
rs756282667 Chr.9: 33038705-33038705 on Build GRCh38
rs778818641 Chr.9: 33038861-33038861 on Build GRCh38
rs752685836 Chr.9: 33038985-33038985 on Build GRCh38
rs11542818 Chr.9: 33039017-33039017 on Build GRCh38
rs762575235 Chr.9: 33038925-33038925 on Build GRCh38
rs759112229 Chr.9: 33039006-33039006 on Build GRCh38
rs759627222 Chr.9: 33038978-33038978 on Build GRCh38
rs769521508 Chr.9: 33038921-33038921 on Build GRCh38
rs776490065 Chr.9: 33038780-33038780 on Build GRCh38
rs754232780 Chr.9: 33038977-33038977 on Build GRCh38
rs368226813 Chr.9: 33038838-33038838 on Build GRCh38
rs773063526 Chr.9: 33038647-33038647 on Build GRCh38
rs749336661 Chr.9: 33038712-33038712 on Build GRCh38
rs1803513 Chr.9: 33038884-33038884 on Build GRCh38
rs772077578 Chr.9: 33038758-33038758 on Build GRCh38
rs766399128 Chr.9: 33038700-33038700 on Build GRCh38
rs74968935 Chr.9: 33038674-33038674 on Build GRCh38
rs778896977 Chr.9: 33038750-33038750 on Build GRCh38
rs773091099 Chr.9: 33038761-33038761 on Build GRCh38
rs945405184 Chr.9: 33039036-33039036 on Build GRCh38
rs746848794 Chr.9: 33038772-33038772 on Build GRCh38
rs750594107 Chr.9: 33038663-33038664 on Build GRCh38
rs185238499 Chr.9: 33038837-33038837 on Build GRCh38
rs374511744 Chr.9: 33038998-33039002 on Build GRCh38
rs148382351 Chr.9: 33038756-33038756 on Build GRCh38
rs772428402 Chr.9: 33038949-33038949 on Build GRCh38
rs367858309 Chr.9: 33038787-33038787 on Build GRCh38
rs752060167 Chr.9: 33038687-33038687 on Build GRCh38
rs747015712 Chr.9: 33038919-33038919 on Build GRCh38
rs890716045 Chr.9: 33038747-33038747 on Build GRCh38
rs770673934 Chr.9: 33038776-33038776 on Build GRCh38
rs755081187 Chr.9: 33038718-33038718 on Build GRCh38

View Full Product Details