Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33110946-33111446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT1
Entrez Gene ID
2683
Gene Name
beta-1,4-galactosyltransferase 1
Gene Aliases
B4GAL-T1, CDG2D, GGTB2, GT1, GTB, beta4Gal-T1
UniGene
Hs.272011
Gene Chromosome Location
Chr.9: 33110641-33167391 on Build GRCh38
Transcripts
NM_001497.3 XM_005251440.4

SNP Detail

SNP ID Chromosome Location
rs774394805 Chr.9: 33111143-33111143 on Build GRCh38
rs147768292 Chr.9: 33111279-33111282 on Build GRCh38
rs75201363 Chr.9: 33111013-33111013 on Build GRCh38
rs920180628 Chr.9: 33110995-33110995 on Build GRCh38
rs895763052 Chr.9: 33111329-33111330 on Build GRCh38
rs184203621 Chr.9: 33111375-33111375 on Build GRCh38
rs978231896 Chr.9: 33111392-33111392 on Build GRCh38
rs989926711 Chr.9: 33111228-33111228 on Build GRCh38
rs994915693 Chr.9: 33111154-33111154 on Build GRCh38
rs1064950 Chr.9: 33111275-33111275 on Build GRCh38
rs886063862 Chr.9: 33111162-33111162 on Build GRCh38
rs533627511 Chr.9: 33111125-33111125 on Build GRCh38
rs78409522 Chr.9: 33111413-33111413 on Build GRCh38
rs993156099 Chr.9: 33111119-33111119 on Build GRCh38
rs1019836635 Chr.9: 33111384-33111384 on Build GRCh38
rs7041783 Chr.9: 33111248-33111248 on Build GRCh38
rs1046328462 Chr.9: 33111263-33111263 on Build GRCh38
rs749146295 Chr.9: 33111107-33111107 on Build GRCh38
rs558569071 Chr.9: 33111035-33111035 on Build GRCh38
rs770356020 Chr.9: 33111277-33111285 on Build GRCh38
rs889269349 Chr.9: 33111262-33111262 on Build GRCh38
rs45480798 Chr.9: 33111276-33111276 on Build GRCh38
rs746956537 Chr.9: 33111282-33111282 on Build GRCh38
rs898800578 Chr.9: 33111146-33111146 on Build GRCh38
rs769799377 Chr.9: 33111279-33111284 on Build GRCh38
rs796930062 Chr.9: 33111271-33111279 on Build GRCh38
rs796346335 Chr.9: 33111266-33111266 on Build GRCh38
rs527240435 Chr.9: 33111410-33111410 on Build GRCh38
rs974295364 Chr.9: 33110993-33110993 on Build GRCh38
rs1052387273 Chr.9: 33111237-33111237 on Build GRCh38
rs925332395 Chr.9: 33111203-33111203 on Build GRCh38
rs759484311 Chr.9: 33111285-33111285 on Build GRCh38
rs1029102981 Chr.9: 33111278-33111278 on Build GRCh38
rs886063861 Chr.9: 33111056-33111056 on Build GRCh38
rs959950114 Chr.9: 33111150-33111150 on Build GRCh38
rs531869620 Chr.9: 33111144-33111145 on Build GRCh38
rs886063864 Chr.9: 33111278-33111282 on Build GRCh38
rs906338622 Chr.9: 33111270-33111270 on Build GRCh38
rs754771244 Chr.9: 33111277-33111279 on Build GRCh38
rs965484177 Chr.9: 33111388-33111388 on Build GRCh38
rs7041909 Chr.9: 33111279-33111279 on Build GRCh38
rs140793406 Chr.9: 33111049-33111050 on Build GRCh38
rs770013565 Chr.9: 33111001-33111001 on Build GRCh38
rs1014697177 Chr.9: 33111133-33111133 on Build GRCh38
rs963061253 Chr.9: 33110990-33110990 on Build GRCh38
rs74995636 Chr.9: 33111201-33111201 on Build GRCh38
rs111345260 Chr.9: 33111277-33111277 on Build GRCh38
rs1053134036 Chr.9: 33111002-33111002 on Build GRCh38
rs978042680 Chr.9: 33111034-33111034 on Build GRCh38
rs374364990 Chr.9: 33111189-33111189 on Build GRCh38
rs576341394 Chr.9: 33111206-33111206 on Build GRCh38
rs985632600 Chr.9: 33111161-33111161 on Build GRCh38
rs936635397 Chr.9: 33111220-33111220 on Build GRCh38
rs561769709 Chr.9: 33111321-33111321 on Build GRCh38
rs745322216 Chr.9: 33111183-33111184 on Build GRCh38
rs761196389 Chr.9: 33111155-33111155 on Build GRCh38
rs537799576 Chr.9: 33111069-33111069 on Build GRCh38
rs978550562 Chr.9: 33111408-33111410 on Build GRCh38
rs16918997 Chr.9: 33111350-33111350 on Build GRCh38
rs116375484 Chr.9: 33111173-33111173 on Build GRCh38
rs775197030 Chr.9: 33111090-33111090 on Build GRCh38
rs374921974 Chr.9: 33111279-33111279 on Build GRCh38
rs895057619 Chr.9: 33111087-33111087 on Build GRCh38
rs74772022 Chr.9: 33111185-33111185 on Build GRCh38
rs898849257 Chr.9: 33111103-33111103 on Build GRCh38
rs947905109 Chr.9: 33111089-33111089 on Build GRCh38
rs367581361 Chr.9: 33111100-33111100 on Build GRCh38
rs1037432896 Chr.9: 33111264-33111264 on Build GRCh38
rs184223660 Chr.9: 33110996-33110996 on Build GRCh38
rs944153101 Chr.9: 33111014-33111014 on Build GRCh38
rs886063863 Chr.9: 33111251-33111254 on Build GRCh38
rs751351046 Chr.9: 33111251-33111265 on Build GRCh38
rs564649137 Chr.9: 33111386-33111386 on Build GRCh38
rs553864348 Chr.9: 33111159-33111159 on Build GRCh38
rs1025972509 Chr.9: 33111307-33111307 on Build GRCh38
rs35504729 Chr.9: 33111251-33111251 on Build GRCh38
rs34791389 Chr.9: 33111010-33111011 on Build GRCh38
rs768566333 Chr.9: 33111129-33111129 on Build GRCh38
rs532160095 Chr.9: 33111387-33111387 on Build GRCh38
rs368519327 Chr.9: 33111216-33111216 on Build GRCh38
rs13289751 Chr.9: 33111269-33111269 on Build GRCh38
rs947989541 Chr.9: 33111233-33111233 on Build GRCh38
rs954474360 Chr.9: 33111158-33111158 on Build GRCh38
rs192108272 Chr.9: 33111075-33111075 on Build GRCh38
rs574554324 Chr.9: 33111099-33111099 on Build GRCh38
rs767384487 Chr.9: 33111420-33111420 on Build GRCh38
rs371831330 Chr.9: 33111192-33111192 on Build GRCh38
rs188764568 Chr.9: 33111052-33111052 on Build GRCh38
rs749777226 Chr.9: 33110983-33110983 on Build GRCh38
rs541787590 Chr.9: 33111232-33111232 on Build GRCh38
rs763903279 Chr.9: 33111273-33111273 on Build GRCh38

View Full Product Details